Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Peptide sequence: EHSPLERDSL MPSDEASESS RQAWTGSSQR SSRHLEEDYA DAYQDLYQPH
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
CACNB3 encodes a regulatory beta subunit of the voltage-dependent calcium channel. Beta subunits are composed of five domains, which contribute to the regulation of surface expression and gating of calcium channels and may also play a role in the regulation of transcription factors and calcium transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAB3; Calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; Cav; Voltage-dependent L-type calcium channel subunit beta-3
Gene Aliases: CAB3; CACNB3; CACNLB3
UniProt ID: (Human) P54284
Entrez Gene ID: (Human) 784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support