Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSSSYAKN GTADGPHSPT SQVARGTTTR RSRLKRSDGS TTSTSFILRQ GSADSYTSRP SDSDVSLEED REAIRQEREQ QAAIQLERAK SKPVAFAVKT NVSYCGALDE DVPVPSTAIS FDAKDFLHIK EKYNNDWWIG RLVKEGCEIG FIPSPLRLEN IRIQQEQKRG RFHGGKSSGN SSSSLGEMVS GTFRATPTST AKQKQKVTEH IPPYDVVPSM RPVVLVGPSL KGYEVTDMMQ KALFDFLKHR FDGRISITRV TADISLAKRS VLNNPSKRAI IERSNTRSSL AEVQSEIERI FELARSLQLV VLDADTINHP AQLIKTSLAP IIVHVKVSSP KVLQRLIKSR G (1-350 aa encoded by BC075049)
This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy and juvenile myoclonic epilepsy. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAB4; calcium channel beta 4 subunit; Calcium channel voltage-dependent subunit beta 4; calcium channel, voltage-dependent, beta 4 subunit; dihydropyridine-sensitive L-type, calcium channel beta-4 subunit; OTTHUMP00000207247; OTTHUMP00000207249; Voltage-dependent L-type calcium channel subunit beta-4
Gene Aliases: 3110038O15Rik; CAB4; CACNB4; CACNLB4; Cchb4; EA5; EIG9; EJM; EJM4; EJM6; lh
UniProt ID: (Human) O00305, (Rat) D4A055, (Mouse) Q8R0S4
Entrez Gene ID: (Human) 785, (Rat) 58942, (Mouse) 12298
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support