Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is human brain tissue lysate.
PA5-20518 can be used with blocking peptide PEP-0638.
CAPS2 and the related protein CAPS1 encode novel neural/endocrine-specific cytosolic and peripheral membrane proteins. Both are essential components of the synaptic vesicle priming machinery and are required for the Ca2+-regulated exocytosis of secretory vesicles; CAPS-deficient neurons contain no or very few fusion competent synaptic vesicles, causing a selective impairment of fast phasic transmitter release. Like CAPS1, numerous isoforms of CAPS2 are known to exist. These isoforms show distinct expression patterns in the brain. For example, CAPS2b showed high expression in the developing cerebellum. Furthermore, one version of CAPS2 mRNA that lacks exon 3 is detected in some autistic patients, suggesting that the differential expression pattern of CAPS2 is involved in neuronal development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ca++-dependent secretion activator 2; Ca2+ dependent secretion activator 2; Ca2+-dependent activator protein for secretion 2; Calcium-dependent activator protein for secretion 2; Calcium-dependent secretion activator 2; CAPS-2; cerebellum postnatal development associated protein 2
Gene Aliases: A230044C21Rik; CADPS2; Caps-2; CAPS2; Cpd2; KIAA1591; RGD1559440
UniProt ID: (Human) Q86UW7, (Mouse) Q8BYR5
Entrez Gene ID: (Human) 93664, (Mouse) 320405, (Rat) 312166
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support