Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Recommended positive controls: Jurkat, Raji, NCI-H929.
Predicted reactivity: Human (99%), Chimpanzee (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen nuclear dot 52 kDa protein; calcium binding and coiled-coil domain 2; Calcium-binding and coiled-coil domain-containing protein 2; Nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; Nuclear domain 10 protein NDP52; Nuclear dot protein 52
Gene Aliases: 2410154J16Rik; C77254; CALCOCO2; NDP52; Ndp52l1; RGD1559974
UniProt ID: (Human) Q13137, (Mouse) A2A6M5
Entrez Gene ID: (Human) 10241, (Mouse) 76815, (Rat) 303479
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support