Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
CRLR is a receptor for calcitonin gene-related peptide type 1. The activity of CRLR is mediated by G proteins which activate adenylyl cyclase. CRLR expression has been reported in brain, lung, blood vessel, liver, and intestinal tract. ESTs have been isolated from B-cell/lung/testis, bome marrow, embryo, lung and synovium libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcitonin gene-related peptide type 1 receptor; calcitonin gene-related peptide type 1 receptor-like; calcitonin receptor-like; Calcitonin receptor-like receptor; calcitonin-like peptide type 1 receptor; CGRP receptor; CGRP type 1 receptor; Cgrp1 receptor; G protein-coupled receptor
Gene Aliases: CALCRL; CGRPR; CLR; CRLR; RATCRLR; RNCLR
UniProt ID: (Bovine) A6QP74, (Human) Q16602, (Pig) Q8WN93, (Rat) Q63118
Entrez Gene ID: (Rabbit) 100354426, (Bovine) 527140, (Human) 10203, (Pig) 397172, (Horse) 100054281, (Dog) 488438, (Goat) 100861156, (Rat) 25029, (Chinese hamster) 100753229
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support