Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T.
Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (84%), Pig (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAMK1D (CaMK1 delta) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. CAMK1D is expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis. Calcium/clamodulin-dependent protein kinase 1D-beta isoform is a novel calmodulin-kinase that exhibits Ca (2+)/CaM-dependent activity. CAM-Kkalpha upregulates CAMK1D-b activity through phosphorylation of its Thr180.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium/calmodulin-dependent protein kinase ID; Calcium/calmodulin-dependent protein kinase type 1D; CaM kinase I delta; CaM kinase ID; caM-KI delta; CAMKI; caMKI delta; CaMKI-delta; CaMKI-like protein kinase; CKLiK; MGC120317; MGC120318; OTTHUMP00000019119; OTTHUMP00000019120; OTTHUMP00000045030; RP11-462F15.1
Gene Aliases: CaM-K1; CAMK1D; CAMKID; CKLiK
UniProt ID: (Human) Q8IU85
Entrez Gene ID: (Human) 57118
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support