Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The antibody 13730-1-AP recognizes all the members of CAMK2 family.
Immunogen sequence: MATITCTRF TEEYQLFEEL GKGAFSVVRR CVKVLAGQEY AAKIINTKKL SARDHQKLER EARICRLLKH PNIVRLHDSI SEEGHHYLIF DLVTGGELFE DIVAREYYSE ADASHCIQQI LEAVLHCHQM GVVHRDLKPE NLLLASKLKG AAVKLADFGL AIEVEGEQQA WFGFAGTPGY LSPEVLRKDP YGKPVDLWAC GVILYILLVG YPPFWDEDQH RLYKQIKAGA YDFPSPEWDT VTPEAKDLIN KMLTINPSKR ITAAEALKHP WI (1-271 aa encoded by BC040457)
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; Calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; Calcium/calmodulin-dependent protein kinase type II subunit delta; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; CaM kinase II subunit alpha; CaM kinase II subunit beta; CaM kinase II subunit delta; CaM kinase II subunit gamma; CaM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; caM-kinase II gamma chain; CaMK II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; caMK-II subunit gamma; CaMKIINalpha; proline rich calmodulin-dependent protein kinase
Gene Aliases: 2810011D23Rik; 8030469K03Rik; [d]-CaMKII; CAM2; CAMK; CaMK II; CAMK-II; CAMK1; CAMK2; CAMK2A; CAMK2B; CAMK2D; CAMK2G; CAMKA; CAMKB; CAMKD; CAMKG; Camki; CaMKII; Ck2b; KIAA0968; Kiaa4163; mKIAA0968; PK2CDD; PKCCD; R74975; RATCAMKI
UniProt ID: (Human) Q9UQM7, (Human) Q13554, (Human) Q13557, (Human) Q8N4I3, (Mouse) P11798, (Rat) P11275, (Rat) P08413, (Mouse) P28652, (Rat) P15791, (Mouse) Q6PHZ2, (Mouse) Q923T9, (Rat) P11730
Entrez Gene ID: (Human) 815, (Human) 816, (Human) 817, (Human) 818, (Mouse) 12322, (Rat) 25400, (Rat) 24245, (Mouse) 12323, (Rat) 24246, (Mouse) 108058, (Mouse) 12325, (Rat) 171140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support