Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MLKVTVPSCS ASSCSSVTAS AAPGTASLVP DYWIDGSNRD ALSDFFEVES ELGRGATSIV YRCKQKGTQK PYALKVLKKT VDKKIVRTEI GVLLRLSHPN IIKLKEIFET PTEISLVLEL VTGGELFDRI VEKGYYSERD AADAVKQILE AVAYLHENGI VHRDLKPENL LYATPAPDAP LKIADFGLSK IVEHQVLMKT VCGTPGYCAP EILRGCAYGP EVDMWSVGII TYILLCGFEP FYDERGDQFM FRRILNCEYY FISPWWDEVS LNAKDLVRKL IVLDPKKRLT TFQALQHPWV TGKAANFVHM DTAQKKLQEF NARRKLKAAV KAVVASSRLG SASSSHGSIQ ESHKASRDPS PIQDGNEDMK AIPEGEKIQG DGAQAAVKGA QAELMKVQAL EKVKGADINA EEAPKMVPKA VEDGIKVADL ELEEGLAEEK LKTVEEAAAP REGQGSSAVG FEVPQQDVIL PEY
CAMK4 (CaMKIV) is a serine/threonine protein kinase that phosphoryates CREB, oncoprotein 18, and SRF. It has limited tissue distribution, and has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells. The nuclear localization of this protein is consistent with its role in mediating calcium-dependent gene expression. CaMKIV is particularly abundant in testis, T-cells, and neurons but is also found in other tissues to varying degrees. In neurons, CaMKIV is thought to play an important role in synaptic plasticity via its gene regulatory effects. In T-cells, this protein plays an important role in calcium signaling which could affect the transcription regulatory protein, nuclear factor of activated T-cells (NFAT). CaMKIV is encoded, along with calspermin, by the CaMKIV gene. It has been found that, in testes, CaMKIV is expressed in germ cells and found to be associated with chromatin. The association of CaMKIV with chromatin suggests a potential role in chromatin remodeling during nuclear condensation in spermatids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain Ca(2+)-calmodulin-dependent protein kinase type IV; brain Ca++-calmodulin-dependent protein kinase type IV; Calcium/calmoduli; calcium/calmodulin-dependent protein kinase IV; Calcium/calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase type IV catalytic chain; CAM kinase IV; CAM kinase- GR; CaM kinase-GR; CaMK IV; EC 2.7.11.17; KCC4; kinase CaMK4; OTTHUMP00000222818
Gene Aliases: CAMK; CaMK IV; CAMK-GR; CAMK4; CAMKIV
UniProt ID: (Human) Q16566
Entrez Gene ID: (Human) 814
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support