Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KADVPVEKY DGESDKEQFD DDQKVCCGFF FKDDQKAEND MAMKRAALLE KRLRREKETQ LRKQQLEAEM EHKKEETRRK TEEERQKKED ERARREFIRQ EYMRRKQLKL MEDMDTVIKP RPQVVKQKKQ RPKSIHRDHI ESPKTPIKGP PVSSLSLASL NTGDNESVHS GKRTPRSESV EGFLSPSRCG SRNGEKDWEN ASTTSSVASG TEYTGPKLYK EPSAKSNKHI IQNALAHCCL AGKVNEGQKK KILEEMEKSD ANNFLILFRD SGCQFRSLYT YCPETEEINK LTGIGPKSIT KKMIEGLYKY NSDRKQFSHI PAKTLSASVD AITIHSHLWQ TKRPVTPKKL LPTKA (551-904 aa encoded by BC065508)
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calmodulin regulated spectrin-associated protein 1-like 1; calmodulin regulated spectrin-associated protein family, member 2; Calmodulin-regulated spectrin-associated protein 1-like protein 1; Calmodulin-regulated spectrin-associated protein 2
Gene Aliases: 1600013L13Rik; 4930541M15Rik; CAMSAP1L1; CAMSAP2; KIAA1078; mKIAA1078; RGD1310950
UniProt ID: (Human) Q08AD1, (Mouse) Q8C1B1, (Rat) D4AEC2
Entrez Gene ID: (Human) 23271, (Mouse) 67886, (Rat) 289400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support