Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CAP2 Antibodies

            Proteintech

            CAP2 Polyclonal Antibody

            6 References
            View all (11) CAP2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # 15865-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (EUR)
            437,00
            150 µL
             
            Check your price
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite CAP2 Polyclonal Antibody

            • Antibody Testing Data (14)
            Application
            CAP2 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 14

            CAP2 Antibody (15865-1-AP) in ICC/IF

            Immunofluorescent analysis of ( -20°C Ethanol) fixed U2OS cells using 15865-1-AP (CAP2 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            CAP2 Antibody in Immunocytochemistry (ICC/IF)
            CAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Western Blot (WB)
            CAP2 Antibody in Immunoprecipitation (IP)

            Product Details

            15865-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:3,000
            View 3 publications 3 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            1:50-1:500
            View 1 publication 1 publication

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication

            Not applicable (N/A)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            CAP2 Fusion Protein Ag8477 (129-477 aa encoded by BC008481)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.19 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MFNHLSAVS ESIPALGWIA VSPKPGPYVK EMNDAATFYT NRVLKDYKHS DLRHVDWVKS YLNIWSELQA YIKEHHTTGL TWSKTGPVAS TVSAFSVLSS GPGLPPPPPP LPPPGPPPLF ENEGKKEESS PSRSALFAQL NQGEAITKGL RHVTDDQKTY KNPSLRAQGG QTQSPTKSHT PSPTSPKSYP SQKHAPVLEL EGKKWRVEYQ EDRNDLVISE TELKQVAYIF KCEKSTIQIK GKVNSIIIDN CKKLGLVFDN VVGIVEVINS QDIQIQVMGR VPTISINKTE GCHIYLSEDA LDCEIVSAKS SEMNILIPQD GDYREFPIPE QFKTAWDGSK LITEPAEIMA (129-477 aa encoded by BC008481)

            Target Information

            This gene was identified by its similarity to the gene for human adenylyl cyclase-associated protein. The function of the protein encoded by this gene is unknown. However, the protein appears to be able to interact with adenylyl cyclase-associated protein and actin.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: 2810452G09Rik; Adenylyl cyclase-associated protein 2; CAP 2

            View more View less

            Gene Aliases: 2810452G09Rik; AV045235; CAP2

            View more View less

            UniProt ID: (Human) P40123, (Rat) P52481, (Mouse) Q9CYT6

            View more View less

            Entrez Gene ID: (Human) 10486, (Rat) 116653, (Mouse) 67252

            View more View less

            Function(s)

            actin binding protein binding adenylate cyclase binding actin or actin-binding cytoskeletal protein cytoskeletal protein

            Process(es)

            cell morphogenesis establishment or maintenance of cell polarity signal transduction activation of adenylate cyclase activity actin polymerization or depolymerization cytoskeleton organization actin cytoskeleton organization
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            • Price & Freight Policy
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • German Supply Chain Due Diligence Act
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Legal Notice
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Germany flag icon
            Germany

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-7765t:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0