Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTPVDVPVT NPAATILPVH VYPLPQQMRV AFSAARTSNL APGTLDQPIV FDLLLNNLGE TFDLQLGRFN CPVNGTYVFI FHMLKLAVNV PLYVNLMKNE EVLVSAYAND GAPDHETASN HAILQLFQGD QIWLRLHRGA IYGSSWKYST FSGYLLYQD (970-1128 aa encoded by BC007520)
The protein encoded by this gene may regulate the transport of mRNA. It may play a role in the differentiation of erythroblasts. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C1q domain containing 1; C1q domain-containing protein 1; caprin family member 2; Caprin-2; caprin2; Cytoplasmic activation/proliferation-associated protein 2; Gastric cancer multidrug resistance-associated protein; Protein EEG-1; RNA granule protein 140
Gene Aliases: C1QDC1; CAPRIN2; EEG-1; EEG1; KIAA1873; RNG140
UniProt ID: (Human) Q6IMN6, (Mouse) Q05A80
Entrez Gene ID: (Human) 65981, (Rat) 686779, (Mouse) 232560
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support