Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Bovine, Pig and Orangutan
CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cap Z; capping protein (actin filament) muscle Z-line, beta; capping protein beta 1; CapZ beta; F-actin-capping protein subunit beta; MGC104401; MGC129749; MGC129750; RP4-657E11.7
Gene Aliases: 1700120C01Rik; AI325129; CAPB; CAPPB; Cappb1; CAPZ; CAPZB; CPB1; CPB2; CPbeat2; CPbeta1; CPbeta2
UniProt ID: (Human) P47756, (Mouse) P47757
Entrez Gene ID: (Human) 832, (Mouse) 12345
Molecular Function:
actin or actin-binding cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support