Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AQEDAELQP RALDIYSAVD DASHEKELIE ALQEVLKKLK SKRVPIYEKK YGQVPMCDAG EQCAVRKGAR IGKLCDCPRG TSCNSFLLKC L (27-116 aa encoded by BC029882)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cocaine and amphetamine regulated transcript; Cocaine- and amphetamine-regulated transcript protein
Gene Aliases: CART; CARTPT
UniProt ID: (Human) Q16568, (Mouse) P56388, (Rat) P49192
Entrez Gene ID: (Human) 9607, (Mouse) 27220, (Rat) 29131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support