Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: EEGLNFPTYD GKDRVVSLSE KNFKQVLKKY DLLCLYYHEP VSSDKVTQKQ FQLKEIVLEL VAQVLEHKAI GFVMVDAKKE AKLAKKLGFD EEGSLYILKG DRTIEFDGEF AADVLVEFLL DLIEDPVEII SSKLEVQAFE RIEDYIKLIG FFKSEDSEYY KAFEEAAEHF QPYIKFFATF DKGVAKKLSL KMNEVDFYEP FMDEPIAIPN KPYTEEELVE FVKEHQRPTL RRLRPEEMFE TWEDDLNGIH IVAFAEKSDP DGYEFLEILK QVARDNTDNP DLSILWIDPD DFPLLVAYWE KTFKIDLFRP QIGVVNVTDA DSVWMEIPDD DDLPTAEELE DWIEDVLSGK INTEDDDEDD DDDDNSDEED NDDSDDDDDE
Calsequestrin (CASQ) is a Ca2+-binding protein present primarily in junctional sarcoplasmic reticulum of skeletal and cardiac muscle; the cardiac form (CASQ2) is encoded by a separate gene. The primary role of CASQ2 is buffering of the sarcoplasmic reticulum Ca2+ ions, but another role for CASQ2 has emerged recently: CASQ2 regulates the open probability of ryanodine receptor 2 (RyR2). Mutations in CASQ2 cause stress-induced polymorphic ventricular tachycardia, also referred to as catecholaminergic polymorphic ventricular tachycardia 2 (CPVT2), a disease characterized by bidirectional ventricular tachycardia that may lead to cardiac arrest.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calsequestrin 2 (cardiac muscle); calsequestrin 2, fast-twitch, cardiac muscle; Calsequestrin, cardiac muscle isoform; Calsequestrin-2
Gene Aliases: CASQ2; PDIB2
UniProt ID: (Human) O14958
Entrez Gene ID: (Human) 845
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support