Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Calcium ions play a primary role in the regulation of sperm motility. Anti-CATSPER2 belongs to a family of putative cation channels that are specific to spermatozoa and localize to the flagellum. The protein family features a single repeat with six membrane-spanning segments and a predicted calcium-selective pore region. A second, closely linked copy of this gene has been identified. Although multiple transcript variants encoding protein isoforms have been characterized, they seem to be only transcribed from this gene. Additional splice variants have been described but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cation channel sperm-associated protein 2; cation channel, sperm associated 2; CatSper2; sperm ion channel
Gene Aliases: CATSPER2
UniProt ID: (Human) Q96P56
Entrez Gene ID: (Human) 117155
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support