Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MGFHHVGQAR LELLTSGDLP ALASQRAGIT VGPEKRVPAM PGSPVEVKIQ SRSSPPTMPP LPPINPGGPR PVSFTPTALS NGINHSPPTL NGAPSPPQRF SNGPASSTSS ALTNQQLPAT CGARQLSKLK RFLTTLQQFG NDISPEIGEK VRTLVLALVN STVTIEEFHC KLQEATNFPL RPFVIPFLKA NLPLLQRELL HCARAAKQTP SQYLAQHEHL LLNTSIASPA DSSELLMEVH GNGKRPSPER REENSFDRDT IAPEPPAKRV CTISPAPRHS PALTVPLMNP GGQFHPTPPP LQHYTLEDIA TSHLYREPNK MLEHREVRDR HHSLGLNGGY QDELVDHRLT EREWADEWKH LDHALNCIME MVEKTRRSMA VLRRCQGSDR EELNYWKRRY NENTELRKTG TELVSRQHSP GSADSLSNDS QREFNSRPGT GYVPVEFWKK TEEAVNKVKI QAMSEVQKAV AEAEQKAFEV IATERARMEQ TIADVKRQAA EDAFLVINEQ EESTENCWNC GRKASETCSG CNIARYCGSF CQHKDWERHH RLCGQNLHGQ SPHGQGRPLL PVGRGSSARS ADCSVPSPAL DKTSATTSRS STPASVTAID TNGL
Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as a complex with MTGR1 and that the complex might be important in promoting leukemogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: core-binding factor, runt domain, alpha subunit 2; translocated to, 2; DKFZp313F2116; ETO homolog on chromosome 20; ETO homologous on chromosome 20; MTG8-like protein; MTG8-related protein 1; myeloid translocation gene-related protein 1; Myeloid translocation-related protein 1; p85; Protein CBFA2T2; RP5-1137F22.1; ZMYND3 CBFA2T2
Gene Aliases: CBFA2T2; EHT; MTGR1; p85; ZMYND3
UniProt ID: (Human) O43439
Entrez Gene ID: (Human) 9139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support