Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: MSSCSRVAL VTGANRGIGL AIARELCRQF SGDVVLTARD VARGQAAVQQ LQAEGLSPRF HQLDIDDLQS IRALRDFLRK EYGGLNVLVN NAAVAFKSDD PMPFDIKAEM TLKTNFFATR NMCNELLPIM KPHGRVVNIS SLQCLRAFEN CSEDLQERFH SETLTEGDLV DLMKKFVEDT KNEVHEREGW PNSPYGVSKL GVTVLSRILA RRLDEKRKAD RILVNACCPG PVKTDMDGKD SIRTVEEGAE TPVYLALLPP DATEPQGQLV HDKVVQNW (1-277 aa encoded by BC002812)
Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carbonyl reductase (NADPH) 3; Carbonyl reductase [NADPH] 3; epididymis secretory protein Li 25; Monomeric carbonyl reductase 3; NADPH-dependent carbonyl reductase 3; Quinone reductase CBR3; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2
Gene Aliases: 1110001J05Rik; C81353; CBR3; hCBR3; HEL-S-25; SDR21C2
UniProt ID: (Human) O75828, (Mouse) Q8K354, (Rat) B2GV72
Entrez Gene ID: (Human) 874, (Mouse) 109857, (Rat) 304078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support