Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MPSETPQAEV GPTGCPHRSG PHSAKGSLEK GSPEDKEAKE PLWIRPDAPS RCTWQLGRPA SESPHHHTAP AKSPKILPDI LKKIGDTPMV RINKIGKKFG LKCELLAKCE FFNAGGSVKD RISLRMIEDA ERDGTLKPGD TIIEPTSGNT GIGLALAAAV RGYRCIIVMP EKMSSEKVDV LRALGAEIVR TPTNARFDSP ESHVGVAWRL KNEIPNSHIL DQYRNASNPL AHYDTTADEI LQQCDGKLDM LVASVGTGGT ITGIARKLKE KCPGCRIIGV DPEGSILAEP EELNQTEQTT YEVEGIGYDF IPTVLDRTVV DKWFKSNDEE AFTFARMLIA QEGLLCGGSA GSTVAVAVKA AQELQEGQRC VVILPDSVRN YMTKFLSDRW MLQKGFLKEE DLTEKKPWWW HLRVQELGLS APLTVLPTIT CGHTIEILRE KGFDQAPVVD EAGVILGMVT LGNMLSSLLA GKVQPSDQVG KVIYKQFKQI RLTDTLGRLS HILEMDHFAL VVHEQIQYHS TGKSSQRQMV FGVVTAIDLL NFVAAQERDQ K
Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-thionase; Cystathionine beta-synthase; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: AI047524; AI303044; CBS; HIP4
UniProt ID: (Human) P35520, (Mouse) Q91WT9
Entrez Gene ID: (Human) 875, (Mouse) 12411
Molecular Function: lyase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support