Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: VAWRLKNEIP NSHILDQYRN ASNPLAHYDT TADEILQQCD GKLDMLVASV GTGGTITGIA RKLKEKCPGC RIIGVDPEGS ILAEPEELNQ TEQTTYEVEG IGYDFIPTVL DRTVVDKWFK SNDEEAFTFA RMLIAQEGLL CGGSAGSTVA VAVKAAQELQ EGQRCVVILP DSVRNYMTKF LSDRWMLQKG FLKEEDLTEK KPWWWHLRVQ ELGLSAPLTV LPTITCGHTI EILREKGFDQ APVVDEAGVI LGMVTLGNML SSLLAGKVQP SDQVGKVIYK QFKQIRLTDT LGRLSHILEM DHFALVVHEQ IQYHSTGKSS QRQMVFGVVT AIDLLNFVAA QERDQK
Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-thionase; Cystathionine beta-synthase; cystathionine-beta-synthase; Hemoprotein H-450; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: CBS; HIP4
UniProt ID: (Human) P35520, (Rat) P32232
Entrez Gene ID: (Human) 875, (Rat) 24250
Molecular Function: lyase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support