Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-thionase; Cystathionine beta-synthase; cystathionine-beta-synthase; Hemoprotein H-450; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: AI047524; AI303044; CBS; HIP4
UniProt ID: (Human) P35520, (Rat) P32232, (Mouse) Q91WT9
Entrez Gene ID: (Human) 875, (Rat) 24250, (Mouse) 12411
Molecular Function: lyase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support