Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: 293, human liver tissue lysate, 293T.
Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-thionase; Cystathionine beta-synthase; methylcysteine synthase; Serine sulfhydrase
Gene Aliases: CBS; HIP4
UniProt ID: (Human) P35520
Entrez Gene ID: (Human) 875
Molecular Function: lyase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support