Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: CBX3.
At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromobox homolog 3; chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox protein homolog 3; heterochromatin protein 1 gamma; heterochromatin protein 1 homolog gamma; HGNC:1553; HP1 gamma; HPy; modifier 2 protein; OTTHUMP00000202463
Gene Aliases: Hp1-gamma; HP1g; M32
Entrez Gene ID: (Mouse) 12417, (Rat) 297093
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support