Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHKRKGPPGP PGRGAAAARQ LGLLVDLSPD GLMIPEDGAN DEELEAEFLA LVGGQPPALE KLKGKGPLPM EAIEKMASLC MRDPDEDEEE GTDEDDLEAD DDLLAELNEV LGEEQKASET PPPVAQPKPE APHPGLETTL QERLALYQTA IESARQAGDS AKMRRYDRGL KTLENLLASI RKGNAIDEAD IPPPVAIGKG PASTPTYSPA PTQPAPRIAS APEPRVTLEG PSATAPASSP GLAKPQMPPG PCSPGPLAQL QSRQRDYKLA ALHAKQQGDT TAAARHFRVA KSFDAVLEAL SRGEPVDLSC LPPPPDQLPP DPPSPPSQPP TPATAPSTTE VPPPPRTLLE ALEQRMERYQ VAAAQAKSKG DQRKARMHER IVKQYQDAIR AHKAGRAVDV AELPVPPGFP PIQGLEATKP TQQSLVGVLE TAMKLANQDE GPEDEEDEVP KKQNSPVAPT AQPKAPPSRT PQSGSAPTAK APPKATSTRA QQQLAFLEGR KKQLLQAALR AKQKNDVEGA KMHLRQAKGL EPMLEASRNG LPVDITKVPP APVNKDDFAL VQRPGPGLSQ EAARRYGELT KLIRQQHEMC LNHSNQFTQL GNITETTKFE KLAEDCKRSM DILKQAFVRG LPTPTARFEQ RTFSVIKIFP DLSSNDMLLF IVKGINLPTP PGLSPGDLDV FVRFDFPYPN VEEAQKDKTS VIKNTDSPEF KEQFKLCINR SHRGFRRAIQ TKGIKFEVVH KGGLFKTDRV LGTAQLKLDA LEIACEVREI LEVLDGRRPT GGRLEVMVRI REPLTAQQLE TTTERWLVID PVPAAVPTQV AGPKGKAPPV PAPARESGNR SARPLHSLSV LAFDQERLER KILALRQARR PVPPEVAQQY QDIMQRSQWQ RAQLEQGGVG IRREYAAQLE RQLQFYTEAA RRLGNDGSRD AAKEALYRRN LVESELQRLR R
This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5'-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Akt kinase-interacting protein 1; Coiled-coil and C2 domain-containing protein 1A; Five prime repressor element under dual repression-binding protein 1; five repressor element under dual repression-binding protein 1; FLJ20241; FRE under dual repression-binding protein 1; mental retardation, nonsyndromic, autosomal recessive, 3; Putative NF-kappa-B-activating protein 023N; putative NFkB activating protein
Gene Aliases: AKI1; CC2D1A; FREUD-1; Freud-1/Aki1; LGD2; MRT3
UniProt ID: (Human) Q9H603
Entrez Gene ID: (Human) 54862
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support