Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MHKRKGPPGP PGRGAAAARQ LGLLVDLSPD GLMIPEDGAN DEELEAEFLA LVGGQPPALE KLKGKGPLPM EAIEKMASLC MRDPDEDEEE GTDEDDLEAD DDLLAELNEV LGEEQKASET PPPVAQPKPE APHPGLETTL QERLALYQTA IESARQAGDS AKMRRYDRGL KTLENLLASI RKGNAIDEAD IPPPVAIGKG PASTPTYSPA PTQPAPRIAS APEPRVTLEG PSATAPASSP GLAKPQMPPG PCSPGPLAQL QSRQRDYKLA ALHAKQQGDT TAAARHFRVA KSFDAVLEAL SRGEPVDLSC LPPPPDQLPP DPPSPPSQPP TPATAPSTTE VPPPPRTLLE A
This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5'-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-HT1A receptor; Akt kinase-interacting protein 1; Coiled-coil and C2 domain-containing protein 1A; Five prime repressor element under dual repression-binding protein 1; five repressor element under dual repression-binding protein 1; five' repressor element under dual repression binding protein 1; FLJ20241; FRE under dual repression-binding protein 1; freud-1; mental retardation, nonsyndromic, autosomal recessive, 3; Putative NF-kappa-B-activating protein 023N; putative NFkB activating protein
Gene Aliases: AKI1; BC016188; CC2D1A; FREUD-1; Freud-1/Aki1; LGD2; MRT3; RGD1306108; Tape
UniProt ID: (Human) Q9H603, (Rat) Q66HA5, (Mouse) Q8K1A6
Entrez Gene ID: (Human) 54862, (Rat) 288908, (Mouse) 212139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support