Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Peptide sequence: YSQRFGTVQR EVKGPTPKVV IVRSKPPKGQ GAEHHLERIR RSHQKHNAIL
Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 77%; Horse: 85%; Human: 100%; Pig: 85%
This gene belongs to the CFAP53 family. It was found to be differentially expressed by the ciliated cells of frog epidermis and in skin fibroblasts from human. Mutations in this gene are associated with visceral heterotaxy-6, which implicates this gene in determination of left-right asymmetric patterning. [provided by RefSeq, Aug 2015]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 53; coiled-coil domain containing 11; Coiled-coil domain-containing protein 11
Gene Aliases: CCDC11; CFAP53; HTX6
UniProt ID: (Human) Q96M91
Entrez Gene ID: (Human) 220136
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support