Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCDC112 (Coiled-Coil Domain Containing 112) is a Protein Coding gene and its exact function is unknown. An important paralog of this gene is CCDC148.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 112; mutated in bladder cancer 1; Mutated in bladder cancer protein 1
Gene Aliases: CCDC112; MBC1
UniProt ID: (Human) Q8NEF3
Entrez Gene ID: (Human) 153733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support