Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TIAEYRAIVM KNKEEEERQR KIEAKEQLLA VMKADQIFWE HEKEKKCKAD KEHQEVQDAH IQQMAKNKFN AKQAKQAELD YCRLT
Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 69%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella- associated protein 210; Coiled-coil domain-containing protein 173
Gene Aliases: C2orf77; CCDC173; CFAP210
UniProt ID: (Human) Q6PJF6
Entrez Gene ID: (Human) 129881
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support