Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
A suggested positive control is mouse heart tissue lysate.
PA5-20855 can be used with blocking peptide PEP-0969.
The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. The CCDC47 gene maps to chromosome 17 at 17q23.3. Little is known about this single-pass membrane protein except that the coiled-coil domain is within the cytosolic domain near the carboxy terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipocyte-specific protein 4; Calumin; ccd47; Coiled-coil domain-containing protein 47; PAT complex subunit CCDC47
Gene Aliases: 2610204L23Rik; Asp4; C88307; calumin; CCDC47; GK001; MSTP041; PSEC0077; RGD1308813
UniProt ID: (Human) Q96A33, (Rat) Q5U2X6, (Mouse) Q9D024
Entrez Gene ID: (Human) 57003, (Rat) 303606, (Mouse) 67163
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support