Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEVSIDQS KLPGVKEVCR DFAVLEDHTL AHSLQEQEIE HHLASNVQRN RLVQHDLQVA KQLQEEDLKA QAQLQKRYKD LEQQDCEIAQ EIQEKLAIEA ERRRIQEKKD EDIARLLQEK ELQEEKKRKK HFPEFPATRA YADSYYYEDG GMKPRVMKEA VSTPSRMAHR DQEWYDAEIA RKLQEEELLA TQVDMRAAQV AQDEEIARLL MAEEKKAYKK AKEREKSSLD KRKQDPEWKP KTAKAANSKS KESDEPHHSK NERPARPPPP IMTDGEDADY THFTNQQSST RHFSKSESSH KGFHYKH (1-306 aa encoded by BC065004)
This protein is a bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 50; Protein Ymer; putative C3orf6 protein homolog; Ymer protein
Gene Aliases: C3orf6; C3orf6h; CCDC50; DFNA44; YMER
UniProt ID: (Human) Q8IVM0, (Mouse) Q810U5, (Rat) Q810U0
Entrez Gene ID: (Human) 152137, (Mouse) 67501, (Rat) 288022
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support