Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MATPPFRLI RKMFSFKVSR WMGLACFRSL AASSPSIRQK KLMHKLQEEK AFREEMKIFR EKIEDFREEM WTFRGKIHAF RGQILGFWEE ERPFWEEEKT FWKEEKSFWE MEKSFREEEK TFWKKYRTFW KEDKAFWKED NALWERDRNL LQEDKALWEE EKALWVEERA LLEGEKALWE DKTSLWEEEN ALWEEERAFW MENNGHIAGE QMLEDGPHNA NRGQRLLAFS RGRA (1-233 aa encoded by BC069770)
CCDC70 (Coiled-Coil Domain Containing 70) is a Protein Coding gene. An important paralog of this gene is SPERT.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 70
Gene Aliases: 1700112P19Rik; CCDC70
UniProt ID: (Human) Q6NSX1, (Mouse) Q9D9B0
Entrez Gene ID: (Human) 83446, (Mouse) 67929
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support