Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.
Immunogen sequence: KEDNALWERD RNLLQEDKAL WEEEKALWVE ERALLEGEKA LWEDKTSLWE EENALWEEER AFWMENNGHI AGEQMLEDG
CCDC70 (Coiled-Coil Domain Containing 70) is a Protein Coding gene. An important paralog of this gene is SPERT.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 70
Gene Aliases: CCDC70
UniProt ID: (Human) Q6NSX1
Entrez Gene ID: (Human) 83446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support