Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: AEVEDRRAA MERQLISMKV KYQSLKKQNV FNREQMQRMK LQIATLLQMK GSQTEFEQQE RLLAMLEQKN GEIKHLLGEI RNLEKFKNLY DSMESKPSVD SGTLEDNTYY TDLLQMKLDN LNKEIESTKG ELSIQRMKAL FESQRALDIE RKLFANERCL QLSESENMKL RAKLDELKLK YEPEETVEVP VLKKRREVLP VDITTAKDAC VNNSALGGEV YRLPPQKEET QSCPNSLEDN NLQLEKSVSI HTPVVSLSPH KNLPVDMQLK KEKKCVKLIG VPADAEALSE RSGNTPNSPR LAAESKLQTE VKEGKETSSK LEKETCKKSH PILYVSSKST PETQCPQQ (259-605 aa encoded by BC012568)
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridizations indicate that this lectin is specifically expressed in keratinocytes. It is expressed at all stages of epidermal differentiation (i.e., in basal and suprabasal layers). The protein was found mainly in stratified squamous epithelium. It is moderately repressed by retinoic acid. The antigen localized to basal keratinocytes, although it was also found at lower levels in the suprabasal layers, where it concentrated to areas of cell-to-cell contact. The cellular localization and its striking down-regulation in cultured keratinocytes imply a role in cell-cell and/or cell-matrix interactions necessary for normal growth control.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arsenite-related gene 1 protein; coiled-coil domain containing 99; Coiled-coil domain-containing protein 99; hSpindly; Protein Spindly; Rhabdomyosarcoma antigen MU-RMS-40.4A; rrhabdomyosarcoma antigen protein MU-RMS-40.4A; SPDL1; Spindle apparatus coiled-coil domain-containing protein 1; spindly homolog
Gene Aliases: CCDC99; SPDL1
UniProt ID: (Human) Q96EA4
Entrez Gene ID: (Human) 54908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support