Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: Unboiled HeLa cell extract.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCK-B receptor; CCK-BR; CCK2 receptor; CCK2-R; CCKB; Cckb receptor; CCKB-R; Cholecystokinin-2 receptor; Cholecystokinin-B; gastrin receptor; Gastrin/cholecystokinin type B receptor; HGNC:1571
Gene Aliases: CCK-B; CCK2R; CCKBR; CCKRB; GASR
UniProt ID: (Human) P32239
Entrez Gene ID: (Human) 887
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support