Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; HCC-4; IL-10-inducible chemokine; LCC-1; liver CC chemokine-1; Liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; Monotactin-1; MTN-1; NCC-4; new CC chemokine 4; small inducible cytokine subfamily A (Cys-Cys), member 16; Small-inducible cytokine A16
Gene Aliases: CCL16; CKb12; HCC-4; ILINCK; LCC-1; LEC; LMC; Mtn-1; NCC-4; NCC4; SCYA16; SCYL4
UniProt ID: (Human) O15467
Entrez Gene ID: (Human) 6360
Molecular Function:
cytokine
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support