Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Eotaxin-3 is a small cytokine belonging to the CC chemokine family that is also called CCL26, Chemokine (C-C motif) ligand 26. Chemokines play pivotal roles in health and disease by controlling migration and tissue localization of specific types of cells expressing their cognate receptors. Eotaxin-3 is chemotactic for eosinophils and basophils and elicits its effects by binding to the cell surface chemokine receptor CCR3. It is expressed by several tissues including heart, lung and ovary, and in endothelial cells that have been stimulated with the cytokine interleukin 4. This gene for chemokine is located on human chromosome 7. Eotaxin-3 induced migration of eosinophils and basophils only at a 10-fold higher concentration than eotaxin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 26; CC chemokine IMAC; chemokine (C-C motif) ligand 26; chemokine N1; Eotaxin-3; Eotaxin3; Macrophage inflammatory protein 4-alpha; MGC126714; MIP-4-alpha; PRO242; SCYA26MIP-4-alpha; small inducible cytokine A26; small inducible cytokine subfamily A (Cys-Cys), member 26; Small-inducible cytokine A26; Thymic stroma chemokine-1; TSC-1; UNQ216
Gene Aliases: CCL26; IMAC; MIP-4a; MIP-4alpha; SCYA26; TSC-1; UNQ216/PRO242
UniProt ID: (Human) A0N0Q5
Entrez Gene ID: (Human) 10344
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support