Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSRRSSRLQA KQQPQPSQTE SPQEAQIIQA KKRKTTQDVK KRREEVTKKH QYEIRNCWPP VLSGGISPCI IIETPHKEIG TSDFSRFTNY RFKNLFINPS PLPDLSWGCS KEVWLNMLKK ESRYVHDKHF EVLHSDLEPQ MRSILLDWLL EVCEVYTLHR ETFYLAQDFF DRFMLTQKDI NKNMLQLIGI TSLFIASKLE EIYAPKLQEF AYVTDGACSE EDILRMELII LKALKWELCP VTIISWLNLF LQVDALKDAP KVLLPQYSQE TFIQIAQLLD LCILAIDSLE FQYRILTAAA LCHFTSIEVV KKASGLEWDS ISECVDWMVP FVNVVKSTSP VKLKTFKKIP MEDRHNIQTH TNYLAMLEEV NYINTFRKGG QLSPVCNGGI MTPPKSTEKP PGKH
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G1/S-specific cyclin-E2
Gene Aliases: CCNE2; CYCE2
UniProt ID: (Human) O96020, (Mouse) Q9Z238
Entrez Gene ID: (Human) 9134, (Mouse) 12448
Molecular Function:
kinase activator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support