Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: MKDLGAEHLA GHEGVQLLGL LNVYLEQEER FQPREKGLSL IEATPENDNT LCPGLRNAKV EDLRSLANFF GSCTETFVLA VNILDRFLAL MKVKPKHLSC IGVCSFLLAA RIVEEDCNIP STHDVIRISQ CKCTASDIKR MEKIISEKLH YELEATTALN FLHLYHTIIL CHTSERKEIL SLDKLEAQLK ACNCRLIFSK AKPSVLALCL LNLEVETLKS VELLEILLLV KKHSKINDTE FFYWRELVSK CLAEYSSPEC CKPDLKKLVW IVSRRTAQNL HNSYYSVPEL PTIPEGGCFD ESESSVAQAG VQWPDLSSFQ PPPTRFKRFS CLSLRSSWDY SSWT
Cyclin G2 may play a role in growth regulation and in negative regulation of cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclin-G2
Gene Aliases: CCNG2
UniProt ID: (Human) Q16589
Entrez Gene ID: (Human) 901
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support