Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCT-epsilon; Chaperonin containing T-complex polypeptide 1 subunit 5; chaperonin containing TCP1, subunit 5 (epsilon); chaperonin subunit 5 (epsilon); epididymis secretory protein Li 69; T-complex protein 1 subunit epsilon; TCP-1-epsilon
Gene Aliases: CCT-epsilon; CCT5; CCTE; HEL-S-69; KIAA0098; mKIAA0098; PNAS-102; TCP-1-epsilon; TCPE
UniProt ID: (Human) P48643, (Mouse) P80316, (Rat) Q68FQ0
Entrez Gene ID: (Human) 22948, (Mouse) 12465, (Rat) 294864
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support