Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DVTGDGTTSN VLIIGELLKQ ADLYISEGLH PRIIAEGFEA AK
Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 95%.
Molecular chaperone; assists the folding of proteins upon ATP hydrolysis. Known to play a role, in vitro, in the folding of actin and tubulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCT-zeta-2; CCT-zeta-like; Chaperonin containing T-complex polypeptide 1 subunit 6B; chaperonin containing TCP1, subunit 6B (zeta 2); chaperonin-containing T-complex polypeptide 1, subunit 6B; T-complex protein 1 subunit zeta-2; TCP-1-zeta-2; TCP-1-zeta-like; Testis-specific protein TSA303; Testis-specific Tcp20
Gene Aliases: CCT-zeta-2; CCT6B; CCTZ-2; Cctz2; TCP-1-zeta-2; TSA303
UniProt ID: (Human) Q92526
Entrez Gene ID: (Human) 10693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support