Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Possible molecular chaperone; assists the folding of proteins upon ATP hydrolysis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chaperonin containing TCP1, subunit 8 (theta)-like 2; putative T-complex protein 1 subunit theta-like 2; T-complex protein 1 subunit theta-like 2
Gene Aliases: CCT8L2; CESK1
UniProt ID: (Human) Q96SF2
Entrez Gene ID: (Human) 150160
Molecular Function:
chaperonin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support