Product References
IKK2 controls the inflammatory potential of tissue-resident regulatory T cells in a murine gain of function model.
Nature communications
Cardinez C,Hao Y,Kwong K,Davies AR,Downes MB,Roberts NA,Price JD,Hernandez RA,Lovell J,Chand R,Feng ZP,Enders A,Vinuesa CG,Miraghazadeh B,Cook MC
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Mon Mar 25 00:00:00 UTC 2024
Profiling of Tregs across tissues reveals plasticity in ST2 expression and hierarchies in tissue-specific phenotypes.
iScience
Spath S,Roan F,Presnell SR,Höllbacher B,Ziegler SF
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Fri Sep 16 00:00:00 UTC 2022
COVID-19 treatments and pathogenesis including anosmia in K18-hACE2 mice.
Nature
Zheng J,Wong LR,Li K,Verma AK,Ortiz ME,Wohlford-Lenane C,Leidinger MR,Knudson CM,Meyerholz DK,McCray PB,Perlman S
17-1031 was used in Flow cytometry/Cell sorting to show that K18-hACE2 mice provide a useful model for studying the pathological basis of both mild and lethal COVID-19 and for assessing therapeutic interventions.
Fri Jan 01 00:00:00 UTC 2021
An integrin αEβ7-dependent mechanism of IgA transcytosis requires direct plasma cell contact with intestinal epithelium.
Mucosal immunology
Guzman M,Lundborg LR,Yeasmin S,Tyler CJ,Zgajnar NR,Taupin V,Dobaczewska K,Mikulski Z,Bamias G,Rivera-Nieves J
17-1031-82 was used in Flow Cytometry to propose that IgA not only reaches pIgR through diffusion, but that αEβ7+ PC dock with E-cadherin-expressing intestinal epithelium to directly relay IgA for transcytosis into the intestinal lumen.
Mon Nov 01 00:00:00 UTC 2021
Guidelines for the use of flow cytometry and cell sorting in immunological studies (third edition).
European journal of immunology
Cossarizza A,Chang HD,Radbruch A,Abrignani S,Addo R,Akdis M,Andrä I,Andreata F,Annunziato F,Arranz E,Bacher P,Bari S,Barnaba V,Barros-Martins J,Baumjohann D,Beccaria CG,Bernardo D,Boardman DA,Borger J,Böttcher C,Brockmann L,Burns M,Busch DH,Cameron G,Cammarata I,Cassotta A,Chang Y,Chirdo FG,Christakou E,Čičin-Šain L,Cook L,Corbett AJ,Cornelis R,Cosmi L,Davey MS,De Biasi S,De Simone G,Del Zotto G,Delacher M,Di Rosa F,Di Santo J,Diefenbach A,Dong J,Dörner T,Dress RJ,Dutertre CA,Eckle SBG,Eede P,Evrard M,Falk CS,Feuerer M,Fillatreau S,Fiz-Lopez A,Follo M,Foulds GA,Fröbel J,Gagliani N,Galletti G,Gangaev A,Garbi N,Garrote JA,Geginat J,Gherardin NA,Gibellini L,Ginhoux F,Godfrey DI,Gruarin P,Haftmann C,Hansmann L,Harpur CM,Hayday AC,Heine G,Hernández DC,Herrmann M,Hoelsken O,Huang Q,Huber S,Huber JE,Huehn J,Hundemer M,Hwang WYK,Iannacone M,Ivison SM,Jäck HM,Jani PK,Keller B,Kessler N,Ketelaars S,Knop L,Knopf J,Koay HF,Kobow K,Kriegsmann K,Kristyanto H,Krueger A,Kuehne JF,Kunze-Schumacher H,Kvistborg P,Kwok I,Latorre D,Lenz D,Levings MK,Lino AC,Liotta F,Long HM,Lugli E,MacDonald KN,Maggi L,Maini MK,Mair F,Manta C,Manz RA,Mashreghi MF,Mazzoni A,McCluskey J,Mei HE,Melchers F,Melzer S,Mielenz D,Monin L,Moretta L,Multhoff G,Muñoz LE,Muñoz-Ruiz M,Muscate F,Natalini A,Neumann K,Ng LG,Niedobitek A,Niemz J,Almeida LN,Notarbartolo S,Ostendorf L,Pallett LJ,Patel AA,Percin GI,Peruzzi G,Pinti M,Pockley AG,Pracht K,Prinz I,Pujol-Autonell I,Pulvirenti N,Quatrini L,Quinn KM,Radbruch H,Rhys H,Rodrigo MB,Romagnani C,Saggau C,Sakaguchi S,Sallusto F,Sanderink L,Sandrock I,Schauer C,Scheffold A,Scherer HU,Schiemann M,Schildberg FA,Schober K,Schoen J,Schuh W,Schüler T,Schulz AR,Schulz S,Schulze J,Simonetti S,Singh J,Sitnik KM,Stark R,Starossom S,Stehle C,Szelinski F,Tan L,Tarnok A,Tornack J,Tree TIM,van Beek JJP,van de Veen W,van Gisbergen K,Vasco C,Verheyden NA,von Borstel A,Ward-Hartstonge KA,Warnatz K,Waskow C,Wiedemann A,Wilharm A,Wing J,Wirz O,Wittner J,Yang JHM,Yang J
17-1031-82 was used in Flow cytometry/Cell sorting to provide the key aspects to consider when performing flow cytometry experiments.
Wed Dec 01 00:00:00 UTC 2021
STING Activated Tumor-Intrinsic Type I Interferon Signaling Promotes CXCR3 Dependent Antitumor Immunity in Pancreatic Cancer.
Cellular and molecular gastroenterology and hepatology
Vonderhaar EP,Barnekow NS,McAllister D,McOlash L,Eid MA,Riese MJ,Tarakanova VL,Johnson BD,Dwinell MB
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Mon Jan 24 00:00:00 UTC 2022
Dendritic cell entry to lymphatic capillaries is orchestrated by CD44 and the hyaluronan glycocalyx.
Life science alliance
Johnson LA,Banerji S,Lagerholm BC,Jackson DG
17-1031 was used in Flow cytometry/Cell sorting to show that hyaluronan is anchored and spatially organized into a 400-500 nm dense glycocalyx by the leukocyte receptor CD44.
Sat May 01 00:00:00 UTC 2021
Myeloid cell subsets that express latency-associated peptide promote cancer growth by modulating T cells.
iScience
Gabriely G,Ma D,Siddiqui S,Sun L,Skillin NP,Abou-El-Hassan H,Moreira TG,Donnelly D,da Cunha AP,Fujiwara M,Walton LR,Patel A,Krishnan R,Levine SS,Healy BC,Rezende RM,Murugaiyan G,Weiner HL
17-1031-82 was used in Flow cytometry/Cell sorting to identify a previously unknown function by which LAPHi MCs promote tumor growth and offer therapeutic intervention to target these cells in cancer.
Fri Nov 19 00:00:00 UTC 2021
Microbiota triggers STING-type I IFN-dependent monocyte reprogramming of the tumor microenvironment.
Cell
Lam KC,Araya RE,Huang A,Chen Q,Di Modica M,Rodrigues RR,Lopès A,Johnson SB,Schwarz B,Bohrnsen E,Cogdill AP,Bosio CM,Wargo JA,Lee MP,Goldszmid RS
17-1031-82 was used in Flow cytometry/Cell sorting to uncover a mechanistic link between the microbiota and the innate TME that can be harnessed to improve cancer therapies.
Thu Oct 14 00:00:00 UTC 2021
Macrophages Maintain Epithelium Integrity by Limiting Fungal Product Absorption.
Cell
Chikina AS,Nadalin F,Maurin M,San-Roman M,Thomas-Bonafos T,Li XV,Lameiras S,Baulande S,Henri S,Malissen B,Lacerda Mariano L,Barbazan J,Blander JM,Iliev ID,Matic Vignjevic D,Lennon-Duménil AM
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Thu Oct 15 00:00:00 UTC 2020
Dermal Adipocyte Lipolysis and Myofibroblast Conversion Are Required for Efficient Skin Repair.
Cell stem cell
Shook BA,Wasko RR,Mano O,Rutenberg-Schoenberg M,Rudolph MC,Zirak B,Rivera-Gonzalez GC,López-Giráldez F,Zarini S,Rezza A,Clark DA,Rendl M,Rosenblum MD,Gerstein MB,Horsley V
17-1031-82 was used in Flow Cytometry to identify an essential role for adipocyte lipolysis in regulating inflammation and repair after injury in skin.
Thu Jun 04 00:00:00 UTC 2020
Contact-dependent delivery of IL-2 by dendritic cells to CD4 T cells in the contraction phase promotes their long-term survival.
Protein & cell
Tong D,Zhang L,Ning F,Xu Y,Hu X,Shi Y
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Sat Feb 01 00:00:00 UTC 2020
Inhibition of the Wnt/β-catenin pathway enhances antitumor immunity in ovarian cancer.
Therapeutic advances in medical oncology
Doo DW,Meza-Perez S,Londoño AI,Goldsberry WN,Katre AA,Boone JD,Moore DJ,Hudson CT,Betella I,McCaw TR,Gangrade A,Bao R,Luke JJ,Yang ES,Birrer MJ,Starenki D,Cooper SJ,Buchsbaum DJ,Norian LA,Randall TD,Arend RC
17-1031 was used in Flow cytometry/Cell sorting to determine whether WNT974 would impair tumor growth, affect gene expression patterns, and improve the immune response in human and murine ovarian cancer models.
Thu Apr 14 00:00:00 UTC 2022
Inhibition of the Wnt/β-catenin pathway enhances antitumor immunity in ovarian cancer.
Therapeutic advances in medical oncology
Doo DW,Meza-Perez S,Londoño AI,Goldsberry WN,Katre AA,Boone JD,Moore DJ,Hudson CT,Betella I,McCaw TR,Gangrade A,Bao R,Luke JJ,Yang ES,Birrer MJ,Starenki D,Cooper SJ,Buchsbaum DJ,Norian LA,Randall TD,Arend RC
17-1031 was used in Flow cytometry/Cell sorting to determine whether WNT974 would impair tumor growth, affect gene expression patterns, and improve the immune response in human and murine ovarian cancer models.
Thu Apr 14 00:00:00 UTC 2022
Human and Mouse Transcriptome Profiling Identifies Cross-Species Homology in Pulmonary and Lymph Node Mononuclear Phagocytes.
Cell reports
Leach SM,Gibbings SL,Tewari AD,Atif SM,Vestal B,Danhorn T,Janssen WJ,Wager TD,Jakubzick CV
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Tue Nov 03 00:00:00 UTC 2020
Inhibiting WNT Ligand Production for Improved Immune Recognition in the Ovarian Tumor Microenvironment.
Cancers
Goldsberry WN,Meza-Perez S,Londoño AI,Katre AA,Mott BT,Roane BM,Goel N,Wall JA,Cooper SJ,Norian LA,Randall TD,Birrer MJ,Arend RC
17-1031 was used in Flow cytometry/Cell sorting to study Wnt activity within the microenvironment of ovarian tumors.
Tue Mar 24 00:00:00 UTC 2020
Ovarian granulosa cell tumor characterization identifies FOXL2 as an immunotherapeutic target.
JCI insight
Pierini S,Tanyi JL,Simpkins F,George E,Uribe-Herranz M,Drapkin R,Burger R,Morgan MA,Facciabene A
17-1031 was used in Flow cytometry/Cell sorting to study can serve as a foundation for trials testing immunotherapeutic approaches in patients with ovarian GCT.
Thu Aug 20 00:00:00 UTC 2020
TLR9 Sensing of Self-DNA Controls Cell-Mediated Immunity to Listeria Infection via Rapid Conversion of Conventional CD4+ T Cells to Treg.
Cell reports
Dolina JS,Lee J,Griswold RQ,Labarta-Bajo L,Kannan S,Greenbaum JA,Bahia El Idrissi N,Pont MJ,Croft M,Schoenberger SP
17-1031 was used in Flow cytometry/Cell sorting to resolve long-standing questions regarding the conditional requirement for TH amongst pathogens and reveal a remarkable degree of plasticity in the function of CD4+ T cells, which can be quickly converted to Tregin vivo by infection-mediated immune modulation.
Tue Apr 07 00:00:00 UTC 2020
Inflammation-Induced Lactate Leads to Rapid Loss of Hepatic Tissue-Resident NK Cells.
Cell reports
Dodard G,Tata A,Erick TK,Jaime D,Miah SMS,Quatrini L,Escalière B,Ugolini S,Vivier E,Brossay L
17-1031 was used in Flow cytometry/Cell sorting to characterize the kinetics and immune response of each of the hepatic NK cell populations to viral challenge and other stimuli.
Tue Jul 07 00:00:00 UTC 2020
Toxin-Triggered Interleukin-1 Receptor Signaling Enables Early-Life Discrimination of Pathogenic versus Commensal Skin Bacteria.
Cell host & microbe
Leech JM,Dhariwala MO,Lowe MM,Chu K,Merana GR,Cornuot C,Weckel A,Ma JM,Leitner EG,Gonzalez JR,Vasquez KS,Diep BA,Scharschmidt TC
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Wed Dec 11 00:00:00 UTC 2019
Toxin-Triggered Interleukin-1 Receptor Signaling Enables Early-Life Discrimination of Pathogenic versus Commensal Skin Bacteria.
Cell host & microbe
Leech JM,Dhariwala MO,Lowe MM,Chu K,Merana GR,Cornuot C,Weckel A,Ma JM,Leitner EG,Gonzalez JR,Vasquez KS,Diep BA,Scharschmidt TC
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Wed Dec 11 00:00:00 UTC 2019
Engulfment of mast cell secretory granules on skin inflammation boosts dendritic cell migration and priming efficiency.
The Journal of allergy and clinical immunology
Dudeck J,Froebel J,Kotrba J,Lehmann CHK,Dudziak D,Speier S,Nedospasov SA,Schraven B,Dudeck A
17-1031 was used in Flow cytometry/Cell sorting to show peripheral mast cells can modify dendritic cell functionality to influence lymphoid tissue-borne adaptive immunity.
Wed May 01 00:00:00 UTC 2019
CSF1R antagonism limits local restimulation of antiviral CD8+ T cells during viral encephalitis.
Journal of neuroinflammation
Funk KE,Klein RS
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Thu Jan 31 00:00:00 UTC 2019
Virus-specific memory T cells populate tumors and can be repurposed for tumor immunotherapy.
Nature communications
Rosato PC,Wijeyesinghe S,Stolley JM,Nelson CE,Davis RL,Manlove LS,Pennell CA,Blazar BR,Chen CC,Geller MA,Vezys V,Masopust D
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Mon Feb 04 00:00:00 UTC 2019
Qa-1-Restricted CD8+ T Cells Can Compensate for the Absence of Conventional T Cells during Viral Infection.
Cell reports
Anderson CK,Reilly EC,Lee AY,Brossay L
17-1031 was used in Flow cytometry/Cell sorting to study compensatory mechanisms in immune responses to viruses when classical T cell immunity has been evaded.
Tue Apr 09 00:00:00 UTC 2019
Interleukin-27 Is Essential for Type 1 Diabetes Development and Sjögren Syndrome-like Inflammation.
Cell reports
Ciecko AE,Foda B,Barr JY,Ramanathan S,Atkinson MA,Serreze DV,Geurts AM,Lieberman SM,Chen YG
17-1031-82 was used in Flow Cytometry to reveal that IL-27 contributes to autoimmunity in NOD mice through multiple mechanisms and provides substantial evidence to support its pathogenic role in human T1D.
Tue Dec 03 00:00:00 UTC 2019
Noc4L-Mediated Ribosome Biogenesis Controls Activation of Regulatory and Conventional T Cells.
Cell reports
Zhu X,Zhang W,Guo J,Zhang X,Li L,Wang T,Yan J,Zhang F,Hou B,Gao N,Gao GF,Zhou X
17-1031 was used in Flow cytometry/Cell sorting to identify the control of Treg activation and immune tolerance maintenance is due to Noc4L mediated ribosome biogenesis.
Tue Apr 23 00:00:00 UTC 2019
CNS lymphatic drainage and neuroinflammation are regulated by meningeal lymphatic vasculature.
Nature neuroscience
Louveau A,Herz J,Alme MN,Salvador AF,Dong MQ,Viar KE,Herod SG,Knopp J,Setliff JC,Lupi AL,Da Mesquita S,Frost EL,Gaultier A,Harris TH,Cao R,Hu S,Lukens JR,Smirnov I,Overall CC,Oliver G,Kipnis J
17-1031 was used in Flow cytometry/Cell sorting to demonstrate that meningeal lymphatics fulfill two critical criteria: they assist in the drainage of cerebrospinal fluid components and enable immune cells to enter draining lymph nodes in a CCR7-dependent manner.
Mon Oct 01 00:00:00 UTC 2018
Irg1 expression in myeloid cells prevents immunopathology during M. tuberculosis infection.
The Journal of experimental medicine
Nair S,Huynh JP,Lampropoulou V,Loginicheva E,Esaulova E,Gounder AP,Boon ACM,Schwarzkopf EA,Bradstreet TR,Edelson BT,Artyomov MN,Stallings CL,Diamond MS
17-1031 was used in Flow cytometry/Cell sorting to study the role of Immune responsive gene 1 in the modulation of inflammation during Mycobacterium tuberculosis infection.
Mon Apr 02 00:00:00 UTC 2018
Non-redundant Functions of IL-6 Produced by Macrophages and Dendritic Cells in Allergic Airway Inflammation.
Frontiers in immunology
Gubernatorova EO,Gorshkova EA,Namakanova OA,Zvartsev RV,Hidalgo J,Drutskaya MS,Tumanov AV,Nedospasov SA
17-1031 was used in Flow cytometry/Cell sorting to indicate that IL-6 plays a pathogenic role in the HDM-induced asthma model and that lung macrophages and dendritic cells are the predominant sources of pathogenic IL-6 but contribute differently to the disease.
Wed Oct 02 00:00:00 UTC 2019
NLRP12 attenuates colon inflammation by maintaining colonic microbial diversity and promoting protective commensal bacterial growth.
Nature immunology
Chen L,Wilson JE,Koenigsknecht MJ,Chou WC,Montgomery SA,Truax AD,Brickey WJ,Packey CD,Maharshak N,Matsushima GK,Plevy SE,Young VB,Sartor RB,Ting JP
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Mon May 01 00:00:00 UTC 2017
NLRP12 attenuates colon inflammation by maintaining colonic microbial diversity and promoting protective commensal bacterial growth.
Nature immunology
Chen L,Wilson JE,Koenigsknecht MJ,Chou WC,Montgomery SA,Truax AD,Brickey WJ,Packey CD,Maharshak N,Matsushima GK,Plevy SE,Young VB,Sartor RB,Ting JP
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Mon May 01 00:00:00 UTC 2017
Deletion of PIKfyve alters alveolar macrophage populations and exacerbates allergic inflammation in mice.
The EMBO journal
Kawasaki T,Ito K,Miyata H,Akira S,Kawai T
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Wed Jun 14 00:00:00 UTC 2017
Anti-CD137 Suppresses Tumor Growth by Blocking Reverse Signaling by CD137 Ligand.
Cancer research
Kang SW,Lee SC,Park SH,Kim J,Kim HH,Lee HW,Seo SK,Kwon BS,Cho HR,Kwon B
17-1031-82 was used in Flow Cytometry to report findings related to reverse signaling by CD137 ligand (CD137L) in antigen-presenting dendritic cells (DC) in tumors that address these paradoxical results.
Wed Nov 01 00:00:00 UTC 2017
Intestinal Batf3-dependent dendritic cells are required for optimal antiviral T-cell responses in adult and neonatal mice.
Mucosal immunology
Sun T,Rojas OL,Li C,Ward LA,Philpott DJ,Gommerman JL
17-1031 was used in Flow cytometry/Cell sorting to study the dendritic cell subset(s) that provoke antiviral responses within the gut.
Mon May 01 00:00:00 UTC 2017
Precision-cut Mouse Lung Slices to Visualize Live Pulmonary Dendritic Cells.
Journal of visualized experiments : JoVE
Lyons-Cohen MR,Thomas SY,Cook DN,Nakano H
17-1031 was used in in vivo experiments to contribute to a comprehensive understanding of cellular events that underlie allergic and inflammatory diseases of the lung.
Wed Apr 05 00:00:00 UTC 2017
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Thu Oct 06 00:00:00 UTC 2016
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Mon Aug 15 00:00:00 UTC 2016
Developmental Progression and Interrelationship of Central and Effector Regulatory T Cell Subsets.
Journal of immunology (Baltimore, Md. : 1950)
Toomer KH,Yuan X,Yang J,Dee MJ,Yu A,Malek TR
17-1031 was used in Flow cytometry/Cell sorting to elucidate the developmental progression and interrelationship of central and effector regulatory T cell subsets.
Sun May 01 00:00:00 UTC 2016
Diet-dependent, microbiota-independent regulation of IL-10-producing lamina propria macrophages in the small intestine.
Scientific reports
Ochi T,Feng Y,Kitamoto S,Nagao-Kitamoto H,Kuffa P,Atarashi K,Honda K,Teitelbaum DH,Kamada N
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Wed Jun 15 00:00:00 UTC 2016
Ablating the aryl hydrocarbon receptor (AhR) in CD11c+ cells perturbs intestinal epithelium development and intestinal immunity.
Scientific reports
Chng SH,Kundu P,Dominguez-Brauer C,Teo WL,Kawajiri K,Fujii-Kuriyama Y,Mak TW,Pettersson S
17-1031 was used in Flow cytometry/Cell sorting to explore the contribution of the Aryl hydrocarbon Receptor signalling pathway to the regulation of innate immunity.
Tue Apr 12 00:00:00 UTC 2016
Cholera Toxin Promotes Th17 Cell Differentiation by Modulating Expression of Polarizing Cytokines and the Antigen-Presenting Potential of Dendritic Cells.
PloS one
Kang JO,Lee JB,Chang J
17-1031 was used in Flow cytometry/Cell sorting to determine how cholera toxin skews the differentiation of CD4 T cells to form IL-17-producing Th17 cells.
Tue Aug 08 00:00:00 UTC 2017
Tetanus toxoid and CCL3 improve dendritic cell vaccines in mice and glioblastoma patients.
Nature
Mitchell DA,Batich KA,Gunn MD,Huang MN,Sanchez-Perez L,Nair SK,Congdon KL,Reap EA,Archer GE,Desjardins A,Friedman AH,Friedman HS,Herndon JE,Coan A,McLendon RE,Reardon DA,Vredenburgh JJ,Bigner DD,Sampson JH
17-1031 was used in Flow cytometry/Cell sorting to show that pre-conditioning with a potent recall antigen significantly improves the homing ability and efficacy of tumour-antigen-specific dendritic cells.
Thu Mar 19 00:00:00 UTC 2015
Altered lymph node composition in diphtheria toxin receptor-based mouse models to ablate dendritic cells.
Journal of immunology (Baltimore, Md. : 1950)
van Blijswijk J,Schraml BU,Rogers NC,Whitney PG,Zelenay S,Acton SE,Reis e Sousa C
17-1031 was used in Flow cytometry/Cell sorting to report that mice in which DTR is expressed on DCs display lymph node hypocellularity and reduced frequency of DCs in the same organs.
Thu Jan 01 00:00:00 UTC 2015
Eosinophils from Murine Lamina Propria Induce Differentiation of Naïve T Cells into Regulatory T Cells via TGF-β1 and Retinoic Acid.
PloS one
Chen HH,Sun AH,Ojcius DM,Hu WL,Ge YM,Lin X,Li LJ,Pan JP,Yan J
17-1031 was used in Flow cytometry/Cell sorting to investigate mechanisms that induce Tregs, showing that eosinophils from murine lamina propria induce differentiation via TGF-β1 and retinoic acid.
Fri Jun 17 00:00:00 UTC 2016
Immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Nature medicine
Clatworthy MR,Aronin CE,Mathews RJ,Morgan NY,Smith KG,Germain RN
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Mon Dec 01 00:00:00 UTC 2014
Identification of a unique TGF-β-dependent molecular and functional signature in microglia.
Nature neuroscience
Butovsky O,Jedrychowski MP,Moore CS,Cialic R,Lanser AJ,Gabriely G,Koeglsperger T,Dake B,Wu PM,Doykan CE,Fanek Z,Liu L,Chen Z,Rothstein JD,Ransohoff RM,Gygi SP,Antel JP,Weiner HL
17-1031 was used in Flow cytometry/Cell sorting to study gene expression in microglia versus myeloid and other immune cells.
Wed Jan 01 00:00:00 UTC 2014
CX₃CR1⁺ mononuclear phagocytes support colitis-associated innate lymphoid cell production of IL-22.
The Journal of experimental medicine
Longman RS,Diehl GE,Victorio DA,Huh JR,Galan C,Miraldi ER,Swaminath A,Bonneau R,Scherl EJ,Littman DR
17-1031 was used in Flow cytometry/Cell sorting to study the role of CX3CR1+ mononuclear phagocytes in the development of colitis and disease progression following Citrobacter rodentium infection.
Mon Jul 28 00:00:00 UTC 2014
Intravascular staining for discrimination of vascular and tissue leukocytes.
Nature protocols
Anderson KG,Mayer-Barber K,Sung H,Beura L,James BR,Taylor JJ,Qunaj L,Griffith TS,Vezys V,Barber DL,Masopust D
17-1031 was used in Flow cytometry/Cell sorting to outline a protocol for the validation and use of intravascular staining to define innate and adaptive immune cells in mice.
Wed Jan 01 00:00:00 UTC 2014
CD31 is a key coinhibitory receptor in the development of immunogenic dendritic cells.
Proceedings of the National Academy of Sciences of the United States of America
Clement M,Fornasa G,Guedj K,Ben Mkaddem S,Gaston AT,Khallou-Laschet J,Morvan M,Nicoletti A,Caligiuri G
17-1031 was used in Flow cytometry/Cell sorting to investigate the effect exerted by CD31 signalling in dendritic cells.
Tue Mar 25 00:00:00 UTC 2014
GM-CSF-licensed CD11b+ lung dendritic cells orchestrate Th2 immunity to Blomia tropicalis.
Journal of immunology (Baltimore, Md. : 1950)
Zhou Q,Ho AW,Schlitzer A,Tang Y,Wong KH,Wong FH,Chua YL,Angeli V,Mortellaro A,Ginhoux F,Kemeny DM
17-1031 was used in Flow cytometry/Cell sorting to identify GM-CSF-licensed CD11b(+) lung DCs as a key component for induction of Th2 responses.
Tue Jul 15 00:00:00 UTC 2014
Lung dendritic cells undergo maturation and polarization towards a T helper type 2-stimulating phenotype in a mouse model of asthma: Role of nerve growth factor.
Experimental and therapeutic medicine
Qin Q,Wang Z,Pan P,Cao Z,Xia Q,Tan H,Hu C
Published figure using CD103 (Integrin alpha E) monoclonal antibody (Product # 17-1031-82) in Flow Cytometry
Sat Nov 01 00:00:00 UTC 2014
Dendritic cell subset distributions in the aorta in healthy and atherosclerotic mice.
PloS one
Busch M,Westhofen TC,Koch M,Lutz MB,Zernecke A
17-1031 was used in Flow cytometry/Cell sorting to perform a systematic analysis of dendritic cells and their accumulation characteristics during atherogenesis.
Tue Oct 21 00:00:00 UTC 2014
Therapeutic evaluation of ex vivo-generated versus natural regulatory T-cells in a mouse model of chronic gut inflammation.
Inflammatory bowel diseases
Karlsson F,Martinez NE,Gray L,Zhang S,Tsunoda I,Grisham MB
17-1031 was used in Flow cytometry/Cell sorting to study the effect of ex vivo-generated and isolated Treg cells on a mouse model of chronic colitis, showing that ex-vivo Tregs were more potent at attenuating colitis, likely due to the associated decrease in IL-6 and IL-17A expression in the colon.
Tue Oct 01 00:00:00 UTC 2013
Mechanism of action of inhibition of allergic immune responses by a novel antedrug TLR7 agonist.
Journal of immunology (Baltimore, Md. : 1950)
Matsui H,Tomizawa H,Eiho K,Kashiwazaki Y,Edwards S,Biffen M,Bell JP,Bahl A,Leishman AJ,Murray CM,Takaku H,Ueda Y
17-1031 was used in Flow cytometry/Cell sorting to characterise the mechanism of action at the cytokine and cellular level involved in the suppression of Th2 allergic responses.
Sat Dec 01 00:00:00 UTC 2012
Vitamin A deficiency impairs vaccine-elicited gastrointestinal immunity.
Journal of immunology (Baltimore, Md. : 1950)
Kaufman DR,De Calisto J,Simmons NL,Cruz AN,Villablanca EJ,Mora JR,Barouch DH
17-1031 was used in Flow cytometry/Cell sorting to suggest that oral vitamin A supplementation may be important for optimizing the success of vaccines against HIV-1 and other mucosal pathogens in the developing world.
Mon Aug 15 00:00:00 UTC 2011
Localization of intestinal intraepithelial T lymphocytes involves regulation of alphaEbeta7 expression by transforming growth factor-beta.
International immunology
Suzuki R,Nakao A,Kanamaru Y,Okumura K,Ogawa H,Ra C
Mon Apr 01 00:00:00 UTC 2002