Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects
              • Instrument Management

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CD103 (Integrin alpha E) Antibodies

            Invitrogen

            CD103 (Integrin alpha E) Monoclonal Antibody (2E7), PerCP-eFluor™ 710, eBioscience™

            18 Published Figures
            23 References
            View all (77) CD103 (Integrin alpha E) antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Promotion Available
            Learn more
            Catalog # 46-1031-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (HKD)
            4,321.00
             
            Check your price & availability
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite CD103 (Integrin alpha E) Monoclonal Antibody (2E7), PerCP-eFluor™ 710, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (18)
            Application
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 19

            CD103 (Integrin alpha E) Antibody (46-1031-82) in Flow

            Staining of C57Bl/6 lymph node cells with Anti-Mouse CD3e FITC (Product # 11-0031-82) and 0.25 µg of Armenian Hamster IgG Isotype Control PerCP-eFluor® 710 (Product # 46-4888-82) (left) or 0.25 µg of Anti-Mouse CD103 (Integrin alpha E) PerCP-eFluor® 710 (right). Cells in the lymphocyte gate were used for analysis.
            View Product

            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)
            CD103 (Integrin alpha E) Antibody in Flow Cytometry (Flow)

            Product Details

            46-1031-82

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (Frozen) (IHC (F))

            -
            View 1 publication 1 publication

            Flow Cytometry (Flow)

            0.5 µg/test
            View 22 publications 22 publications
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Human, Mouse

            Host/Isotype

            Armenian hamster / IgG

            Recommended Isotype Control

            Armenian Hamster IgG Isotype Control (eBio299Arm), PerCP-eFluor™ 710, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            2E7

            Conjugate

            PerCP-eFluor™ 710
            • Unconjugated
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 563
            • Brilliant UV 615
            • Brilliant UV 737
            • Brilliant Violet 421
            • Brilliant Violet 480
            • Brilliant Violet 711
            • eFluor 450
            • FITC
            • Functional Grade
            • NovaFluor Blue 510
            • PE 
            • PE-eFluor 610
            • Super Bright 600
            • Request custom conjugation

            Excitation/Emission Max

            482/708 nm View spectra spectra

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2573704

            Product Specific Information

            Description: The 2E7 monoclonal antibody reacts with mouse CD103, the alphaE integrin. CD103 non-covalently associates with integrin beta7 and is expressed on all intraepithelial lymphocytes, a small subset of peripheral lymphocytes, and dendritic epidermal T cells (DEC). Epithelial cell antigen E-cadherin binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium.

            Applications Reported: This 2E7 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This 2E7 antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.

            Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

            Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            CD103 (ITGAE, Integrin alpha E) is molecule that is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. In particular, CD103 includes a 150kDa alpha chain and a 120kDa beta chain. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is part of the integrin family which are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. ITGAE encodes an I-domain-containing alpha integrin that undergoes post-translational cleavage in the extracellular domain, yielding disulfide-linked heavy and light chains. In combination with the beta 7 integrin, CD103 protein forms the E-cadherin binding integrin known as the human mucosal lymphocyte-1 antigen. CD103 is preferentially expressed in human intestinal intraepithelial lymphocytes (IEL), and in addition to a role in adhesion, it may serve as an accessory molecule for IEL activation. Diseases associated with CD103 dysfunction include hairy cell leukemia and splenic marginal zone lymphoma.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Bioinformatics

            Protein Aliases: CD103; DC; DCs; Integrin alpha M290; Integrin alpha-E; Intregrin alpha E2; MGC141996

            View more View less

            Gene Aliases: A530055J10; alpha-E1; alpha-M290; aM290; CD103; Itgae

            View more View less

            UniProt ID: (Mouse) Q60677

            View more View less

            Entrez Gene ID: (Mouse) 16407

            View more View less

            Function(s)

            metal ion binding integrin

            Process(es)

            cell adhesion integrin-mediated signaling pathway
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            hong kong flag icon
            Hong Kong

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-k8qjk:80/100.66.77.209:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0