Product References
Stimulated Human Umbilical Cord Mesenchymal Stem Cells Enhance the Osteogenesis and Cranial Bone Regeneration through IL-32 Mediated P38 Signaling Pathway.
Stem cells international
Zhang X,Zheng Y,Wang G,Liu Y,Wang Y,Jiang X,Liang Y,Zhao X,Li P,Zhang Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2024
Generation of Highly Functional Hepatocyte-like Organoids from Human Adipose-Derived Mesenchymal Stem Cells Cultured with Endothelial Cells.
Cells
Chen S,Saito Y,Waki Y,Ikemoto T,Teraoku H,Yamada S,Morine Y,Shimada M
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Mar 20 00:00:00 UTC 2024
Development of a membrane and a bilayer of chitosan, gelatin, and polyhydroxybutyrate to be used as wound dressing for the regeneration of rat excisional wounds.
Journal of biomedical materials research. Part A
Barbosa JL,de Melo MIA,da Silva Cunha P,de Miranda MC,Barrioni BR,Moreira CDF,da Fonseca Ferreira A,Arantes RME,de Sá MA,de Magalhães Pereira M,Rodrigues MA,Novikoff S,Gomes DA,de Goes AM
12-1057-42 was used in Flow Cytometry to demonstrate the potential of these new biomaterials as dressings in wound healing studies, favoring tissue regeneration.
Mon Jan 01 00:00:00 UTC 2024
Comparison of retinal degeneration treatment with four types of different mesenchymal stem cells, human induced pluripotent stem cells and RPE cells in a rat retinal degeneration model.
Journal of translational medicine
Liu Q,Liu J,Guo M,Sung TC,Wang T,Yu T,Tian Z,Fan G,Wu W,Higuchi A
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Dec 14 00:00:00 UTC 2023
Enhanced chondrogenic differentiation of iPS cell-derived mesenchymal stem/stromal cells via neural crest cell induction for hyaline cartilage repair.
Frontiers in cell and developmental biology
Zujur D,Al-Akashi Z,Nakamura A,Zhao C,Takahashi K,Aritomi S,Theoputra W,Kamiya D,Nakayama K,Ikeya M
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Sun May 28 00:00:00 UTC 2023
Characteristics of multilineage-differentiating stress-enduring cell clusters in different culture conditions.
Skin research and technology : official journal of International Society for Bioengineering and the Skin (ISBS) [and] International Society for Digital Imaging of Skin (ISDIS) [and] International Society for Skin Imaging (ISSI)
Ning J,Cao YY,Zhang RZ,Li Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2023
Exercise combined with administration of adipose-derived stem cells ameliorates neuropathic pain after spinal cord injury.
Neural regeneration research
Cheng X,Mao GP,Hu WJ,Yu ZR,Xu YY,Chen W,Li X,Zeng XL,Zhang WW,Chen JW,Wan Y,Wang L
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
Single-cell analysis of immune cells on gingiva-derived mesenchymal stem cells in experimental autoimmune uveitis.
iScience
Gao Y,Duan R,Li H,Jiang L,Tao T,Liu X,Zhu L,Li Z,Chen B,Zheng S,Lin X,Su W
12-1057-42 was used in Flow cytometry/Cell sorting to generate a lymph node single-cell transcriptomic atlas of GMSC-treated experimental autoimmune uveitis mice.
Fri May 19 00:00:00 UTC 2023
Stromal heterogeneity may explain increased incidence of metaplastic breast cancer in women of African descent.
Nature communications
Kumar B,Khatpe AS,Guanglong J,Batic K,Bhat-Nakshatri P,Granatir MM,Addison RJ,Szymanski M,Baldridge LA,Temm CJ,Sandusky G,Althouse SK,Cote ML,Miller KD,Storniolo AM,Nakshatri H
12-1057-42 was used in Flow Cytometry to demonstrate that these cells have properties of fibroadipogenic/mesenchymal stromal cells that express PROCR and PDGFRα and transdifferentiate into adipogenic and osteogenic lineages.
Thu Sep 14 00:00:00 UTC 2023
Carboxymethyl chitin or chitosan for osteoinduction effect on the human periodontal ligament stem cells.
Dental materials journal
Fan C,Li Z,Ji Q,Sun H,Liang Y,Yang P
12-1057-42 was used in Flow Cytometry to elucidate carboxymethyl chitosan (CMCTS) and carboxymethyl chitin (CMCT) effect on osteogenic differentiation.
Tue May 31 00:00:00 UTC 2022
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Mar 03 00:00:00 UTC 2022
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Mar 03 00:00:00 UTC 2022
Production enhancement of human adipose-derived mesenchymal stem cells by low-intensity ultrasound stimulation.
Scientific reports
Min S,Byeon Y,Kim M,Lee Y,Lee SH,Lee Y,Farooqi HMU,Lee HK,Paeng DG
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Dec 21 00:00:00 UTC 2022
Isolation and characterization mesenchymal stem cells from red panda (Ailurus fulgens styani) endometrium.
Conservation physiology
Wang DH,Wu XM,Chen JS,Cai ZG,An JH,Zhang MY,Li Y,Li FP,Hou R,Liu YL
12-1057-42 was used in Flow Cytometry to show that the new multipotential stem cell could be beneficial not only for the germ plasm resources conservation of red panda, but also for basic or pre-clinical studies in the future.
Sat Jan 01 00:00:00 UTC 2022
Induced Neurodifferentiation of hBM-MSCs through Activation of the ERK/CREB Pathway via Pulsed Electromagnetic Fields and Physical Stimulation Promotes Neurogenesis in Cerebral Ischemic Models.
International journal of molecular sciences
Park HJ,Choi JH,Nam MH,Seo YK
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Jan 21 00:00:00 UTC 2022
Mesenchymal stem cells derived from patients with premature aging syndromes display hallmarks of physiological aging.
Life science alliance
Trani JP,Chevalier R,Caron L,El Yazidi C,Broucqsault N,Toury L,Thomas M,Annab K,Binetruy B,De Sandre-Giovannoli A,Levy N,Magdinier F,Robin JD
12-1057-42 was used in Flow cytometry/Cell sorting to report the presence of an altered mitochondrial pattern; an increased response to double-strand DNA damage; and telomere erosion in HGPS, HGPS-L, and APS MSCs, suggesting converging pathways, independent of progerin accumulation, but a distinct DNA methylation profile in HGPS and HGPS-L compared with APS cells.
Wed Sep 14 00:00:00 UTC 2022
Mesenchymal stem cells derived from patients with premature aging syndromes display hallmarks of physiological aging.
Life science alliance
Trani JP,Chevalier R,Caron L,El Yazidi C,Broucqsault N,Toury L,Thomas M,Annab K,Binetruy B,De Sandre-Giovannoli A,Levy N,Magdinier F,Robin JD
12-1057-42 was used in Flow cytometry/Cell sorting to report the presence of an altered mitochondrial pattern; an increased response to double-strand DNA damage; and telomere erosion in HGPS, HGPS-L, and APS MSCs, suggesting converging pathways, independent of progerin accumulation, but a distinct DNA methylation profile in HGPS and HGPS-L compared with APS cells.
Wed Sep 14 00:00:00 UTC 2022
Aberrant Expression of COX-2 and FOXG1 in Infrapatellar Fat Pad-Derived ASCs from Pre-Diabetic Donors.
Cells
O'Donnell BT,Monjure TA,Al-Ghadban S,Ives CJ,L'Ecuyer MP,Rhee C,Romero-Lopez M,Li Z,Goodman SB,Lin H,Tuan RS,Bunnell BA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
In-cytoplasm mitochondrial transplantation for mesenchymal stem cells engineering and tissue regeneration.
Bioengineering & translational medicine
Yao X,Ma Y,Zhou W,Liao Y,Jiang Z,Lin J,He Q,Wu H,Wei W,Wang X,Björklund M,Ouyang H
12-1057-42 was used in Flow Cytometry to develop a strategy for energetic engineering of mesenchymal stem cells by transfer of exogeneous mitochondria.
Sat Jan 01 00:00:00 UTC 2022
Neuronal Cell Differentiation of Human Dental Pulp Stem Cells on Synthetic Polymeric Surfaces Coated With ECM Proteins.
Frontiers in cell and developmental biology
Gao Y,Tian Z,Liu Q,Wang T,Ban LK,Lee HH,Umezawa A,Almansour AI,Arumugam N,Kumar RS,Ye Q,Higuchi A,Chen H,Sung TC
12-1057-42 was used in Flow cytometry/Cell sorting to investigate the effect of cell culture biomaterials on the differentiation of dental pulp stem cells (DPSCs) into neuronal cells.
Sat Jul 16 00:00:00 UTC 2022
Mechanical Compression by Simulating Orthodontic Tooth Movement in an In Vitro Model Modulates Phosphorylation of AKT and MAPKs via TLR4 in Human Periodontal Ligament Cells.
International journal of molecular sciences
Roth CE,Craveiro RB,Niederau C,Malyaran H,Neuss S,Jankowski J,Wolf M
12-1057-42 was used in Flow cytometry/Cell sorting to better understand the cell signaling downstream of key molecules involved in the process of sterile inflammation via TLR4.
Fri Jul 22 00:00:00 UTC 2022
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Tue Aug 31 00:00:00 UTC 2021
Development, characterization, and hematopoietic differentiation of Griscelli syndrome type 2 induced pluripotent stem cells.
Stem cell research & therapy
Güney-Esken G,Erol ÖD,Pervin B,Gürhan Sevinç G,Önder T,Bilgiç E,Korkusuz P,Günel-Özcan A,Uçkan-Çetinkaya D,Aerts-Kaya F
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu May 13 00:00:00 UTC 2021
Human Amniotic Mesenchymal Stem Cells Inhibit aGVHD by Regulating Balance of Treg and T Effector Cells.
Journal of inflammation research
Gao Y,Li W,Bu X,Xu Y,Cai S,Zhong J,Du M,Sun H,Huang L,He Y,Hu X,Liu Q,Jin H,Wang Q,Ping B
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Tue Apr 26 00:00:00 UTC 2022
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Jul 15 00:00:00 UTC 2021
Glycemic control by umbilical cord-derived mesenchymal stem cells promotes effects of fasting-mimicking diet on type 2 diabetic mice.
Stem cell research & therapy
Zhao N,Gao YF,Bao L,Lei J,An HX,Pu FX,Cheng RP,Chen J,Ni H,Sui BD,Ji FP,Hu CH
12-1057-42 was used in Flow Cytometry to establish a strategy that combined UC-MSC infusion and FMD and was effective in treating T2D, which provided potential approaches for developing novel clinical T2D therapies.
Tue Jul 13 00:00:00 UTC 2021
Differentiation potential and mRNA profiles of human dedifferentiated adipose cells and adipose‑derived stem cells from young donors.
Molecular medicine reports
Nie F,Bi H,Zhang C,Ding P
12-1057-42 was used in Flow cytometry/Cell sorting to provide a foundation for further clinical administration of stem cells derived from adipose tissues in therapy.
Fri Jan 01 00:00:00 UTC 2021
ZKSCAN3 counteracts cellular senescence by stabilizing heterochromatin.
Nucleic acids research
Hu H,Ji Q,Song M,Ren J,Liu Z,Wang Z,Liu X,Yan K,Hu J,Jing Y,Wang S,Zhang W,Liu GH,Qu J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Jun 19 00:00:00 UTC 2020
Rescue of premature aging defects in Cockayne syndrome stem cells by CRISPR/Cas9-mediated gene correction.
Protein & cell
Wang S,Min Z,Ji Q,Geng L,Su Y,Liu Z,Hu H,Wang L,Zhang W,Suzuiki K,Huang Y,Zhang P,Tang TS,Qu J,Yu Y,Liu GH,Qiao J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Mon Sep 28 00:00:00 UTC 2020
The Secretome Derived From Mesenchymal Stromal Cells Cultured in a Xeno-Free Medium Promotes Human Cartilage Recovery in vitro.
Frontiers in bioengineering and biotechnology
Palamà MEF,Shaw GM,Carluccio S,Reverberi D,Sercia L,Persano L,Pisignano D,Cortese K,Barry FP,Murphy JM,Gentili C
12-1057-42 was used in Flow Cytometry to investigate the therapeutic potential of conditioned media (CM) and extracellular vesicles (EVs) isolated from hBMSC and grown in a xeno-free culture system (XFS) compared to the conventional fetal bovine serum-culture system (FBS) in an in vitro model of OA.
Mon Sep 28 00:00:00 UTC 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Tue Oct 06 00:00:00 UTC 2020
Resveratrol rescues TNF‑α‑induced inhibition of osteogenesis in human periodontal ligament stem cells via the ERK1/2 pathway.
Molecular medicine reports
Yuan J,Wang X,Ma D,Gao H,Zheng D,Zhang J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
Impairment of endothelial progenitor cells function in patient with mustard gas intoxication.
Inhalation toxicology
Siavashi V,Cheraghi H,Pourmohammad P,Nooshirvani P,Abdolahi S,Solghani A,Nassiri SM,Ghazanfari T
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Sat Feb 01 00:00:00 UTC 2020
Human umbilical cord-derived mesenchymal stem cells and human cord blood mononuclear cells protect against cisplatin-induced acute kidney injury in rat models.
Experimental and therapeutic medicine
Xu Q,Yan P,Duan XJ,Wu X,Chen XJ,Luo M,Peng JC,Feng LX,Liu J,Zhong HL,Cheng W,Zou QY,Duan SB
12-1057-42 was used in Flow Cytometry to determine the protective effects of hUCMSCs and hCBMNCs transplantation therapies on an established CP-induced rat model and explore their underlying mechanisms of action.
Tue Dec 01 00:00:00 UTC 2020
Vasculogenesis from Human Dental Pulp Stem Cells Grown in Matrigel with Fully Defined Serum-Free Culture Media.
Biomedicines
Luzuriaga J,Irurzun J,Irastorza I,Unda F,Ibarretxe G,Pineda JR
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Mon Nov 09 00:00:00 UTC 2020
Vasculogenesis from Human Dental Pulp Stem Cells Grown in Matrigel with Fully Defined Serum-Free Culture Media.
Biomedicines
Luzuriaga J,Irurzun J,Irastorza I,Unda F,Ibarretxe G,Pineda JR
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Mon Nov 09 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
Chondrogenic Differentiation from Induced Pluripotent Stem Cells Using Non-Viral Minicircle Vectors.
Cells
Rim YA,Nam Y,Park N,Jung H,Lee K,Lee J,Ju JH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Sun Mar 01 00:00:00 UTC 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
12-1057 was used in Flow cytometry/Cell sorting to investigate the potential of biologically active compounds, namely, angiotensin II, retinoic acid (RA), epigallocatechin-3-gallate (EGCG), vitamin C alone, and the combinations of RA, EGCG, and vitamin C with angiotensin II to induce cardiomyogenic differentiation of AF-MSCs.
Thu Nov 19 00:00:00 UTC 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
12-1057 was used in Flow cytometry/Cell sorting to investigate the potential of biologically active compounds, namely, angiotensin II, retinoic acid (RA), epigallocatechin-3-gallate (EGCG), vitamin C alone, and the combinations of RA, EGCG, and vitamin C with angiotensin II to induce cardiomyogenic differentiation of AF-MSCs.
Thu Nov 19 00:00:00 UTC 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
12-1057 was used in Flow cytometry/Cell sorting to demonstrate that transplanted MSCs promoted cutaneous wound healing partially through releasing apoptotic bodies which could convert the macrophages towards an anti-inflammatory phenotype that plays a crucial role in the tissue repair process.
Fri Nov 27 00:00:00 UTC 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
12-1057 was used in Flow cytometry/Cell sorting to demonstrate that transplanted MSCs promoted cutaneous wound healing partially through releasing apoptotic bodies which could convert the macrophages towards an anti-inflammatory phenotype that plays a crucial role in the tissue repair process.
Fri Nov 27 00:00:00 UTC 2020
Migration Inhibitory Factor in Conditioned Medium from Human Umbilical Cord Blood-Derived Mesenchymal Stromal Cells Stimulates Hair Growth.
Cells
Oh HA,Kwak J,Kim BJ,Jin HJ,Park WS,Choi SJ,Oh W,Um S
12-1057 was used in Flow cytometry/Cell sorting to suggest that P-CM can improve the efficacy in hair growth via a paracrine mechanism and that MIF in P-CM exerts hair growth-promoting effects via a VEGF-related β-catenin and p-GSK-3β [SER9] signaling pathway.
Thu May 28 00:00:00 UTC 2020
Telomere-dependent and telomere-independent roles of RAP1 in regulating human stem cell homeostasis.
Protein & cell
Zhang X,Liu Z,Liu X,Wang S,Zhang Y,He X,Sun S,Ma S,Shyh-Chang N,Liu F,Wang Q,Wang X,Liu L,Zhang W,Song M,Liu GH,Qu J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Sun Sep 01 00:00:00 UTC 2019
Up-regulation of FOXD1 by YAP alleviates senescence and osteoarthritis.
PLoS biology
Fu L,Hu Y,Song M,Liu Z,Zhang W,Yu FX,Wu J,Wang S,Izpisua Belmonte JC,Chan P,Qu J,Tang F,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu May 13 00:00:00 UTC 2021
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2019
Bone mesenchymal stromal cells exhibit functional inhibition but no chromosomal aberrations in chronic myelogenous leukemia.
Oncology letters
Xie J,Chen J,Wang B,He X,Huang H
12-1057-42 was used in Flow Cytometry to demonstrate functional inhibition of CML-BMSCs; however, no signs of chromosomal aberrations were observed, thereby providing insight into the changes occurring in the CML-BMM.
Tue Jan 01 00:00:00 UTC 2019
Ultrastructural morphology is distinct among primary progenitor cell isolates from normal, inflamed, and cryopreserved equine hoof tissue and CD105+K14+ progenitor cells.
In vitro cellular & developmental biology. Animal
Yang Q,Lopez MJ
12-1057 was used in Flow cytometry/Cell sorting to provide accurate ultrastructural depictions of progenitor cells isolated from inflamed tissue and normal tissue before and after cryopreservation and following selection of cells expressing both keratin (K) 14 (ectodermal) and cluster of differentiation (CD) 105 (mesodermal).
Sun Sep 01 00:00:00 UTC 2019
Ultrastructural morphology is distinct among primary progenitor cell isolates from normal, inflamed, and cryopreserved equine hoof tissue and CD105+K14+ progenitor cells.
In vitro cellular & developmental biology. Animal
Yang Q,Lopez MJ
12-1057 was used in Flow cytometry/Cell sorting to provide accurate ultrastructural depictions of progenitor cells isolated from inflamed tissue and normal tissue before and after cryopreservation and following selection of cells expressing both keratin (K) 14 (ectodermal) and cluster of differentiation (CD) 105 (mesodermal).
Sun Sep 01 00:00:00 UTC 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
12-1057 was used in Flow cytometry/Cell sorting to determine whether primary MenSCs exist in the sedimentation of the deciduous endometrium after DGC and further to evaluate the isolation of MenSCs by direct red blood cell lysis treatment.
Thu May 02 00:00:00 UTC 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
12-1057 was used in Flow cytometry/Cell sorting to determine whether primary MenSCs exist in the sedimentation of the deciduous endometrium after DGC and further to evaluate the isolation of MenSCs by direct red blood cell lysis treatment.
Thu May 02 00:00:00 UTC 2019
Stabilizing heterochromatin by DGCR8 alleviates senescence and osteoarthritis.
Nature communications
Deng L,Ren R,Liu Z,Song M,Li J,Wu Z,Ren X,Fu L,Li W,Zhang W,Guillen P,Izpisua Belmonte JC,Chan P,Qu J,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Jul 26 00:00:00 UTC 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Dec 20 00:00:00 UTC 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2019
Investigating the potential of the secretome of mesenchymal stem cells derived from sickle cell disease patients.
PloS one
Ribeiro TO,Silveira BM,Meira MC,Carreira ACO,Sogayar MC,Meyer R,Fortuna V
12-1057 was used in Flow cytometry/Cell sorting to find that hypoxically conditioned cells presented increased expression and secretion of several well-characterized trophic growth factors (VEGF, IL8, MCP-1, ANG) directly linked to angiogenesis and tissue repair.
Wed Mar 11 00:00:00 UTC 2020
Differential stem cell aging kinetics in Hutchinson-Gilford progeria syndrome and Werner syndrome.
Protein & cell
Wu Z,Zhang W,Song M,Wang W,Wei G,Li W,Lei J,Huang Y,Sang Y,Chan P,Chen C,Qu J,Suzuki K,Belmonte JCI,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2018
Antagonizing CD105 enhances radiation sensitivity in prostate cancer.
Oncogene
Madhav A,Andres A,Duong F,Mishra R,Haldar S,Liu Z,Angara B,Gottlieb R,Zumsteg ZS,Bhowmick NA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Aug 01 00:00:00 UTC 2018
Isolation and characterization of stem cells from differentially degenerated human lumbar zygapophyseal articular cartilage.
Molecular medicine reports
Xiao L,Xu S,Wang X,Jin Z,Wang J,Yang B,Xu H
12-1057 was used in Flow cytometry/Cell sorting to show stem cells with multilineage differentiation potential and clonal properties were identified in human LZAC, and these characteristics were more prominent in mildly degenerated as compared with severely degenerated articular cartilage.
Sat Dec 01 00:00:00 UTC 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
BNIP3/Bcl-2-mediated apoptosis induced by cyclic tensile stretch in human cartilage endplate-derived stem cells.
Experimental and therapeutic medicine
Yuan C,Pu L,He Z,Wang J
12-1057-42 was used in Flow Cytometry to investigate the molecular mechanisms of cyclic stretch-induced apoptosis in human intervertebral disc cartilage endplate-derived stem cells (CESCs).
Mon Jan 01 00:00:00 UTC 2018
ATF6 safeguards organelle homeostasis and cellular aging in human mesenchymal stem cells.
Cell discovery
Wang S,Hu B,Ding Z,Dang Y,Wu J,Li D,Liu X,Xiao B,Zhang W,Ren R,Lei J,Hu H,Chen C,Chan P,Li D,Qu J,Tang F,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Generation of human embryonic stem cell line with heterozygous RB1 deletion by CRIPSR/Cas9 nickase.
Stem cell research
Tu J,Huo Z,Liu M,Wang D,Xu A,Zhou R,Zhu D,Gingold J,Shen J,Zhao R,Lee DF
12-1057-42 was used in Immunocytochemistry to demonstrate generation of a heterozygous deletion of the RB1 gene in the H1 human embryonic stem cell line using CRISPR/Cas9 nickase genome editing.
Sun Apr 01 00:00:00 UTC 2018
The ICAM-1 expression level determines the susceptibility of human endothelial cells to simulated microgravity.
Journal of cellular biochemistry
Buravkova LB,Rudimov EG,Andreeva ER,Grigoriev AI
12-1057-42 was used in Flow cytometry/Cell sorting to study the expression profile of the multi-step cascade of adhesion molecules in umbilical cord endothelial cells.
Thu Mar 01 00:00:00 UTC 2018
Endoglin: a novel target for therapeutic intervention in acute leukemias revealed in xenograft mouse models.
Blood
Dourado KMC,Baik J,Oliveira VKP,Beltrame M,Yamamoto A,Theuer CP,Figueiredo CAV,Verneris MR,Perlingeiro RCR
12-1057 was used in Flow cytometry/Cell sorting to show that endoglin (CD105) is a biomarker for acute leukaemia that is necessary for leukaemogenic activity.
Thu May 04 00:00:00 UTC 2017
Insulin-Like Growth Factor Binding Protein-6 Alters Skeletal Muscle Differentiation of Human Mesenchymal Stem Cells.
Stem cells international
Aboalola D,Han VKM
12-1057 was used in Flow cytometry/Cell sorting to indicate that insulin-like growth factor binding protein-6 influences mesenchymal stem cell pluripotency and myogenic differentiation.
Tue Apr 11 00:00:00 UTC 2023
The Improvement of Respiratory Performance After Phototherapy-Induced EPC Mobilization in Preterm Infants With RDS.
Journal of cellular biochemistry
Siavashi V,Asadian S,Taheri-Asl M,Babaei H,Keshavarz S,Bazaei M,Nassiri SM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
Circulation Enrichment of Functional Endothelial Progenitor Cells by Infantile Phototherapy.
Journal of cellular biochemistry
Siavashi V,Asadian S,Sharifi A,Esmaeilivand M,Norouzinia R,Azadbakht M,Nassiri SM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2017
A high-yield isolation and enrichment strategy for human lung microvascular endothelial cells.
Pulmonary circulation
Gaskill C,Majka SM
12-1057 was used in Flow cytometry/Cell sorting to describe a protocol to select large quantities of enriched primary human lung microvascular endothelial cells, free from mesenchymal contamination.
Wed Mar 01 00:00:00 UTC 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2017
Elimination of undifferentiated human embryonic stem cells by cardiac glycosides.
Scientific reports
Lin YT,Wang CK,Yang SC,Hsu SC,Lin H,Chang FP,Kuo TC,Shen CN,Chiang PM,Hsiao M,Lu FL,Lu J
Thu Jul 13 00:00:00 UTC 2017
Effects of acute exposure to low-dose radiation on the characteristics of human bone marrow mesenchymal stromal/stem cells.
Inflammation and regeneration
Fujishiro A,Miura Y,Iwasa M,Fujii S,Sugino N,Andoh A,Hirai H,Maekawa T,Ichinohe T
12-1057 was used in Flow cytometry/Cell sorting to determine whether low dose irradiation affects the function of mesenchymal stromal/stem cells.
Fri Mar 11 00:00:00 UTC 2022
Feasibility of placenta-derived mesenchymal stem cells as a tool for studying pregnancy-related disorders.
Scientific reports
Fuchi N,Miura K,Doi H,Li TS,Masuzaki H
12-1057 was used in Flow cytometry/Cell sorting to evaluate the feasibility of using placenta-derived mesenchymal stem cells as a tool to study pregnancy related disorders.
Wed Apr 12 00:00:00 UTC 2017
SIRT6 safeguards human mesenchymal stem cells from oxidative stress by coactivating NRF2.
Cell research
Pan H,Guan D,Liu X,Li J,Wang L,Wu J,Zhou J,Zhang W,Ren R,Zhang W,Li Y,Yang J,Hao Y,Yuan T,Yuan G,Wang H,Ju Z,Mao Z,Li J,Qu J,Tang F,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2016
Lymphatic Contribution to the Cellular Niche in Heterotopic Ossification.
Annals of surgery
Loder S,Agarwal S,Sorkin M,Breuler C,Li J,Peterson J,Gardenier J,Hsieh HH,Wang SC,Mehrara BJ,Levi B
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2016
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Tue Jan 19 00:00:00 UTC 2016
NR2F2 regulates bone marrow-derived mesenchymal stem cell-promoted proliferation of Reh cells.
Molecular medicine reports
Zhu N,Wang H,Wei J,Wang B,Shan W,Lai X,Zhao Y,Yu J,Huang H
12-1057 was used in Flow cytometry/Cell sorting to demonstrate NR2F2 mediates bone marrow-derived mesenchymal stem cell induced proliferation of Reh cells via regulation of vascular endothelial growth factor A.
Mon Aug 01 00:00:00 UTC 2016
Angiogenic activity mediates bone repair from human pluripotent stem cell-derived osteogenic cells.
Scientific reports
Zou L,Chen Q,Quanbeck Z,Bechtold JE,Kaufman DS
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Mar 16 00:00:00 UTC 2016
Influence of aging on the quantity and quality of human cardiac stem cells.
Scientific reports
Nakamura T,Hosoyama T,Kawamura D,Takeuchi Y,Tanaka Y,Samura M,Ueno K,Nishimoto A,Kurazumi H,Suzuki R,Ito H,Sakata K,Mikamo A,Li TS,Hamano K
12-1057 was used in Flow cytometry/Cell sorting to compare the growth rate, senescence, DNA damage, and growth factor production of cardiosphere-derived cells from patients of various ages.
Mon Mar 07 00:00:00 UTC 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
12-1057 was used in Flow cytometry/Cell sorting to show MIF plays a role in regulating the differentiation fate of human cartilage endplate stem cells under hypoxia.
Tue Aug 09 00:00:00 UTC 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
Potency of umbilical cord blood- and Wharton's jelly-derived mesenchymal stem cells for scarless wound healing.
Scientific reports
Doi H,Kitajima Y,Luo L,Yan C,Tateishi S,Ono Y,Urata Y,Goto S,Mori R,Masuzaki H,Shimokawa I,Hirano A,Li TS
12-1057 was used in Flow cytometry/Cell sorting to determine that umbilical cord blood offers a potential source of MSCs for therapeutic scarless wound healing.
Tue Jan 05 00:00:00 UTC 2016
Modeling familial cancer with induced pluripotent stem cells.
Cell
Lee DF,Su J,Kim HS,Chang B,Papatsenko D,Zhao R,Yuan Y,Gingold J,Xia W,Darr H,Mirzayans R,Hung MC,Schaniel C,Lemischka IR
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Apr 09 00:00:00 UTC 2015
The role of Toll-like receptor 3 and 4 in regulating the function of mesenchymal stem cells isolated from umbilical cord.
International journal of molecular medicine
Zhang L,Liu D,Pu D,Wang Y,Li L,He Y,Li Y,Li L,Qiu Z,Zhao S,Li W
12-1057-42 was used in Flow Cytometry to examine the role of the TLR3 and TLR4 pathways in modulating the immune properties of human umbilical cord-derived MSCs.
Wed Apr 01 00:00:00 UTC 2015
Adipose-derived Mesenchymal Stem Cells Are Phenotypically Superior for Regeneration in the Setting of Osteonecrosis of the Femoral Head.
Clinical orthopaedics and related research
Wyles CC,Houdek MT,Crespo-Diaz RJ,Norambuena GA,Stalboerger PG,Terzic A,Behfar A,Sierra RJ
12-1057 was used in Flow cytometry/Cell sorting to demonstrate that aMSCs outperform bmMSCs in growth rate and bone differentiation potential in osteonecrosis.
Thu Oct 01 00:00:00 UTC 2015
Impaired function of bone marrow stromal cells in systemic mastocytosis.
Stem cell research
Nemeth K,Wilson TM,Ren JJ,Sabatino M,Stroncek DM,Krepuska M,Bai Y,Robey PG,Metcalfe DD,Mezey E
12-1057 was used in Flow cytometry/Cell sorting to implicate epigenetic change in mesenchymal stem cells as a driver of some of the symptoms associated with systemic mastocytosis.
Wed Jul 01 00:00:00 UTC 2015
Human adipose-derived stem cells partially rescue the stroke syndromes by promoting spatial learning and memory in mouse middle cerebral artery occlusion model.
Stem cell research & therapy
Zhou F,Gao S,Wang L,Sun C,Chen L,Yuan P,Zhao H,Yi Y,Qin Y,Dong Z,Cao L,Ren H,Zhu L,Li Q,Lu B,Liang A,Xu GT,Zhu H,Gao Z,Ma J,Xu J,Chen X
12-1057 was used in Flow cytometry/Cell sorting to demonstrate human adipose derived stem cell (hDAC) intervention with MCAO mice could apparently ameliorate stroke symptoms by direct cell replacement, enhanced immnunosuppression and increasing the viability of endogenous neurons.
Sat May 09 00:00:00 UTC 2015
Human Gingival Integration-Free iPSCs; a Source for MSC-Like Cells.
International journal of molecular sciences
Umezaki Y,Hashimoto Y,Nishishita N,Kawamata S,Baba S
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Mon Jun 15 00:00:00 UTC 2015
Efficacy of autologous stem cell-based therapy for osteonecrosis of the femoral head in sickle cell disease: a five-year follow-up study.
Stem cell research & therapy
Daltro GC,Fortuna V,de Souza ES,Salles MM,Carreira AC,Meyer R,Freire SM,Borojevic R
12-1057 was used in Flow cytometry/Cell sorting to report a significant improvement in clinical joint symptoms and pain relief in patients after bone marrow-derived mononuclear cell implantation.
Fri May 29 00:00:00 UTC 2015
Low-oxygen tension and IGF-I promote proliferation and multipotency of placental mesenchymal stem cells (PMSCs) from different gestations via distinct signaling pathways.
Endocrinology
Youssef A,Iosef C,Han VK
12-1057 was used in Flow cytometry/Cell sorting to demonstrate that low-oxygen tension regulates the fate of PMSCs from early and late gestations in response to IGF-I, both independently and dependently, via specific signal transduction mechanisms.
Tue Apr 01 00:00:00 UTC 2014
Undifferentiated human adipose-derived stromal/stem cells loaded onto wet-spun starch-polycaprolactone scaffolds enhance bone regeneration: nude mice calvarial defect in vivo study.
Journal of biomedical materials research. Part A
Carvalho PP,Leonor IB,Smith BJ,Dias IR,Reis RL,Gimble JM,Gomes ME
12-1057 was used in Flow cytometry/Cell sorting to assess the ossification ability of adipose-derived stem cells.
Mon Sep 01 00:00:00 UTC 2014
Serum- and growth-factor-free three-dimensional culture system supports cartilage tissue formation by promoting collagen synthesis via Sox9-Col2a1 interaction.
Tissue engineering. Part A
Ahmed N,Iu J,Brown CE,Taylor DW,Kandel RA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Aug 01 00:00:00 UTC 2014
Serum- and growth-factor-free three-dimensional culture system supports cartilage tissue formation by promoting collagen synthesis via Sox9-Col2a1 interaction.
Tissue engineering. Part A
Ahmed N,Iu J,Brown CE,Taylor DW,Kandel RA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Fri Aug 01 00:00:00 UTC 2014
Epithelial cell differentiation of human mesenchymal stromal cells in decellularized lung scaffolds.
Tissue engineering. Part A
Mendez JJ,Ghaedi M,Steinbacher D,Niklason LE
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Sun Jun 01 00:00:00 UTC 2014
Differentiation of human adipose-derived stem cells into neuron-like cells which are compatible with photocurable three-dimensional scaffolds.
Tissue engineering. Part A
Gao S,Zhao P,Lin C,Sun Y,Wang Y,Zhou Z,Yang D,Wang X,Xu H,Zhou F,Cao L,Zhou W,Ning K,Chen X,Xu J
12-1057 was used in Immunocytochemistry to investigate the differentiation of human adipose-derived stem cells into neuron-like cells which are compatible with photocurable 3D scaffolds.
Tue Apr 01 00:00:00 UTC 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Thu Dec 11 00:00:00 UTC 2014
Resolving cancer-stroma interfacial signalling and interventions with micropatterned tumour-stromal assays.
Nature communications
Shen K,Luk S,Hicks DF,Elman JS,Bohr S,Iwamoto Y,Murray R,Pena K,Wang F,Seker E,Weissleder R,Yarmush ML,Toner M,Sgroi D,Parekkadan B
12-1057 was used in Immunofluorescence to investigate μTSA as a platform for studying tumour microenvironmental interactions and cancer field effects with applications in drug discovery and development.
Tue Dec 09 00:00:00 UTC 2014
Lipopolysaccharide activates Toll-like receptor 4 (TLR4)-mediated NF-κB signaling pathway and proinflammatory response in human pericytes.
The Journal of biological chemistry
Guijarro-Muñoz I,Compte M,Álvarez-Cienfuegos A,Álvarez-Vallina L,Sanz L
12-1057 was used in Flow cytometry/Cell sorting to suggest pericytes should be considered as active players in the inflammatory cascade with potential physiopathological implications.
Fri Jan 24 00:00:00 UTC 2014
In vitro mesenchymal trilineage differentiation and extracellular matrix production by adipose and bone marrow derived adult equine multipotent stromal cells on a collagen scaffold.
Stem cell reviews and reports
Xie L,Zhang N,Marsano A,Vunjak-Novakovic G,Zhang Y,Lopez MJ
12-1057 was used in Flow cytometry/Cell sorting to evaluate the capability of equine multipotent stromal cells on a type I collagen scaffold to undergo chondrogenic, osteogenic and adipogenic differentiation and form extracellular matrix in vitro.
Sun Dec 01 00:00:00 UTC 2013
Early intervention with gene-modified mesenchymal stem cells overexpressing interleukin-4 enhances anti-inflammatory responses and functional recovery in experimental autoimmune demyelination.
Cell adhesion & migration
Payne NL,Dantanarayana A,Sun G,Moussa L,Caine S,McDonald C,Herszfeld D,Bernard CC,Siatskas C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 12-1057-42) in Flow Cytometry
Wed Dec 19 00:00:00 UTC 2012
Neuroprotective effects of mesenchymal stem cells derived from human embryonic stem cells in transient focal cerebral ischemia in rats.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Liu YP,Seçkin H,Izci Y,Du ZW,Yan YP,Başkaya MK
12-1057 was used in Flow cytometry/Cell sorting to show intravenously administered embryonic mesenchymal stem cells differentiated into neuronal and endothelial cells, and were neuroprotective in transient focal cerebral ischemia.
Wed Apr 01 00:00:00 UTC 2009
Tumor immunotherapy using gene-modified human mesenchymal stem cells loaded into synthetic extracellular matrix scaffolds.
Stem cells (Dayton, Ohio)
Compte M,Cuesta AM,Sánchez-Martín D,Alonso-Camino V,Vicario JL,Sanz L,Alvarez-Vallina L
12-1057 was used in Flow cytometry/Cell sorting to report that human MSCs genetically engineered to secrete a bispecific diabody, seeded in an sECM scaffold and implanted in a location distant from the primary tumor.
Sun Mar 01 00:00:00 UTC 2009
Electrophysiological properties of human adipose tissue-derived stem cells.
American journal of physiology. Cell physiology
Bai X,Ma J,Pan Z,Song YH,Freyberg S,Yan Y,Vykoukal D,Alt E
12-1057 was used in Flow cytometry/Cell sorting to investigate ionic channel currents present in undifferentiated human adipose tissue-derived stem cells (hASCs) and their impact on hASC proliferation.
Thu Nov 01 00:00:00 UTC 2007