Product References
Stimulated Human Umbilical Cord Mesenchymal Stem Cells Enhance the Osteogenesis and Cranial Bone Regeneration through IL-32 Mediated P38 Signaling Pathway.
Stem cells international
Zhang X,Zheng Y,Wang G,Liu Y,Wang Y,Jiang X,Liang Y,Zhao X,Li P,Zhang Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2024
Generation of Highly Functional Hepatocyte-like Organoids from Human Adipose-Derived Mesenchymal Stem Cells Cultured with Endothelial Cells.
Cells
Chen S,Saito Y,Waki Y,Ikemoto T,Teraoku H,Yamada S,Morine Y,Shimada M
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Mar 20 00:00:00 UTC 2024
Comparison of retinal degeneration treatment with four types of different mesenchymal stem cells, human induced pluripotent stem cells and RPE cells in a rat retinal degeneration model.
Journal of translational medicine
Liu Q,Liu J,Guo M,Sung TC,Wang T,Yu T,Tian Z,Fan G,Wu W,Higuchi A
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Dec 14 00:00:00 UTC 2023
Enhanced chondrogenic differentiation of iPS cell-derived mesenchymal stem/stromal cells via neural crest cell induction for hyaline cartilage repair.
Frontiers in cell and developmental biology
Zujur D,Al-Akashi Z,Nakamura A,Zhao C,Takahashi K,Aritomi S,Theoputra W,Kamiya D,Nakayama K,Ikeya M
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sun May 28 00:00:00 UTC 2023
Characteristics of multilineage-differentiating stress-enduring cell clusters in different culture conditions.
Skin research and technology : official journal of International Society for Bioengineering and the Skin (ISBS) [and] International Society for Digital Imaging of Skin (ISDIS) [and] International Society for Skin Imaging (ISSI)
Ning J,Cao YY,Zhang RZ,Li Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2023
Exercise combined with administration of adipose-derived stem cells ameliorates neuropathic pain after spinal cord injury.
Neural regeneration research
Cheng X,Mao GP,Hu WJ,Yu ZR,Xu YY,Chen W,Li X,Zeng XL,Zhang WW,Chen JW,Wan Y,Wang L
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
Selective vulnerability of human-induced pluripotent stem cells to dihydroorotate dehydrogenase inhibition during mesenchymal stem/stromal cell purification.
Frontiers in cell and developmental biology
Al-Akashi Z,Zujur D,Kamiya D,Kato T,Kondo T,Ikeya M
17-1057-42 was used in Flow cytometry/Cell sorting to demonstrates the Brequinar (BRQ)-induced inhibition of dihydroorotate dehydrogenase (DHODH), a key enzyme in de novo pyrimidine biosynthesis, selectively induces apoptosis, cell cycle arrest, and differentiation; furthermore, it promotes transcriptional changes and prevents the growth of 3-dimensional hiPSC aggregates.
Fri Feb 24 00:00:00 UTC 2023
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Mar 03 00:00:00 UTC 2022
Production enhancement of human adipose-derived mesenchymal stem cells by low-intensity ultrasound stimulation.
Scientific reports
Min S,Byeon Y,Kim M,Lee Y,Lee SH,Lee Y,Farooqi HMU,Lee HK,Paeng DG
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Dec 21 00:00:00 UTC 2022
Isolation and characterization mesenchymal stem cells from red panda (Ailurus fulgens styani) endometrium.
Conservation physiology
Wang DH,Wu XM,Chen JS,Cai ZG,An JH,Zhang MY,Li Y,Li FP,Hou R,Liu YL
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sat Jan 01 00:00:00 UTC 2022
Induced Neurodifferentiation of hBM-MSCs through Activation of the ERK/CREB Pathway via Pulsed Electromagnetic Fields and Physical Stimulation Promotes Neurogenesis in Cerebral Ischemic Models.
International journal of molecular sciences
Park HJ,Choi JH,Nam MH,Seo YK
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Jan 21 00:00:00 UTC 2022
Mesenchymal stem cells derived from patients with premature aging syndromes display hallmarks of physiological aging.
Life science alliance
Trani JP,Chevalier R,Caron L,El Yazidi C,Broucqsault N,Toury L,Thomas M,Annab K,Binetruy B,De Sandre-Giovannoli A,Levy N,Magdinier F,Robin JD
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Sep 14 00:00:00 UTC 2022
Aberrant Expression of COX-2 and FOXG1 in Infrapatellar Fat Pad-Derived ASCs from Pre-Diabetic Donors.
Cells
O'Donnell BT,Monjure TA,Al-Ghadban S,Ives CJ,L'Ecuyer MP,Rhee C,Romero-Lopez M,Li Z,Goodman SB,Lin H,Tuan RS,Bunnell BA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
Short-Term Autophagy Preconditioning Upregulates the Expression of COX2 and PGE2 and Alters the Immune Phenotype of Human Adipose-Derived Stem Cells In Vitro.
Cells
Wise RM,Al-Ghadban S,Harrison MAA,Sullivan BN,Monaco ER,Aleman SJ,Donato UM,Bunnell BA
17-1057-42 was used in Flow cytometry/Cell sorting to suggest that a 4-h Rapa preconditioning strategy may bestow the greatest improvement to hASC expression of cytokines known to promote tissue repair and regeneration and may hold promise for augmenting the therapeutic potential of hASCs for inflammation-driven pathological conditions.
Tue Apr 19 00:00:00 UTC 2022
Stabilization of heterochromatin by CLOCK promotes stem cell rejuvenation and cartilage regeneration.
Cell research
Liang C,Liu Z,Song M,Li W,Wu Z,Wang Z,Wang Q,Wang S,Yan K,Sun L,Hishida T,Cai Y,Belmonte JCI,Guillen P,Chan P,Zhou Q,Zhang W,Qu J,Liu GH
17-1057 was used in Flow cytometry/Cell sorting to identify a transcription factor-independent role of CLOCK, a core component of the molecular circadian clock machinery, in counteracting human mesenchymal stem cell (hMSC) decay.
Mon Feb 01 00:00:00 UTC 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Tue Aug 31 00:00:00 UTC 2021
Development, characterization, and hematopoietic differentiation of Griscelli syndrome type 2 induced pluripotent stem cells.
Stem cell research & therapy
Güney-Esken G,Erol ÖD,Pervin B,Gürhan Sevinç G,Önder T,Bilgiç E,Korkusuz P,Günel-Özcan A,Uçkan-Çetinkaya D,Aerts-Kaya F
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu May 13 00:00:00 UTC 2021
Human Amniotic Mesenchymal Stem Cells Inhibit aGVHD by Regulating Balance of Treg and T Effector Cells.
Journal of inflammation research
Gao Y,Li W,Bu X,Xu Y,Cai S,Zhong J,Du M,Sun H,Huang L,He Y,Hu X,Liu Q,Jin H,Wang Q,Ping B
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Tue Apr 26 00:00:00 UTC 2022
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Jul 15 00:00:00 UTC 2021
ZKSCAN3 counteracts cellular senescence by stabilizing heterochromatin.
Nucleic acids research
Hu H,Ji Q,Song M,Ren J,Liu Z,Wang Z,Liu X,Yan K,Hu J,Jing Y,Wang S,Zhang W,Liu GH,Qu J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Jun 19 00:00:00 UTC 2020
Rescue of premature aging defects in Cockayne syndrome stem cells by CRISPR/Cas9-mediated gene correction.
Protein & cell
Wang S,Min Z,Ji Q,Geng L,Su Y,Liu Z,Hu H,Wang L,Zhang W,Suzuiki K,Huang Y,Zhang P,Tang TS,Qu J,Yu Y,Liu GH,Qiao J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2020
SIRT7 antagonizes human stem cell aging as a heterochromatin stabilizer.
Protein & cell
Bi S,Liu Z,Wu Z,Wang Z,Liu X,Wang S,Ren J,Yao Y,Zhang W,Song M,Liu GH,Qu J
17-1057 was used in Flow cytometry/Cell sorting to investigate if SIRT7 expression declines during human mesenchymal stem cell (hMSC) aging and if SIRT7 deficiency accelerates senescence.
Wed Jul 01 00:00:00 UTC 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
17-1057 was used in Flow cytometry/Cell sorting to demonstrate miR-221-3p can improve angiogenesis, migration and proliferation of mesenchymal stem cells while suppressing apoptosis.
Mon Sep 28 00:00:00 UTC 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
17-1057 was used in Flow cytometry/Cell sorting to demonstrate miR-221-3p can improve angiogenesis, migration and proliferation of mesenchymal stem cells while suppressing apoptosis.
Mon Sep 28 00:00:00 UTC 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Tue Oct 06 00:00:00 UTC 2020
Resveratrol rescues TNF‑α‑induced inhibition of osteogenesis in human periodontal ligament stem cells via the ERK1/2 pathway.
Molecular medicine reports
Yuan J,Wang X,Ma D,Gao H,Zheng D,Zhang J
17-1057-42 was used in Flow cytometry/Cell sorting to investigate the effect of tumor necrosis factor‚α, resveratrol, or the combination of both on the osteogenic differentiation of hPDLSCs, as well as the underlying mechanisms involved.
Fri May 01 00:00:00 UTC 2020
Resveratrol rescues TNF‑α‑induced inhibition of osteogenesis in human periodontal ligament stem cells via the ERK1/2 pathway.
Molecular medicine reports
Yuan J,Wang X,Ma D,Gao H,Zheng D,Zhang J
17-1057-42 was used in Flow cytometry/Cell sorting to investigate the effect of tumor necrosis factor‚α, resveratrol, or the combination of both on the osteogenic differentiation of hPDLSCs, as well as the underlying mechanisms involved.
Fri May 01 00:00:00 UTC 2020
Silencing of RBP‑JK promotes the differentiation of bone marrow mesenchymal stem cells into vascular endothelial cells.
Molecular medicine reports
Luo Z,Dai Y,Chen M,Zhu C,Wu K,Li G,Shang X
17-1057 was used in Flow cytometry/Cell sorting to demonstrate that silencing of RBP-JK promotes differentiation of bone marrow mesenchymal stem cells into vascular endothelial cells, a promising therapeutic basis for the treatment of bone defects.
Wed Jan 01 00:00:00 UTC 2020
Vasculogenesis from Human Dental Pulp Stem Cells Grown in Matrigel with Fully Defined Serum-Free Culture Media.
Biomedicines
Luzuriaga J,Irurzun J,Irastorza I,Unda F,Ibarretxe G,Pineda JR
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Mon Nov 09 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
Chondrogenic Differentiation from Induced Pluripotent Stem Cells Using Non-Viral Minicircle Vectors.
Cells
Rim YA,Nam Y,Park N,Jung H,Lee K,Lee J,Ju JH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sun Mar 01 00:00:00 UTC 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Nov 19 00:00:00 UTC 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Nov 27 00:00:00 UTC 2020
Increase in Leptin and PPAR-γ Gene Expression in Lipedema Adipocytes Differentiated in vitro from Adipose-Derived Stem Cells.
Cells
Al-Ghadban S,Diaz ZT,Singer HJ,Mert KB,Bunnell BA
17-1057-42 was used in Flow cytometry/Cell sorting to characterize the adipose-derived stem cells of healthy and lipedema patients by the expression of stemness markers and the adipogenic and osteogenic differentiation potential.
Wed Feb 12 00:00:00 UTC 2020
Telomere-dependent and telomere-independent roles of RAP1 in regulating human stem cell homeostasis.
Protein & cell
Zhang X,Liu Z,Liu X,Wang S,Zhang Y,He X,Sun S,Ma S,Shyh-Chang N,Liu F,Wang Q,Wang X,Liu L,Zhang W,Song M,Liu GH,Qu J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sun Sep 01 00:00:00 UTC 2019
Chemical screen identifies a geroprotective role of quercetin in premature aging.
Protein & cell
Geng L,Liu Z,Zhang W,Li W,Wu Z,Wang W,Ren R,Su Y,Wang P,Sun L,Ju Z,Chan P,Song M,Qu J,Liu GH
17-1057 was used in Flow cytometry/Cell sorting to identify quercetin as a geroprotective agent against accelerated and natural aging in human mesenchymal stem cells, providing a potential therapeutic intervention for treating age-associated disorders.
Sat Jun 01 00:00:00 UTC 2019
Up-regulation of FOXD1 by YAP alleviates senescence and osteoarthritis.
PLoS biology
Fu L,Hu Y,Song M,Liu Z,Zhang W,Yu FX,Wu J,Wang S,Izpisua Belmonte JC,Chan P,Qu J,Tang F,Liu GH
17-1057 was used in Flow cytometry/Cell sorting to reveal YAP-FOXD1, a novel aging-associated regulatory axis, as a potential target for gene therapy to alleviate osteoarthritis.
Mon Apr 01 00:00:00 UTC 2019
Up-regulation of FOXD1 by YAP alleviates senescence and osteoarthritis.
PLoS biology
Fu L,Hu Y,Song M,Liu Z,Zhang W,Yu FX,Wu J,Wang S,Izpisua Belmonte JC,Chan P,Qu J,Tang F,Liu GH
17-1057 was used in Flow cytometry/Cell sorting to reveal YAP-FOXD1, a novel aging-associated regulatory axis, as a potential target for gene therapy to alleviate osteoarthritis.
Mon Apr 01 00:00:00 UTC 2019
Overexpression of ERBB4 rejuvenates aged mesenchymal stem cells and enhances angiogenesis via PI3K/AKT and MAPK/ERK pathways.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Liang X,Ding Y,Lin F,Zhang Y,Zhou X,Meng Q,Lu X,Jiang G,Zhu H,Chen Y,Lian Q,Fan H,Liu Z
17-1057-42 was used in Flow Cytometry to investigate whether Erb-B2 receptor tyrosine kinase 4 ERBB4 rejuvenates aged mesenchymal stem cells MSC and how ERBB4 enhances therapeutic efficacy of aged MSC in treating myocardial infarction (MI).
Fri Mar 01 00:00:00 UTC 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu May 13 00:00:00 UTC 2021
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2019
Ultrastructural morphology is distinct among primary progenitor cell isolates from normal, inflamed, and cryopreserved equine hoof tissue and CD105+K14+ progenitor cells.
In vitro cellular & developmental biology. Animal
Yang Q,Lopez MJ
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sun Sep 01 00:00:00 UTC 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu May 02 00:00:00 UTC 2019
Stabilizing heterochromatin by DGCR8 alleviates senescence and osteoarthritis.
Nature communications
Deng L,Ren R,Liu Z,Song M,Li J,Wu Z,Ren X,Fu L,Li W,Zhang W,Guillen P,Izpisua Belmonte JC,Chan P,Qu J,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Jul 26 00:00:00 UTC 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Dec 20 00:00:00 UTC 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2019
Differential stem cell aging kinetics in Hutchinson-Gilford progeria syndrome and Werner syndrome.
Protein & cell
Wu Z,Zhang W,Song M,Wang W,Wei G,Li W,Lei J,Huang Y,Sang Y,Chan P,Chen C,Qu J,Suzuki K,Belmonte JCI,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2018
Differential stem cell aging kinetics in Hutchinson-Gilford progeria syndrome and Werner syndrome.
Protein & cell
Wu Z,Zhang W,Song M,Wang W,Wei G,Li W,Lei J,Huang Y,Sang Y,Chan P,Chen C,Qu J,Suzuki K,Belmonte JCI,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2018
Antagonizing CD105 enhances radiation sensitivity in prostate cancer.
Oncogene
Madhav A,Andres A,Duong F,Mishra R,Haldar S,Liu Z,Angara B,Gottlieb R,Zumsteg ZS,Bhowmick NA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Aug 01 00:00:00 UTC 2018
Antagonizing CD105 enhances radiation sensitivity in prostate cancer.
Oncogene
Madhav A,Andres A,Duong F,Mishra R,Haldar S,Liu Z,Angara B,Gottlieb R,Zumsteg ZS,Bhowmick NA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Aug 01 00:00:00 UTC 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
ATF6 safeguards organelle homeostasis and cellular aging in human mesenchymal stem cells.
Cell discovery
Wang S,Hu B,Ding Z,Dang Y,Wu J,Li D,Liu X,Xiao B,Zhang W,Ren R,Lei J,Hu H,Chen C,Chan P,Li D,Qu J,Tang F,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
The Improvement of Respiratory Performance After Phototherapy-Induced EPC Mobilization in Preterm Infants With RDS.
Journal of cellular biochemistry
Siavashi V,Asadian S,Taheri-Asl M,Babaei H,Keshavarz S,Bazaei M,Nassiri SM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
Circulation Enrichment of Functional Endothelial Progenitor Cells by Infantile Phototherapy.
Journal of cellular biochemistry
Siavashi V,Asadian S,Sharifi A,Esmaeilivand M,Norouzinia R,Azadbakht M,Nassiri SM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2017
Comparing the in vivo and in vitro effects of hypoxia (3% O2) on directly derived cells from murine cardiac explants versus murine cardiosphere derived cells.
Journal of stem cells & regenerative medicine
Amirrasouli MM,Shamsara M
17-1057 was used in Flow cytometry/Cell sorting to compare explant-derived cells with cardiosphere-derived cells, and assess the effect of hypoxic reconditioning on their pro-angiogenic potential.
Tue Sep 29 00:00:00 UTC 2020
Region and cell-type resolved quantitative proteomic map of the human heart.
Nature communications
Doll S,Dreßen M,Geyer PE,Itzhak DN,Braun C,Doppler SA,Meier F,Deutsch MA,Lahm H,Lange R,Krane M,Mann M
17-1057 was used in Flow cytometry/Cell sorting to determine the healthy human heart proteome.
Mon Nov 13 00:00:00 UTC 2017
SIRT6 safeguards human mesenchymal stem cells from oxidative stress by coactivating NRF2.
Cell research
Pan H,Guan D,Liu X,Li J,Wang L,Wu J,Zhou J,Zhang W,Ren R,Zhang W,Li Y,Yang J,Hao Y,Yuan T,Yuan G,Wang H,Ju Z,Mao Z,Li J,Qu J,Tang F,Liu GH
17-1057 was used in Flow cytometry/Cell sorting to investigate whether human SIRT6 has a direct role in maintaining the homeostasis of mesodermal tissues, showing that it safeguards against oxidative stress by coactivating NRF2.
Mon Feb 01 00:00:00 UTC 2016
Lymphatic Contribution to the Cellular Niche in Heterotopic Ossification.
Annals of surgery
Loder S,Agarwal S,Sorkin M,Breuler C,Li J,Peterson J,Gardenier J,Hsieh HH,Wang SC,Mehrara BJ,Levi B
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2016
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Tue Jan 19 00:00:00 UTC 2016
Angiogenic activity mediates bone repair from human pluripotent stem cell-derived osteogenic cells.
Scientific reports
Zou L,Chen Q,Quanbeck Z,Bechtold JE,Kaufman DS
17-1057 was used in Flow cytometry/Cell sorting to explore the role of the pro-angiogenic potential of transplanted osteogenic cells for effective cell-mediated bone repair.
Wed Mar 16 00:00:00 UTC 2016
Influence of aging on the quantity and quality of human cardiac stem cells.
Scientific reports
Nakamura T,Hosoyama T,Kawamura D,Takeuchi Y,Tanaka Y,Samura M,Ueno K,Nishimoto A,Kurazumi H,Suzuki R,Ito H,Sakata K,Mikamo A,Li TS,Hamano K
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Tue Aug 09 00:00:00 UTC 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
Modeling familial cancer with induced pluripotent stem cells.
Cell
Lee DF,Su J,Kim HS,Chang B,Papatsenko D,Zhao R,Yuan Y,Gingold J,Xia W,Darr H,Mirzayans R,Hung MC,Schaniel C,Lemischka IR
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Apr 09 00:00:00 UTC 2015
Mesenchymal stromal cells form vascular tubes when placed in fibrin sealant and accelerate wound healing in vivo.
Biomaterials
Mendez JJ,Ghaedi M,Sivarapatna A,Dimitrievska S,Shao Z,Osuji CO,Steinbacher DM,Leffell DJ,Niklason LE
17-1057 was used in Flow cytometry/Cell sorting to show that ADSCs are able to form tubular structures within fibrin gels, and may also contribute to faster wound healing.
Sun Feb 01 00:00:00 UTC 2015
Impaired function of bone marrow stromal cells in systemic mastocytosis.
Stem cell research
Nemeth K,Wilson TM,Ren JJ,Sabatino M,Stroncek DM,Krepuska M,Bai Y,Robey PG,Metcalfe DD,Mezey E
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2015
Human adipose-derived stem cells partially rescue the stroke syndromes by promoting spatial learning and memory in mouse middle cerebral artery occlusion model.
Stem cell research & therapy
Zhou F,Gao S,Wang L,Sun C,Chen L,Yuan P,Zhao H,Yi Y,Qin Y,Dong Z,Cao L,Ren H,Zhu L,Li Q,Lu B,Liang A,Xu GT,Zhu H,Gao Z,Ma J,Xu J,Chen X
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sat May 09 00:00:00 UTC 2015
Human Gingival Integration-Free iPSCs; a Source for MSC-Like Cells.
International journal of molecular sciences
Umezaki Y,Hashimoto Y,Nishishita N,Kawamata S,Baba S
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Mon Jun 15 00:00:00 UTC 2015
PTEN deficiency reprogrammes human neural stem cells towards a glioblastoma stem cell-like phenotype.
Nature communications
Duan S,Yuan G,Liu X,Ren R,Li J,Zhang W,Wu J,Xu X,Fu L,Li Y,Yang J,Zhang W,Bai R,Yi F,Suzuki K,Gao H,Esteban CR,Zhang C,Izpisua Belmonte JC,Chen Z,Wang X,Jiang T,Qu J,Tang F,Liu GH
17-1057 was used in Flow cytometry to uncover a potential mechanism by which PTEN safeguards neural stem cells (NSCs), and establish a cellular platform to identify factors involved in NSC transformation.
Thu Dec 03 00:00:00 UTC 2015
Human adipose stem cell and ASC-derived cardiac progenitor cellular therapy improves outcomes in a murine model of myocardial infarction.
Stem cells and cloning : advances and applications
Davy PM,Lye KD,Mathews J,Owens JB,Chow AY,Wong L,Moisyadi S,Allsopp RC
17-1057 was used in Flow cytometry/Cell sorting to investigate the potential use of adipose-derived stem cells.
Wed Nov 25 00:00:00 UTC 2015
Embelin inhibits endothelial mitochondrial respiration and impairs neoangiogenesis during tumor growth and wound healing.
EMBO molecular medicine
Coutelle O,Hornig-Do HT,Witt A,Andree M,Schiffmann LM,Piekarek M,Brinkmann K,Seeger JM,Liwschitz M,Miwa S,Hallek M,Krönke M,Trifunovic A,Eming SA,Wiesner RJ,Hacker UT,Kashkar H
17-1057 was used in Flow cytometry/Cell sorting to demonstrate a novel therapy to control pathological neoangiogenesis while sparing normal vasculature.
Thu May 01 00:00:00 UTC 2014
Interaction of mesenchymal stem cells with fibroblast-like synoviocytes via cadherin-11 promotes angiogenesis by enhanced secretion of placental growth factor.
Journal of immunology (Baltimore, Md. : 1950)
Park SJ,Kim KJ,Kim WU,Cho CS
17-1057 was used in Flow cytometry/Cell sorting to study how cadherin-11 allows mesenchymal stem cells and fibroblast-like synoviocytes to interact, promoting angiogenesis.
Tue Apr 01 00:00:00 UTC 2014
Serum- and growth-factor-free three-dimensional culture system supports cartilage tissue formation by promoting collagen synthesis via Sox9-Col2a1 interaction.
Tissue engineering. Part A
Ahmed N,Iu J,Brown CE,Taylor DW,Kandel RA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Fri Aug 01 00:00:00 UTC 2014
Epithelial cell differentiation of human mesenchymal stromal cells in decellularized lung scaffolds.
Tissue engineering. Part A
Mendez JJ,Ghaedi M,Steinbacher D,Niklason LE
Sun Jun 01 00:00:00 UTC 2014
Lineage depletion of stromal vascular fractions isolated from human adipose tissue: a novel approach towards cell enrichment technology.
Cytotechnology
Indumathi S,Mishra R,Harikrishnan R,Rajkumar JS,Kantawala N,Dhanasekaran M
17-1057 was used in Flow cytometry/Cell sorting to understand the comparative efficacy of lineage depleted adipose derived stromal vascular fraction (SVF), to help rationalise tissue repair and regeneration using stem cells.
Sat Mar 01 00:00:00 UTC 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Thu Dec 11 00:00:00 UTC 2014
Modelling Fanconi anemia pathogenesis and therapeutics using integration-free patient-derived iPSCs.
Nature communications
Liu GH,Suzuki K,Li M,Qu J,Montserrat N,Tarantino C,Gu Y,Yi F,Xu X,Zhang W,Ruiz S,Plongthongkum N,Zhang K,Masuda S,Nivet E,Tsunekawa Y,Soligalla RD,Goebl A,Aizawa E,Kim NY,Kim J,Dubova I,Li Y,Ren R,Benner C,Del Sol A,Bueren J,Trujillo JP,Surralles J,Cappelli E,Dufour C,Esteban CR,Belmonte JCI
Mon Jul 07 00:00:00 UTC 2014
FOXF1 mediates mesenchymal stem cell fusion-induced reprogramming of lung cancer cells.
Oncotarget
Wei HJ,Nickoloff JA,Chen WH,Liu HY,Lo WC,Chang YT,Yang PC,Wu CW,Williams DF,Gelovani JG,Deng WP
17-1057 was used in Flow cytometry/Cell sorting to study the effect of fusion of mesenchymal stem cells with lung cancer cells, showing that fusion is mediated by FOXF1 and leads to a more benign state due to complementation of tumorigenic defects such as the restoration of p21 function.
Wed Oct 15 00:00:00 UTC 2014
Vasculogenic conditioning of peripheral blood mononuclear cells promotes endothelial progenitor cell expansion and phenotype transition of anti-inflammatory macrophage and T lymphocyte to cells with regenerative potential.
Journal of the American Heart Association
Masuda H,Tanaka R,Fujimura S,Ishikawa M,Akimaru H,Shizuno T,Sato A,Okada Y,Iida Y,Itoh J,Itoh Y,Kamiguchi H,Kawamoto A,Asahara T
17-1057-42 was used in Flow Cytometry to investigate the therapeutic potential of peripheral blood (PB) MNCs, operated by recently developed quality and quantity (QQ) culture of endothelial progenitor cells (EPCs).
Wed Jun 25 00:00:00 UTC 2014
Genetic stability of bone marrow-derived human mesenchymal stromal cells in the Quantum System.
Cytotherapy
Jones M,Varella-Garcia M,Skokan M,Bryce S,Schowinsky J,Peters R,Vang B,Brecheisen M,Startz T,Frank N,Nankervis B
17-1057 was used in Flow cytometry/Cell sorting to determine the genetic stability of bone marrow-derived human mesenchymal stromal cells selected and produced with the Quantum Cell Expansion System.
Fri Nov 01 00:00:00 UTC 2013
Early intervention with gene-modified mesenchymal stem cells overexpressing interleukin-4 enhances anti-inflammatory responses and functional recovery in experimental autoimmune demyelination.
Cell adhesion & migration
Payne NL,Dantanarayana A,Sun G,Moussa L,Caine S,McDonald C,Herszfeld D,Bernard CC,Siatskas C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Wed Dec 19 00:00:00 UTC 2012
Surface antigenic profiling of stem cells from human omentum fat in comparison with subcutaneous fat and bone marrow.
Cytotechnology
Dhanasekaran M,Indumathi S,Kanmani A,Poojitha R,Revathy KM,Rajkumar JS,Sudarsanam D
17-1057 was used in Flow cytometry/Cell sorting to investigate surface antigenic profiling of stem cells from human omentum fat, compared with subcutaneous fat and bone marrow..
Wed Feb 01 00:00:00 UTC 2012
Identification of a candidate proteomic signature to discriminate multipotent and non-multipotent stromal cells.
PloS one
Rosu-Myles M,She YM,Fair J,Muradia G,Mehic J,Menendez P,Prasad SS,Cyr TD
17-1057 was used in Flow cytometry/Cell sorting to provide evidence of heterogeneity among bone marrow stromal cells and identify potential candidate proteins for quantifying MSCs in vitro.
Thu Dec 13 00:00:00 UTC 2012
Unravelling the retention of proliferation and differentiation potency in extensive culture of human subcutaneous fat-derived mesenchymal stem cells in different media.
Cell proliferation
Dhanasekaran M,Indumathi S,Rashmi M,Rajkumar JS,Sudarsanam D
17-1057-42 was used in Flow Cytometry to evaluate SF-MSCs with reference to phenotypic characterization, proliferative ability, karyotype stability and differentiation potency with early (P3) and late passage (P20) conditions, using four different media, DMEM-LG, ALPHA-MEM, DMEM-F12 and DMEM-KO.
Sat Dec 01 00:00:00 UTC 2012
A differentiation-based microRNA signature identifies leiomyosarcoma as a mesenchymal stem cell-related malignancy.
The American journal of pathology
Danielson LS,Menendez S,Attolini CS,Guijarro MV,Bisogna M,Wei J,Socci ND,Levine DA,Michor F,Hernando E
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 17-1057-42) in Flow Cytometry
Sun Aug 01 00:00:00 UTC 2010