Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CSF1R (CD115) Antibodies

            Invitrogen

            CD115 (c-fms) Monoclonal Antibody (12-3A3-1B10), PE, eBioscience™

            1 Published Figure
            7 References
            View all (82) CSF1R (CD115) antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 12-1159-42
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            366.35
            Special offer
            Online exclusive
            Ends: 01-Jul-2025
            431.00
            Save 64.65 (15%)
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CD115 (c-fms) Monoclonal Antibody (12-3A3-1B10), PE, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (1)
            Application
            CD115 (c-fms) Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            CD115 (c-fms) Antibody (12-1159-42) in Flow

            Staining of normal human peripheral blood cells with Rat IgG2a Isotype Contol PE (Product # 12-4321-80) (open histogram) or Anti-Human CD115 (c-fms) PE (filled histogram). Cells in the monocyte gate were used for analysis.
            View Product

            CD115 (c-fms) Antibody in Flow Cytometry (Flow)
            CD115 (c-fms) Antibody in Flow Cytometry (Flow)

            Product Details

            12-1159-42

            Applications
            Tested Dilution
            Publications

            Flow Cytometry (Flow)

            5 µL (0.03 µg)/test
            View 7 publications 7 publications
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human

            Host/Isotype

            Rat / IgG2a, kappa

            Recommended Isotype Control

            Rat IgG2a kappa Isotype Control (eBR2a), PE, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            12-3A3-1B10

            Conjugate

            PE
            • Unconjugated
            • Biotin 
            • PE-Cyanine7
            • Super Bright 436
            • Request custom conjugation

            Excitation/Emission Max

            565/576 nm View spectra spectra

            Form

            Liquid

            Concentration

            5 µL/Test

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2, with BSA

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_10717675

            Product Specific Information

            Description: CD115 or Colony-Stimulating Factor-1 Receptor (CSF-1R) is expressed on cells committed to the monocyte lineage and osteoclasts. CSF-1R is a disulfide-linked homodimer, which upon binding of its ligand, CSF-1, undergoes tyrosine autophosphorylation and subsequently, phosphorylates other membrane-proximal downstream targets which results in cytoskeletal remodeling, gene transcription and protein translation. CSF-1R activation promotes the survival, proliferation and differentiation of mononuclear phagocytes and the spreading and motility of macrophages. In osteoclasts, CSF-1R synergizes with RANKL to regulate the differentiation of mononuclear phagocytes to osteoclasts.

            Applications Reported: This 12-3A3-1B10 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This 12-3A3-1B10 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood. This can be used at 5 µL (0.03 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

            Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 8:21
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -8:21
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Bioinformatics

            Protein Aliases: CD115; CD115 antigen; CSF-1 receptor; CSF-1-R; CSFIR; EC 2.7.10.1; FMS proto-oncogene; kinase CSFR; macrophage colony stimulating factor I receptor; Macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; OTTHUMP00000224093; Proto-oncogene c-Fms

            View more View less

            Gene Aliases: C-FMS; CD115; CSF-1R; CSF1R; CSFR; FIM2; FMS; HDLS; M-CSF-R

            View more View less

            UniProt ID: (Human) P07333

            View more View less

            Entrez Gene ID: (Human) 1436

            View more View less

            Function(s)

            macrophage colony-stimulating factor receptor activity ATP binding protein phosphatase binding cytokine binding protein homodimerization activity transmembrane signal receptor

            Process(es)

            positive regulation of protein phosphorylation inflammatory response signal transduction transmembrane receptor protein tyrosine kinase signaling pathway multicellular organism development axon guidance skeletal muscle tissue development cell proliferation positive regulation of cell proliferation negative regulation of cell proliferation regulation of cell shape peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway olfactory bulb development forebrain neuron differentiation hemopoiesis monocyte differentiation macrophage differentiation osteoclast differentiation positive regulation of cell migration ruffle organization cellular response to macrophage colony-stimulating factor stimulus macrophage colony-stimulating factor signaling pathway positive regulation of tyrosine phosphorylation of Stat3 protein negative regulation of apoptotic process innate immune response regulation of bone resorption cell-cell junction maintenance positive regulation of osteoclast differentiation phosphatidylinositol metabolic process protein autophosphorylation phosphatidylinositol-mediated signaling mammary gland duct morphogenesis positive regulation of protein tyrosine kinase activity positive regulation of ERK1 and ERK2 cascade cellular response to cytokine stimulus positive regulation of protein serine/threonine kinase activity positive regulation of chemokine secretion positive regulation of cell motility regulation of actin cytoskeleton reorganization
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-c4vbx:80/100.66.73.217:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0