Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CSF2RA (CD116) (Met1-Gly320) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD116; CD116 antigen; CDw116; colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage); CSF 2R; GM CSF R alpha; GM-CSF receptor alpha subunit; GM-CSF-R-alpha; GMCSFR-alpha; GMR-alpha; granulocyte-macrophage colony-stimulating factor receptor alpha chain; Granulocyte-macrophage colony-stimulating factor receptor subunit alpha; MGC3848; MGC4838
Gene Aliases: CD116; CDw116; CSF2R; CSF2RA; CSF2RAX; CSF2RAY; CSF2RX; CSF2RY; GM-CSF-R-alpha; GMCSFR; GMR; SMDP4
UniProt ID: (Human) P15509
Entrez Gene ID: (Human) 1438
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support