Product References
Calcineurin Aα Contributes to IgE-Dependent Mast-Cell Mediator Secretion in Allergic Inflammation.
Journal of innate immunity
Leong E,Pang Z,Stadnyk AW,Lin TJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Jul 07 00:00:00 EDT 2022
Calcineurin Aα Contributes to IgE-Dependent Mast-Cell Mediator Secretion in Allergic Inflammation.
Journal of innate immunity
Leong E,Pang Z,Stadnyk AW,Lin TJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Jul 07 00:00:00 EDT 2022
Gastruloids as in vitro models of embryonic blood development with spatial and temporal resolution.
Scientific reports
Rossi G,Giger S,Hübscher T,Lutolf MP
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Aug 04 00:00:00 EDT 2022
Specific overexpression of SIRT1 in mesenchymal stem cells rescues hematopoiesis niche in BMI1 knockout mice through promoting CXCL12 expression.
International journal of biological sciences
Li J,Li X,Sun W,Zhang J,Yan Q,Wu J,Jin J,Lu R,Miao D
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Apr 15 00:00:00 EDT 2022
Centrosome function is critical during terminal erythroid differentiation.
The EMBO journal
Tátrai P,Gergely F
11-1171-82 was used in Flow cytometry/Cell sorting to show that disruption of centrosome and spindle function initiates macrocytic anaemias due to nutritional deficiency or exposure to chemotherapy.
Mon Jul 18 00:00:00 EDT 2022
Preventive Effects of a Human Hematopoietic Mesenchymal Stem Cell (hHMSC) Therapy in Ovalbumin-Induced Food Allergy.
Biomedicines
Lee DG,Lee YJ,Park SH,Park HR,Kang H,Kim JE
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Feb 21 00:00:00 EST 2022
Bone marrow-derived myeloid progenitors in the leptomeninges of adult mice.
Stem cells (Dayton, Ohio)
Koeniger T,Bell L,Mifka A,Enders M,Hautmann V,Mekala SR,Kirchner P,Ekici AB,Schulz C,Wörsdörfer P,Mencl S,Kleinschnitz C,Ergün S,Kuerten S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunocytochemistry
Mon Feb 01 00:00:00 EST 2021
Inaccessible LCG Promoters Act as Safeguards to Restrict T Cell Development to Appropriate Notch Signaling Environments.
Stem cell reports
Furuyama S,Wu QV,Varnum-Finney B,Sandstrom R,Meuleman W,Stamatoyannopoulos JA,Bernstein ID
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Apr 13 00:00:00 EDT 2021
Mitochondrial transcription factor A in RORγt+ lymphocytes regulate small intestine homeostasis and metabolism.
Nature communications
Fu Z,Dean JW,Xiong L,Dougherty MW,Oliff KN,Chen ZE,Jobin C,Garrett TJ,Zhou L
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Jul 22 00:00:00 EDT 2021
SNAP23 is essential for platelet and mast cell development and required in connective tissue mast cells for anaphylaxis.
The Journal of biological chemistry
Cardenas RA,Gonzalez R,Sanchez E,Ramos MA,Cardenas EI,Rodarte AI,Alcazar-Felix RJ,Isaza A,Burns AR,Heidelberger R,Adachi R
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
mMCP7, a Mouse Ortholog of δ Tryptase, Mediates Pelvic Tactile Allodynia in a Model of Chronic Pelvic Pain.
Frontiers in pain research (Lausanne, Switzerland)
Pattabiraman G,Liu Z,Paul M,Schaeffer AJ,Thumbikat P
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Oct 20 00:00:00 EDT 2023
Analysis of Radiation-Induced Changes in Cell Cycle and DNA Damage of Murine Hematopoietic Stem Cells by Multi-Color Flow Cytometry.
Current protocols
Majumdar D,Pietras EM,Pawar SA
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Targeting FTO Suppresses Cancer Stem Cell Maintenance and Immune Evasion.
Cancer cell
Su R,Dong L,Li Y,Gao M,Han L,Wunderlich M,Deng X,Li H,Huang Y,Gao L,Li C,Zhao Z,Robinson S,Tan B,Qing Y,Qin X,Prince E,Xie J,Qin H,Li W,Shen C,Sun J,Kulkarni P,Weng H,Huang H,Chen Z,Zhang B,Wu X,Olsen MJ,Müschen M,Marcucci G,Salgia R,Li L,Fathi AT,Li Z,Mulloy JC,Wei M,Horne D,Chen J
11-1171-82 was used in Flow Cytometry to demonstrate if fat mass and obesity-associated protein plays critical roles in cancer stem cell self-renewal and immune evasion.
Mon Jul 13 00:00:00 EDT 2020
Bone Marrow Mesenchymal Stem Cells Support Acute Myeloid Leukemia Bioenergetics and Enhance Antioxidant Defense and Escape from Chemotherapy.
Cell metabolism
Forte D,García-Fernández M,Sánchez-Aguilera A,Stavropoulou V,Fielding C,Martín-Pérez D,López JA,Costa ASH,Tronci L,Nikitopoulou E,Barber M,Gallipoli P,Marando L,Fernández de Castillejo CL,Tzankov A,Dietmann S,Cavo M,Catani L,Curti A,Vázquez J,Frezza C,Huntly BJ,Schwaller J,Méndez-Ferrer S
11-1171 was used in Flow cytometry/Cell sorting to study the contribution of nestin+ BM mesenchymal stem cells to MLL-AF9-driven acute myeloid leukemia development and chemoresistance in vivo.
Tue Nov 03 00:00:00 EST 2020
Infiltrating Mast Cell-Mediated Stimulation of Estrogen Receptor Activity in Breast Cancer Cells Promotes the Luminal Phenotype.
Cancer research
Majorini MT,Cancila V,Rigoni A,Botti L,Dugo M,Triulzi T,De Cecco L,Fontanella E,Jachetti E,Tagliabue E,Chiodoni C,Tripodo C,Colombo MP,Lecis D
11-1171-82 was used in Flow Cytometry to support the notion that MCs influence the phenotype of breast cancer cells by stimulating a luminal phenotype and ultimately modifying the outcome of the disease.
Mon Jun 01 00:00:00 EDT 2020
Reduced Nhe1 (Na+-H+ Exchanger-1) Function Protects ApoE-Deficient Mice From Ang II (Angiotensin II)-Induced Abdominal Aortic Aneurysms.
Hypertension (Dallas, Tex. : 1979)
Liu CL,Liu X,Wang Y,Deng Z,Liu T,Sukhova GK,Wojtkiewicz GR,Tang R,Zhang JY,Achilefu S,Nahrendorf M,Libby P,Wang X,Shi GP
11-1171-82 was used in Immunohistochemistry to establish a direct role of Nhe1 in AAA pathogenesis.
Wed Jul 01 00:00:00 EDT 2020
Explant culture: A relevant tool for the study of telocytes.
Cell biology international
Sanches BDA,Maldarine JDS,Tamarindo GH,Da Silva ADT,Lima MLD,Rahal P,Góes RM,Taboga SR,Carvalho HF
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunocytochemistry
Tue Dec 01 00:00:00 EST 2020
Explant culture: A relevant tool for the study of telocytes.
Cell biology international
Sanches BDA,Maldarine JDS,Tamarindo GH,Da Silva ADT,Lima MLD,Rahal P,Góes RM,Taboga SR,Carvalho HF
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunocytochemistry
Tue Dec 01 00:00:00 EST 2020
XIAOPI formula inhibits the pre-metastatic niche formation in breast cancer via suppressing TAMs/CXCL1 signaling.
Cell communication and signaling : CCS
Zheng Y,Wang N,Wang S,Yang B,Situ H,Zhong L,Lin Y,Wang Z
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Mar 26 00:00:00 EDT 2020
USP15 Deubiquitinase Safeguards Hematopoiesis and Genome Integrity in Hematopoietic Stem Cells and Leukemia Cells.
Cell reports
van den Berk P,Lancini C,Company C,Serresi M,Sanchez-Bailon MP,Hulsman D,Pritchard C,Song JY,Schmitt MJ,Tanger E,Popp O,Mertins P,Huijbers IJ,Jacobs H,van Lohuizen M,Gargiulo G,Citterio E
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Dec 29 00:00:00 EST 2020
Authentication of Primary Murine Cell Lines by a Microfluidics-Based Lab-On-Chip System.
Biomedicines
Hong Y,Singh N,Bamopoulos S,Gjerga E,Schmalbrock LK,Balabanian K,Schick M,Keller U,Wirth M
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Wed Dec 09 00:00:00 EST 2020
Type I Interferon α/β Receptor-Mediated Signaling Negatively Regulates Antiviral Cytokine Responses in Murine Bone-Marrow-Derived Mast Cells and Protects the Cells from Virus-Induced Cell Death.
International journal of molecular sciences
Darzianiazizi M,Mehrani Y,Chan L,Mould RC,Kulkarni RR,Sharif S,Bridle BW,Karimi K
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Nov 27 00:00:00 EST 2020
Type I Interferon α/β Receptor-Mediated Signaling Negatively Regulates Antiviral Cytokine Responses in Murine Bone-Marrow-Derived Mast Cells and Protects the Cells from Virus-Induced Cell Death.
International journal of molecular sciences
Darzianiazizi M,Mehrani Y,Chan L,Mould RC,Kulkarni RR,Sharif S,Bridle BW,Karimi K
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Nov 27 00:00:00 EST 2020
Remodeling of Bone Marrow Hematopoietic Stem Cell Niches Promotes Myeloid Cell Expansion during Premature or Physiological Aging.
Cell stem cell
Ho YH,Del Toro R,Rivera-Torres J,Rak J,Korn C,García-García A,Macías D,González-Gómez C,Del Monte A,Wittner M,Waller AK,Foster HR,López-Otín C,Johnson RS,Nerlov C,Ghevaert C,Vainchenker W,Louache F,Andrés V,Méndez-Ferrer S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Sep 05 00:00:00 EDT 2019
Dynamic changes in epithelial cell morphology control thymic organ size during atrophy and regeneration.
Nature communications
Venables T,Griffith AV,DeAraujo A,Petrie HT
11-1171 was used in Flow cytometry/Cell sorting to show that accelerated thymic atrophy reflects contraction of complex cell projections unique to cortical epithelial cells, while regeneration requires their regrowth.
Fri Sep 27 00:00:00 EDT 2019
Defective germline reprogramming rewires the spermatogonial transcriptome.
Nature structural & molecular biology
Vasiliauskaitė L,Berrens RV,Ivanova I,Carrieri C,Reik W,Enright AJ,O'Carroll D
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2018
Detection of Subclinical Arthritis in Mice by a Thrombin Receptor-Derived Imaging Agent.
Arthritis & rheumatology (Hoboken, N.J.)
Friedman B,Whitney MA,Savariar EN,Caneda C,Steinbach P,Xiong Q,Hingorani DV,Crisp J,Adams SR,Kenner M,Lippert CN,Nguyen QT,Guma M,Tsien RY,Corr M
11-1171 was used in Immunohistochemistry-immunofluorescence to examine coagulation protease activities in arthritic mice with a dual-fluorescence ratiometric activatable cell-penetrating peptide that has a linker, norleucine-TPRSFL, with a cleavage site for thrombin.
Mon Jan 01 00:00:00 EST 2018
JAK2 is dispensable for maintenance of JAK2 mutant B-cell acute lymphoblastic leukemias.
Genes & development
Kim SK,Knight DA,Jones LR,Vervoort S,Ng AP,Seymour JF,Bradner JE,Waibel M,Kats L,Johnstone RW
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
miR-143/145 differentially regulate hematopoietic stem and progenitor activity through suppression of canonical TGFβ signaling.
Nature communications
Lam J,van den Bosch M,Wegrzyn J,Parker J,Ibrahim R,Slowski K,Chang L,Martinez-Høyer S,Condorelli G,Boldin M,Deng Y,Umlandt P,Fuller M,Karsan A
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Wed Jun 20 00:00:00 EDT 2018
A stochastic epigenetic switch controls the dynamics of T-cell lineage commitment.
eLife
Ng KK,Yui MA,Mehta A,Siu S,Irwin B,Pease S,Hirose S,Elowitz MB,Rothenberg EV,Kueh HY
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Nov 20 00:00:00 EST 2018
The H3K4 methyltransferase Setd1b is essential for hematopoietic stem and progenitor cell homeostasis in mice.
eLife
Schmidt K,Zhang Q,Tasdogan A,Petzold A,Dahl A,Arneth BM,Slany R,Fehling HJ,Kranz A,Stewart AF,Anastassiadis K
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Jun 19 00:00:00 EDT 2018
Generation of Vascular Endothelial Cells and Hematopoietic Cells by Blastocyst Complementation.
Stem cell reports
Hamanaka S,Umino A,Sato H,Hayama T,Yanagida A,Mizuno N,Kobayashi T,Kasai M,Suchy FP,Yamazaki S,Masaki H,Yamaguchi T,Nakauchi H
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Oct 09 00:00:00 EDT 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Mar 20 00:00:00 EDT 2018
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus.
eLife
Moretti FA,Klapproth S,Ruppert R,Margraf A,Weber J,Pick R,Scheiermann C,Sperandio M,Fässler R,Moser M
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Sep 06 00:00:00 EDT 2018
Positive and Negative Regulatory Roles of C-Terminal Src Kinase (CSK) in FcεRI-Mediated Mast Cell Activation, Independent of the Transmembrane Adaptor PAG/CSK-Binding Protein.
Frontiers in immunology
Potuckova L,Draberova L,Halova I,Paulenda T,Draber P
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Oct 07 00:00:00 EDT 2019
Myeloid progenitor cluster formation drives emergency and leukaemic myelopoiesis.
Nature
Hérault A,Binnewies M,Leong S,Calero-Nieto FJ,Zhang SY,Kang YA,Wang X,Pietras EM,Chu SH,Barry-Holson K,Armstrong S,Göttgens B,Passegué E
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Apr 06 00:00:00 EDT 2017
GATA Factor-Regulated Samd14 Enhancer Confers Red Blood Cell Regeneration and Survival in Severe Anemia.
Developmental cell
Hewitt KJ,Katsumura KR,Matson DR,Devadas P,Tanimura N,Hebert AS,Coon JJ,Kim JS,Dewey CN,Keles S,Hao S,Paulson RF,Bresnick EH
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Aug 07 00:00:00 EDT 2017
Eosinophils are required to suppress Th2 responses in Peyer's patches during intestinal infection by nematodes.
Mucosal immunology
Strandmark J,Steinfelder S,Berek C,Kühl AA,Rausch S,Hartmann S
11-1171 was used in Flow cytometry/Cell sorting to study whether eosinophils affect the local T-cell and antibody response in the gut-associated lymphoid tissue during enteric infections.
Mon May 01 00:00:00 EDT 2017
Erythro-myeloid progenitors can differentiate from endothelial cells and modulate embryonic vascular remodeling.
Scientific reports
Kasaai B,Caolo V,Peacock HM,Lehoux S,Gomez-Perdiguero E,Luttun A,Jones EA
11117182 was used in immunocytochemistry to show erythro-myeloid progenitors directly differentiate into endothelial cells
Wed Mar 08 00:00:00 EST 2017
Erythro-myeloid progenitors can differentiate from endothelial cells and modulate embryonic vascular remodeling.
Scientific reports
Kasaai B,Caolo V,Peacock HM,Lehoux S,Gomez-Perdiguero E,Luttun A,Jones EA
11117182 was used in immunocytochemistry to show erythro-myeloid progenitors directly differentiate into endothelial cells
Wed Mar 08 00:00:00 EST 2017
Erythro-myeloid progenitors can differentiate from endothelial cells and modulate embryonic vascular remodeling.
Scientific reports
Kasaai B,Caolo V,Peacock HM,Lehoux S,Gomez-Perdiguero E,Luttun A,Jones EA
11117182 was used in immunocytochemistry to show erythro-myeloid progenitors directly differentiate into endothelial cells
Wed Mar 08 00:00:00 EST 2017
Targeted genome editing restores T cell differentiation in a humanized X-SCID pluripotent stem cell disease model.
Scientific reports
Alzubi J,Pallant C,Mussolino C,Howe SJ,Thrasher AJ,Cathomen T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Sep 29 00:00:00 EDT 2017
Sorting protein VPS33B regulates exosomal autocrine signaling to mediate hematopoiesis and leukemogenesis.
The Journal of clinical investigation
Gu H,Chen C,Hao X,Wang C,Zhang X,Li Z,Shao H,Zeng H,Yu Z,Xie L,Xia F,Zhang F,Liu X,Zhang Y,Jiang H,Zhu J,Wan J,Wang C,Weng W,Xie J,Tao M,Zhang CC,Liu J,Chen GQ,Zheng J
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
A New Method to Stabilize C-Kit Expression in Reparative Cardiac Mesenchymal Cells.
Frontiers in cell and developmental biology
Wysoczynski M,Dassanayaka S,Zafir A,Ghafghazi S,Long BW,Noble C,DeMartino AM,Brittian KR,Bolli R,Jones SP
11-1171 was used in Magnetic cell separation to investigate a new method for c-kit expressing cell isolation and stabilisation in reparative cardiac mesenchymal cells.
Thu Aug 18 00:00:00 EDT 2016
Induction of Indoleamine 2,3-dioxygenase by Pre-treatment with Poly(I:C) May Enhance the Efficacy of MSC Treatment in DSS-induced Colitis.
Immune network
Ryu DB,Lim JY,Lee SE,Park G,Min CK
11-1171 was used in Flow cytometry/Cell sorting to determine whether TLR ligands can enhance the therapeutic efficacy of mesenchymal stem cells for treating inflammatory disorders.
Thu Dec 01 00:00:00 EST 2016
Physiological and hypoxic oxygen concentration differentially regulates human c-Kit+ cardiac stem cell proliferation and migration.
American journal of physiology. Heart and circulatory physiology
Bellio MA,Rodrigues CO,Landin AM,Hatzistergos KE,Kuznetsov J,Florea V,Valasaki K,Khan A,Hare JM,Schulman IH
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Arrested Hematopoiesis and Vascular Relaxation Defects in Mice with a Mutation in Dhfr.
Molecular and cellular biology
Thoms JA,Knezevic K,Liu JJ,Glaros EN,Thai T,Qiao Q,Campbell H,Packham D,Huang Y,Papathanasiou P,Tunningley R,Whittle B,Yeung AW,Chandrakanthan V,Hesson L,Chen V,Wong JW,Purton LE,Ward RL,Thomas SR,Pimanda JE
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Apr 01 00:00:00 EDT 2016
New Regulatory Roles of Galectin-3 in High-Affinity IgE Receptor Signaling.
Molecular and cellular biology
Bambouskova M,Polakovicova I,Halova I,Goel G,Draberova L,Bugajev V,Doan A,Utekal P,Gardet A,Xavier RJ,Draber P
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
The miR-17∼92 cluster contributes to MLL leukemia through the repression of MEIS1 competitor PKNOX1.
Leukemia research
Mian YA,Zeleznik-Le NJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
PRDM14 promotes RAG-dependent Notch1 driver mutations in mouse T-ALL.
Biology open
Carofino BL,Ayanga B,Tracey LJ,Brooke-Bisschop T,Justice MJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Sun May 15 00:00:00 EDT 2016
BMP and Hedgehog Regulate Distinct AGM Hematopoietic Stem Cells Ex Vivo.
Stem cell reports
Crisan M,Solaimani Kartalaei P,Neagu A,Karkanpouna S,Yamada-Inagawa T,Purini C,Vink CS,van der Linden R,van Ijcken W,Chuva de Sousa Lopes SM,Monteiro R,Mummery C,Dzierzak E
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Mar 08 00:00:00 EST 2016
LTβR controls thymic portal endothelial cells for haematopoietic progenitor cell homing and T-cell regeneration.
Nature communications
Shi Y,Wu W,Chai Q,Li Q,Hou Y,Xia H,Ren B,Xu H,Guo X,Jin C,Lv M,Wang Z,Fu YX,Zhu M
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Aug 05 00:00:00 EDT 2016
Deep imaging of bone marrow shows non-dividing stem cells are mainly perisinusoidal.
Nature
Acar M,Kocherlakota KS,Murphy MM,Peyer JG,Oguro H,Inra CN,Jaiyeola C,Zhao Z,Luby-Phelps K,Morrison SJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunofluorescence
Thu Oct 01 00:00:00 EDT 2015
Deep imaging of bone marrow shows non-dividing stem cells are mainly perisinusoidal.
Nature
Acar M,Kocherlakota KS,Murphy MM,Peyer JG,Oguro H,Inra CN,Jaiyeola C,Zhao Z,Luby-Phelps K,Morrison SJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunofluorescence
Thu Oct 01 00:00:00 EDT 2015
Mast cells mediate malignant pleural effusion formation.
The Journal of clinical investigation
Giannou AD,Marazioti A,Spella M,Kanellakis NI,Apostolopoulou H,Psallidas I,Prijovich ZM,Vreka M,Zazara DE,Lilis I,Papaleonidopoulos V,Kairi CA,Patmanidi AL,Giopanou I,Spiropoulou N,Harokopos V,Aidinis V,Spyratos D,Teliousi S,Papadaki H,Taraviras S,Snyder LA,Eickelberg O,Kardamakis D,Iwakura Y,Feyerabend TB,Rodewald HR,Kalomenidis I,Blackwell TS,Agalioti T,Stathopoulos GT
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Mast cells mediate malignant pleural effusion formation.
The Journal of clinical investigation
Giannou AD,Marazioti A,Spella M,Kanellakis NI,Apostolopoulou H,Psallidas I,Prijovich ZM,Vreka M,Zazara DE,Lilis I,Papaleonidopoulos V,Kairi CA,Patmanidi AL,Giopanou I,Spiropoulou N,Harokopos V,Aidinis V,Spyratos D,Teliousi S,Papadaki H,Taraviras S,Snyder LA,Eickelberg O,Kardamakis D,Iwakura Y,Feyerabend TB,Rodewald HR,Kalomenidis I,Blackwell TS,Agalioti T,Stathopoulos GT
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Endothelium and NOTCH specify and amplify aorta-gonad-mesonephros-derived hematopoietic stem cells.
The Journal of clinical investigation
Hadland BK,Varnum-Finney B,Poulos MG,Moon RT,Butler JM,Rafii S,Bernstein ID
11-1171 was used in Flow cytometry/Cell sorting to investigate the critical niche components and resident signals required for hematopoietic stem cell induction and self-renewal ex vivo.
Fri May 01 00:00:00 EDT 2015
Cis-regulatory mechanisms governing stem and progenitor cell transitions.
Science advances
Johnson KD,Kong G,Gao X,Chang YI,Hewitt KJ,Sanalkumar R,Prathibha R,Ranheim EA,Dewey CN,Zhang J,Bresnick EH
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Mast cells boost myeloid-derived suppressor cell activity and contribute to the development of tumor-favoring microenvironment.
Cancer immunology research
Danelli L,Frossi B,Gri G,Mion F,Guarnotta C,Bongiovanni L,Tripodo C,Mariuzzi L,Marzinotto S,Rigoni A,Blank U,Colombo MP,Pucillo CE
11-1171 was used in Flow cytometry/Cell sorting to examine the interactions between myeloid-derived suppressor cells and mast cells in the mucosa of patients with colon carcinoma.
Thu Jan 01 00:00:00 EST 2015
Spred1, a Suppressor of the Ras-ERK Pathway, Negatively Regulates Expansion and Function of Group 2 Innate Lymphoid Cells.
Journal of immunology (Baltimore, Md. : 1950)
Suzuki M,Morita R,Hirata Y,Shichita T,Yoshimura A
11-1171 was used in Flow cytometry/Cell sorting to demonstrate that Spred1, a negative regulator of the Ras-ERK pathway, plays an important role in the proliferation and apoptosis of ILC2s and in cytokine secretion from ILC2s.
Sat Aug 01 00:00:00 EDT 2015
Mouse bone marrow-derived mesenchymal stem cells inhibit leukemia/lymphoma cell proliferation in vitro and in a mouse model of allogeneic bone marrow transplant.
International journal of molecular medicine
Song N,Gao L,Qiu H,Huang C,Cheng H,Zhou H,Lv S,Chen L,Wang J
11-1171 was used in Flow cytometry/Cell sorting to determine that MSCs may be safe and effective when used in allogeneic BMT for the treatment of haemotological malignancies.
Wed Jul 01 00:00:00 EDT 2015
Chlamydia pneumoniae promotes dysfunction of pancreatic beta cells.
Cellular immunology
Rodriguez AR,Plascencia-Villa G,Witt CM,Yu JJ,José-Yacamán M,Chambers JP,Perry G,Guentzel MN,Arulanandam BP
11-1171 was used in Flow cytometry/Cell sorting to reveal the negative effect of C. pneumoniae on mast cells, and the consequential impact on pancreatic beta cell function and viability.
Mon Jun 01 00:00:00 EDT 2015
Merlin Isoforms 1 and 2 Both Act as Tumour Suppressors and Are Required for Optimal Sperm Maturation.
PloS one
Zoch A,Mayerl S,Schulz A,Greither T,Frappart L,Rübsam J,Heuer H,Giovannini M,Morrison H
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon May 23 00:00:00 EDT 2016
Notch signal strength controls cell fate in the haemogenic endothelium.
Nature communications
Gama-Norton L,Ferrando E,Ruiz-Herguido C,Liu Z,Guiu J,Islam AB,Lee SU,Yan M,Guidos CJ,López-Bigas N,Maeda T,Espinosa L,Kopan R,Bigas A
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunofluorescence
Wed Oct 14 00:00:00 EDT 2015
Abi3bp regulates cardiac progenitor cell proliferation and differentiation.
Circulation research
Hodgkinson CP,Gomez JA,Payne AJ,Zhang L,Wang X,Dal-Pra S,Pratt RE,Dzau VJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Dec 05 00:00:00 EST 2014
Abi3bp regulates cardiac progenitor cell proliferation and differentiation.
Circulation research
Hodgkinson CP,Gomez JA,Payne AJ,Zhang L,Wang X,Dal-Pra S,Pratt RE,Dzau VJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Dec 05 00:00:00 EST 2014
WASH is required for the differentiation commitment of hematopoietic stem cells in a c-Myc-dependent manner.
The Journal of experimental medicine
Xia P,Wang S,Huang G,Zhu P,Li M,Ye B,Du Y,Fan Z
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Sep 22 00:00:00 EDT 2014
Pharmacological mobilization of endogenous stem cells significantly promotes skin regeneration after full-thickness excision: the synergistic activity of AMD3100 and tacrolimus.
The Journal of investigative dermatology
Lin Q,Wesson RN,Maeda H,Wang Y,Cui Z,Liu JO,Cameron AM,Gao B,Montgomery RA,Williams GM,Sun Z
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
A crucial role for the ubiquitously expressed transcription factor Sp1 at early stages of hematopoietic specification.
Development (Cambridge, England)
Gilmour J,Assi SA,Jaegle U,Kulu D,van de Werken H,Clarke D,Westhead DR,Philipsen S,Bonifer C
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
Identification of hematopoietic-specific regulatory elements from the CD45 gene and use for lentiviral tracking of transplanted cells.
Experimental hematology
Duong KL,Das S,Yu S,Barr JY,Jena S,Kim E,Zavazava N,Colgan JD,Xue HH,Levasseur DN
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
F-box protein FBXL16 binds PP2A-B55α and regulates differentiation of embryonic stem cells along the FLK1+ lineage.
Molecular & cellular proteomics : MCP
Honarpour N,Rose CM,Brumbaugh J,Anderson J,Graham RL,Sweredoski MJ,Hess S,Coon JJ,Deshaies RJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Sat Mar 01 00:00:00 EST 2014
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
The expression of Sox17 identifies and regulates haemogenic endothelium.
Nature cell biology
Clarke RL,Yzaguirre AD,Yashiro-Ohtani Y,Bondue A,Blanpain C,Pear WS,Speck NA,Keller G
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013
CX(3)CR1(+) macrophages support IL-22 production by innate lymphoid cells during infection with Citrobacter rodentium.
Mucosal immunology
Manta C,Heupel E,Radulovic K,Rossini V,Garbi N,Riedel CU,Niess JH
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Distinct development and functions of resident and recruited liver Kupffer cells/macrophages.
Journal of leukocyte biology
Ikarashi M,Nakashima H,Kinoshita M,Sato A,Nakashima M,Miyazaki H,Nishiyama K,Yamamoto J,Seki S
11-1171 was used in Flow cytometry/Cell sorting to study how liver CD68+ Kupffer cells and CD11b+ Kupffer cells/macrophages are developmentally and functionally distinct subsets.
Sun Dec 01 00:00:00 EST 2013
A mouse model for inducible overexpression of Prdm14 results in rapid-onset and highly penetrant T-cell acute lymphoblastic leukemia (T-ALL).
Disease models & mechanisms
Carofino BL,Ayanga B,Justice MJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2013
Calcitonin gene-related peptide and cyclic adenosine 5'-monophosphate/protein kinase A pathway promote IL-9 production in Th9 differentiation process.
Journal of immunology (Baltimore, Md. : 1950)
Mikami N,Miyagi Y,Sueda K,Takatsuji M,Fukada S,Yamamoto H,Tsujikawa K
11-1171 was used in Flow cytometry/Cell sorting to reveal a novel regulatory mechanism comprising G protein-coupled receptor ligands and nervous system-derived substances in Th9 cell differentiation and type I hypersensitivity.
Mon Apr 15 00:00:00 EDT 2013
NK cells impede glioblastoma virotherapy through NKp30 and NKp46 natural cytotoxicity receptors.
Nature medicine
Alvarez-Breckenridge CA,Yu J,Price R,Wojton J,Pradarelli J,Mao H,Wei M,Wang Y,He S,Hardcastle J,Fernandez SA,Kaur B,Lawler SE,Vivier E,Mandelboim O,Moretta A,Caligiuri MA,Chiocca EA
11-1171 was used in Flow cytometry/Cell sorting to demonstrate that glioblastoma virotherapy is limited partially by an antiviral NK cell response involving specific NCRs, uncovering new potential targets to enhance cancer virotherapy.
Sat Dec 01 00:00:00 EST 2012
IKKα-mediated signaling circuitry regulates early B lymphopoiesis during hematopoiesis.
Blood
Balkhi MY,Willette-Brown J,Zhu F,Chen Z,Liu S,Guttridge DC,Karin M,Hu Y
11-1171-82 was used in Flow Cytometry to demonstrate that IKKα regulates B-lineage commitment via combined canonical and noncanonical NF-κB transcriptional activities to target Pax5 expression during hematopoiesis.
Thu Jun 07 00:00:00 EDT 2012
CTC1 deletion results in defective telomere replication, leading to catastrophic telomere loss and stem cell exhaustion.
The EMBO journal
Gu P,Min JN,Wang Y,Huang C,Peng T,Chai W,Chang S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Wed May 16 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunofluorescence
Wed Aug 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunofluorescence
Wed Aug 01 00:00:00 EDT 2012
Mast cell TLR2 signaling is crucial for effective killing of Francisella tularensis.
Journal of immunology (Baltimore, Md. : 1950)
Rodriguez AR,Yu JJ,Guentzel MN,Navara CS,Klose KE,Forsthuber TG,Chambers JP,Berton MT,Arulanandam BP
11-1171 was used in Immunocytochemistry-immunoflourescence to provide evidence of mast cells and TLR2-mediated signalling on early innate processes in the lung following pulmonary F. tularensis infection.
Fri Jun 01 00:00:00 EDT 2012
Mast cell TLR2 signaling is crucial for effective killing of Francisella tularensis.
Journal of immunology (Baltimore, Md. : 1950)
Rodriguez AR,Yu JJ,Guentzel MN,Navara CS,Klose KE,Forsthuber TG,Chambers JP,Berton MT,Arulanandam BP
11-1171 was used in Immunocytochemistry-immunoflourescence to provide evidence of mast cells and TLR2-mediated signalling on early innate processes in the lung following pulmonary F. tularensis infection.
Fri Jun 01 00:00:00 EDT 2012
Partial characterization of the Sox2+ cell population in an adult murine model of digit amputation.
Tissue engineering. Part A
Agrawal V,Siu BF,Chao H,Hirschi KK,Raborn E,Johnson SA,Tottey S,Hurley KB,Medberry CJ,Badylak SF
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2012
RNAi screen identifies Brd4 as a therapeutic target in acute myeloid leukaemia.
Nature
Zuber J,Shi J,Wang E,Rappaport AR,Herrmann H,Sison EA,Magoon D,Qi J,Blatt K,Wunderlich M,Taylor MJ,Johns C,Chicas A,Mulloy JC,Kogan SC,Brown P,Valent P,Bradner JE,Lowe SW,Vakoc CR
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Wed Aug 03 00:00:00 EDT 2011
Essential roles for Pot1b in HSC self-renewal and survival.
Blood
Wang Y,Shen MF,Chang S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011
Smad1 signaling restricts hematopoietic potential after promoting hemangioblast commitment.
Blood
Cook BD,Liu S,Evans T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Jun 16 00:00:00 EDT 2011
Mast cell/IL-4 control of Francisella tularensis replication and host cell death is associated with increased ATP production and phagosomal acidification.
Mucosal immunology
Rodriguez AR,Yu JJ,Murthy AK,Guentzel MN,Klose KE,Forsthuber TG,Chambers JP,Berton MT,Arulanandam BP
11-1171 was used in Flow cytometry/Cell sorting to demonstrate a plausible mechanism by which IL-4-mediated inhibition of F. tularensis replication is associated with increased ATP production and improved acidification of organelles containing bacteria.
Tue Mar 01 00:00:00 EST 2011
Identification of a novel antigen cross-presenting cell type in spleen.
Journal of cellular and molecular medicine
Tan JK,Quah BJ,Griffiths KL,Periasamy P,Hey YY,O'Neill HC
11-1171 was used in Flow cytometry/Cell sorting to study a novel DC type produced by in vitro haematopoiesis which has distinct antigen-presenting potential and reflects a DC subset present also in vivo in spleen.
Sun May 01 00:00:00 EDT 2011
Intramuscular VEGF activates an SDF1-dependent progenitor cell cascade and an SDF1-independent muscle paracrine cascade for cardiac repair.
American journal of physiology. Heart and circulatory physiology
Zisa D,Shabbir A,Mastri M,Taylor T,Aleksic I,McDaniel M,Suzuki G,Lee T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011
Sox3 functions in a cell-autonomous manner to regulate spermatogonial differentiation in mice.
Endocrinology
Laronda MM,Jameson JL
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Apr 01 00:00:00 EDT 2011
Contribution of IL-13 to early exocrinopathy in Id3-/- mice.
Molecular immunology
Mahlios J,Zhuang Y
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunofluorescence
Sat Oct 01 00:00:00 EDT 2011
A high omega-3 fatty acid diet has different effects on early and late stage myeloid progenitors.
Lipids
Varney ME,Buchanan JT,Dementieva Y,Hardman WE,Sollars VE
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2011
Decitabine maintains hematopoietic precursor self-renewal by preventing repression of stem cell genes by a differentiation-inducing stimulus.
Molecular cancer therapeutics
Hu Z,Negrotto S,Gu X,Mahfouz R,Ng KP,Ebrahem Q,Copelan E,Singh H,Maciejewski JP,Saunthararajah Y
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2010
Impaired mobilization of hematopoietic stem/progenitor cells in C5-deficient mice supports the pivotal involvement of innate immunity in this process and reveals novel promobilization effects of granulocytes.
Leukemia
Lee HM,Wu W,Wysoczynski M,Liu R,Zuba-Surma EK,Kucia M,Ratajczak J,Ratajczak MZ
11-1171 was used in Flow cytometry/Cell sorting to examine the central role of innate immunity in the mobilisation of haematopoietic stem/progenitor cells.
Sun Nov 01 00:00:00 EDT 2009
Heart failure therapy mediated by the trophic activities of bone marrow mesenchymal stem cells: a noninvasive therapeutic regimen.
American journal of physiology. Heart and circulatory physiology
Shabbir A,Zisa D,Suzuki G,Lee T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2009
Intramuscular VEGF repairs the failing heart: role of host-derived growth factors and mobilization of progenitor cells.
American journal of physiology. Regulatory, integrative and comparative physiology
Zisa D,Shabbir A,Mastri M,Suzuki G,Lee T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Immunofluorescence
Sun Nov 01 00:00:00 EDT 2009
Reporter gene-expressing bone marrow-derived stromal cells are immune-tolerated following implantation in the central nervous system of syngeneic immunocompetent mice.
BMC biotechnology
Bergwerf I,De Vocht N,Tambuyzer B,Verschueren J,Reekmans K,Daans J,Ibrahimi A,Van Tendeloo V,Chatterjee S,Goossens H,Jorens PG,Baekelandt V,Ysebaert D,Van Marck E,Berneman ZN,Linden AV,Ponsaerts P
11-1171 was used in Flow cytometry/Cell sorting to implant reporter gene-expression bone marrow-derived stromal cells in the central nervous system of syngeneic immunocompetent mice.
Wed Jan 07 00:00:00 EST 2009
CD4+CD25+ regulatory T cells suppress mast cell degranulation and allergic responses through OX40-OX40L interaction.
Immunity
Gri G,Piconese S,Frossi B,Manfroi V,Merluzzi S,Tripodo C,Viola A,Odom S,Rivera J,Colombo MP,Pucillo CE
11-1171 was used in Flow cytometry/Cell sorting to explore whether Treg cells influence the immediate hypersensitivity response of mast cells.
Fri Nov 14 00:00:00 EST 2008
Identification of chromatin remodeling genes Arid4a and Arid4b as leukemia suppressor genes.
Journal of the National Cancer Institute
Wu MY,Eldin KW,Beaudet AL
11-1171 was used in Flow cytometry/Cell sorting to investigate the role of epigenetic alterations in tumorigenesis.
Wed Sep 03 00:00:00 EDT 2008
Analysis of the IKKbeta/NF-kappaB signaling pathway during embryonic angiogenesis.
Developmental dynamics : an official publication of the American Association of Anatomists
Hou Y,Li F,Karin M,Ostrowski MC
11-1171 was used in Flow cytometry/Cell sorting to describe the IKKβ/NF-κB signalling pathway involved in embryonic angiogenesis, which Ikkβ locus disruption in foetal liver vasculature revealed to be responsible for regulation of hepatocyte proliferation and apoptosis, however not for hematopoiesis.
Wed Oct 01 00:00:00 EDT 2008
Essential role of phospholipase C gamma 2 in early B-cell development and Myc-mediated lymphomagenesis.
Molecular and cellular biology
Wen R,Chen Y,Bai L,Fu G,Schuman J,Dai X,Zeng H,Yang C,Stephan RP,Cleveland JL,Wang D
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 11-1171-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2006