Product References
ABIN1 (Q478) is Required to Prevent Hematopoietic Deficiencies through Regulating Type I IFNs Expression.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wu X,Wang Y,Chen B,Liu Y,Li F,Ou Y,Zhang H,Wu X,Li X,Wang L,Rong W,Liu J,Xing M,Zhao X,Liu H,Ge L,Lv A,Wang L,Wang Z,Li M,Zhang H
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Mon Jan 01 00:00:00 UTC 2024
Engineered bone marrow as a clinically relevant ex vivo model for primary bone cancer research and drug screening.
Proceedings of the National Academy of Sciences of the United States of America
Griffin KH,Thorpe SW,Sebastian A,Hum NR,Coonan TP,Sagheb IS,Loots GG,Randall RL,Leach JK
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Tue Sep 26 00:00:00 UTC 2023
Landscape of mast cell populations across organs in mice and humans.
The Journal of experimental medicine
Tauber M,Basso L,Martin J,Bostan L,Pinto MM,Thierry GR,Houmadi R,Serhan N,Loste A,Blériot C,Kamphuis JBJ,Grujic M,Kjellén L,Pejler G,Paul C,Dong X,Galli SJ,Reber LL,Ginhoux F,Bajenoff M,Gentek R,Gaudenzio N
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Mon Oct 02 00:00:00 UTC 2023
Morphology of migrating telocytes and their potential role in stem cell differentiation during cartilage development in catfish (Clarias gariepinus).
Microscopy research and technique
Abd-Elhafeez HH,Rutland CS,Soliman SA
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunohistochemistry
Fri Sep 01 00:00:00 UTC 2023
Two distinctive types of telocytes in gills of fish: A light, immunohistochemical and ultra-structure study.
Microscopy research and technique
Soliman SA,Sobh A,Ali LA,Abd-Elhafeez HH
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunohistochemistry
Tue Nov 01 00:00:00 UTC 2022
The AML-associated K313 mutation enhances C/EBPα activity by leading to C/EBPα overexpression.
Cell death & disease
Gentle IE,Moelter I,Badr MT,Döhner K,Lübbert M,Häcker G
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Mon Jul 05 00:00:00 UTC 2021
The lifespan quantitative trait locus gene Securin controls hematopoietic progenitor cell function.
Haematologica
Brown A,Schuetz D,Han Y,Daria D,Nattamai KJ,Eiwen K,Sakk V,Pospiech J,Saller T,van Zant G,Wagner W,Geiger H
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Tue Apr 27 00:00:00 UTC 2021
The lifespan quantitative trait locus gene Securin controls hematopoietic progenitor cell function.
Haematologica
Brown A,Schuetz D,Han Y,Daria D,Nattamai KJ,Eiwen K,Sakk V,Pospiech J,Saller T,van Zant G,Wagner W,Geiger H
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Tue Apr 27 00:00:00 UTC 2021
Pyruvate kinase M2 in lung APCs regulates Alternaria-induced airway inflammation.
Immunobiology
Jaiswal AK,Makhija S,Stahr N,Sandey M,Suryawanshi A,Mishra A
56-1172-82 was used in Flow Cytometry to support a model in which Alternaria sensitization in airways induce a circuitry of glycolysis and PKM2 regulation that confers an acute activation of APCs in the lung, whose targeting might represent a strategy for asthma treatment.
Wed Jul 01 00:00:00 UTC 2020
Morphological and immunohistochemical phenotype of TCs in the intestinal bulb of Grass carp and their potential role in intestinal immunity.
Scientific reports
Abd-Elhafeez HH,Abou-Elhamd AS,Soliman SA
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunohistochemistry
Thu Aug 20 00:00:00 UTC 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Thu Sep 17 00:00:00 UTC 2020
Inhibition of FGF2-Mediated Signaling in GIST-Promising Approach for Overcoming Resistance to Imatinib.
Cancers
Boichuk S,Galembikova A,Mikheeva E,Bikinieva F,Aukhadieva A,Dunaev P,Khalikov D,Petrov S,Kurtasanov R,Valeeva E,Kireev I,Dugina V,Lushnikova A,Novikova M,Kopnin P
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunohistochemistry
Wed Jun 24 00:00:00 UTC 2020
Fish telocytes and their relation to rodlet cells in ruby-red-fin shark (rainbow shark) Epalzeorhynchos frenatum (Teleostei: Cyprinidae).
Scientific reports
Abd-Elhafeez HH,Abdo W,Kamal BM,Soliman SA
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunohistochemistry
Tue Nov 03 00:00:00 UTC 2020
Loss of FGFR3 Delays Acute Myeloid Leukemogenesis by Programming Weakly Pathogenic CD117-Positive Leukemia Stem-Like Cells.
Frontiers in pharmacology
Guo C,Ran Q,Sun C,Zhou T,Yang X,Zhang J,Pang S,Xiao Y
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Tue Feb 16 00:00:00 UTC 2021
Id1 and Id3 Maintain Steady-State Hematopoiesis by Promoting Sinusoidal Endothelial Cell Survival and Regeneration.
Cell reports
Gadomski S,Singh SK,Singh S,Sarkar T,Klarmann KD,Berenschot M,Seaman S,Jakubison B,Gudmundsson KO,Lockett S,Keller JR
56-1172 was used in Flow cytometry/Cell sorting to investigate the effects of loss of ld genes on adult endothelial cell.
Tue Apr 28 00:00:00 UTC 2020
Hsd3b2 associated in modulating steroid hormone synthesis pathway regulates the differentiation of chicken embryonic stem cells into spermatogonial stem cells.
Journal of cellular biochemistry
Zhang C,Wang M,He N,Ahmed MF,Wang Y,Zhao R,Yu X,Jin J,Song J,Zuo Q,Zhang Y,Li B
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunocytochemistry
Mon Jan 01 00:00:00 UTC 2018
Electroacupuncture at ST36 Increases Bone Marrow-Derived Interstitial Cells of Cajal via the SDF-1/CXCR4 and mSCF/Kit-ETV1 Pathways in the Stomach of Diabetic Mice.
Evidence-based complementary and alternative medicine : eCAM
Zhao J,An J,Liu S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunofluorescence
Wed Mar 16 00:00:00 UTC 2022
Bioengineering functional smooth muscle with spontaneous rhythmic contraction in vitro.
Scientific reports
Kobayashi M,Khalil HA,Lei NY,Wang Q,Wang K,Wu BM,Dunn JCY
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunofluorescence
Mon Sep 10 00:00:00 UTC 2018
GATA4-dependent organ-specific endothelial differentiation controls liver development and embryonic hematopoiesis.
The Journal of clinical investigation
Géraud C,Koch PS,Zierow J,Klapproth K,Busch K,Olsavszky V,Leibing T,Demory A,Ulbrich F,Diett M,Singh S,Sticht C,Breitkopf-Heinlein K,Richter K,Karppinen SM,Pihlajaniemi T,Arnold B,Rodewald HR,Augustin HG,Schledzewski K,Goerdt S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
Conditional genetic deletion of Ano1 in interstitial cells of Cajal impairs Ca2+ transients and slow waves in adult mouse small intestine.
American journal of physiology. Gastrointestinal and liver physiology
Malysz J,Gibbons SJ,Saravanaperumal SA,Du P,Eisenman ST,Cao C,Oh U,Saur D,Klein S,Ordog T,Farrugia G
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunofluorescence
Wed Mar 01 00:00:00 UTC 2017
Low Testosterone Alters the Activity of Mouse Prostate Stem Cells.
The Prostate
Zhou Y,Copeland B,Otto-Duessel M,He M,Markel S,Synold TW,Jones JO
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2017
A Functional Bioluminescent Zebrafish Screen for Enhancing Hematopoietic Cell Homing.
Stem cell reports
Astuti Y,Kramer AC,Blake AL,Blazar BR,Tolar J,Taisto ME,Lund TC
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Tue Jan 10 00:00:00 UTC 2017
Mutant U2AF1-expressing cells are sensitive to pharmacological modulation of the spliceosome.
Nature communications
Shirai CL,White BS,Tripathi M,Tapia R,Ley JN,Ndonwi M,Kim S,Shao J,Carver A,Saez B,Fulton RS,Fronick C,O'Laughlin M,Lagisetti C,Webb TR,Graubert TA,Walter MJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Mon Jan 09 00:00:00 UTC 2017
Longitudinal intravital imaging of the femoral bone marrow reveals plasticity within marrow vasculature.
Nature communications
Reismann D,Stefanowski J,Günther R,Rakhymzhan A,Matthys R,Nützi R,Zehentmeier S,Schmidt-Bleek K,Petkau G,Chang HD,Naundorf S,Winter Y,Melchers F,Duda G,Hauser AE,Niesner RA
56-1172 was used in Flow cytometry/Cell sorting to develop longitudinal intravital imaging of the bone marrow to analyse cellular dynamics within the deep marrow.
Mon Dec 18 00:00:00 UTC 2017
Core Circadian Clock Genes Regulate Leukemia Stem Cells in AML.
Cell
Puram RV,Kowalczyk MS,de Boer CG,Schneider RK,Miller PG,McConkey M,Tothova Z,Tejero H,Heckl D,Järås M,Chen MC,Li H,Tamayo A,Cowley GS,Rozenblatt-Rosen O,Al-Shahrour F,Regev A,Ebert BL
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Thu Apr 07 00:00:00 UTC 2016
Stimulus-induced pacemaker activity in interstitial cells of Cajal associated with the deep muscular plexus of the small intestine.
Neurogastroenterology and motility
Zhu YF,Wang XY,Parsons SP,Huizinga JD
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunocytochemistry
Fri Jul 01 00:00:00 UTC 2016
Regulation of B-cell development and tolerance by different members of the miR-17∼92 family microRNAs.
Nature communications
Lai M,Gonzalez-Martin A,Cooper AB,Oda H,Jin HY,Shepherd J,He L,Zhu J,Nemazee D,Xiao C
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Tue Aug 02 00:00:00 UTC 2016
Different growth and metastatic phenotypes associated with a cell-intrinsic change of Met in metastatic melanoma.
Oncotarget
Adachi E,Sakai K,Nishiuchi T,Imamura R,Sato H,Matsumoto K
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Tue Oct 25 00:00:00 UTC 2016
ILC3 GM-CSF production and mobilisation orchestrate acute intestinal inflammation.
eLife
Pearson C,Thornton EE,McKenzie B,Schaupp AL,Huskens N,Griseri T,West N,Tung S,Seddon BP,Uhlig HH,Powrie F
56-1172 was used in Flow cytometry/Cell sorting to investigate how a small population of innate lymphoid cells (ILCs) has large effects on immune homeostasis, showing that ILC3s produce GM-CSF to orchestrate acute intestinal inflammation.
Mon Jan 18 00:00:00 UTC 2016
The origin of segmentation motor activity in the intestine.
Nature communications
Huizinga JD,Chen JH,Zhu YF,Pawelka A,McGinn RJ,Bardakjian BL,Parsons SP,Kunze WA,Wu RY,Bercik P,Khoshdel A,Chen S,Yin S,Zhang Q,Yu Y,Gao Q,Li K,Hu X,Zarate N,Collins P,Pistilli M,Ma J,Zhang R,Chen D
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunofluorescence
Mon Nov 02 00:00:00 UTC 2015
TRPA1-dependent pruritus in IL-13-induced chronic atopic dermatitis.
Journal of immunology (Baltimore, Md. : 1950)
Oh MH,Oh SY,Lu J,Lou H,Myers AC,Zhu Z,Zheng T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Sun Dec 01 00:00:00 UTC 2013
TRPA1-dependent pruritus in IL-13-induced chronic atopic dermatitis.
Journal of immunology (Baltimore, Md. : 1950)
Oh MH,Oh SY,Lu J,Lou H,Myers AC,Zhu Z,Zheng T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Flow Cytometry
Sun Dec 01 00:00:00 UTC 2013
Mast cell deficiency attenuates progression of atherosclerosis and hepatic steatosis in apolipoprotein E-null mice.
American journal of physiology. Heart and circulatory physiology
Smith DD,Tan X,Raveendran VV,Tawfik O,Stechschulte DJ,Dileepan KN
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunohistochemistry
Fri Jun 15 00:00:00 UTC 2012
An information theoretic, microfluidic-based single cell analysis permits identification of subpopulations among putatively homogeneous stem cells.
PloS one
Glotzbach JP,Januszyk M,Vial IN,Wong VW,Gelbard A,Kalisky T,Thangarajah H,Longaker MT,Quake SR,Chu G,Gurtner GC
56-1172 was used in Flow cytometry/Cell sorting to develop a novel method to characterize and predict transcriptional programs across individual homogeneous stem cells.
Wed Nov 02 00:00:00 UTC 2011
Homeostatic role of transforming growth factor-beta in the oral cavity and esophagus of mice and its expression by mast cells in these tissues.
The American journal of pathology
Vitsky A,Waire J,Pawliuk R,Bond A,Matthews D,Lacasse E,Hawes ML,Nelson C,Richards S,Piepenhagen PA,Garman RD,Andrews L,Thurberg BL,Lonning S,Ledbetter S,Ruzek MC
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 56-1172-82) in Immunofluorescence
Mon Jun 01 00:00:00 UTC 2009