Product References
Senescence Rewires Microenvironment Sensing to Facilitate Antitumor Immunity.
Cancer discovery
Chen HA,Ho YJ,Mezzadra R,Adrover JM,Smolkin R,Zhu C,Woess K,Bernstein N,Schmitt G,Fong L,Luan W,Wuest A,Tian S,Li X,Broderick C,Hendrickson RC,Egeblad M,Chen Z,Alonso-Curbelo D,Lowe SW
Published figure using CD119 (IFN gamma Receptor 1) monoclonal antibody (Product # 12-1191-82) in Flow Cytometry
Mon Feb 06 00:00:00 UTC 2023
STUB1 is an intracellular checkpoint for interferon gamma sensing.
Scientific reports
Ng S,Lim S,Sim ACN,Mangadu R,Lau A,Zhang C,Martinez SB,Chandramohan A,Lim UM,Ho SSW,Chang SC,Gopal P,Hong LZ,Schwaid A,Fernandis AZ,Loboda A,Li C,Phan U,Henry B,Partridge AW
12-1191-82 was used in Flow cytometry/Cell sorting to elucidate STUB1 as a barrier to IFNγ sensing, prompting further investigations to assess if broader inactivation of human STUB1 in both tumors and immune cells could overcome ICB resistance.
Thu Aug 18 00:00:00 UTC 2022
STUB1 is an intracellular checkpoint for interferon gamma sensing.
Scientific reports
Ng S,Lim S,Sim ACN,Mangadu R,Lau A,Zhang C,Martinez SB,Chandramohan A,Lim UM,Ho SSW,Chang SC,Gopal P,Hong LZ,Schwaid A,Fernandis AZ,Loboda A,Li C,Phan U,Henry B,Partridge AW
12-1191-82 was used in Flow cytometry/Cell sorting to elucidate STUB1 as a barrier to IFNγ sensing, prompting further investigations to assess if broader inactivation of human STUB1 in both tumors and immune cells could overcome ICB resistance.
Thu Aug 18 00:00:00 UTC 2022
Tet2 deficiency drives liver microbiome dysbiosis triggering Tc1 cell autoimmune hepatitis.
Cell host & microbe
Pandey SP,Bender MJ,McPherson AC,Phelps CM,Sanchez LM,Rana M,Hedden L,Sangani KA,Chen L,Shapira JH,Siller M,Goel C,Verdú EF,Jabri B,Chang A,Chandran UR,Mullett SJ,Wendell SG,Singhi AD,Tilstra JS,Pierre JF,Arteel GE,Hinterleitner R,Meisel M
12-1191-82 was used in Flow cytometry/Cell sorting to demonstrate that I3A is required for L. reuteri-induced Tc1 cell differentiation in vitro and AIH-like pathology in vivo, both of which are restrained by Tet2 within CD8 T cells.
Wed Jul 13 00:00:00 UTC 2022
Loss of Optineurin Drives Cancer Immune Evasion via Palmitoylation-Dependent IFNGR1 Lysosomal Sorting and Degradation.
Cancer discovery
Du W,Hua F,Li X,Zhang J,Li S,Wang W,Zhou J,Wang W,Liao P,Yan Y,Li G,Wei S,Grove S,Vatan L,Zgodziński W,Majewski M,Wallner G,Chen H,Kryczek I,Fang JY,Zou W
12-1191-82 was used in Flow Cytometry to show that optineurin is a shared node between the IFN and MHC pathways and predicts colorectal cancer patient outcome.
Thu Jul 01 00:00:00 UTC 2021
Mycolactone displays anti-inflammatory effects on the nervous system.
PLoS neglected tropical diseases
Isaac C,Mauborgne A,Grimaldi A,Ade K,Pohl M,Limatola C,Boucher Y,Demangel C,Guenin-Macé L
Published figure using CD119 (IFN gamma Receptor 1) monoclonal antibody (Product # 12-1191-82) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2017
Erythropoietin treatment in murine multiple myeloma: immune gain and bone loss.
Scientific reports
Deshet-Unger N,Hiram-Bab S,Haim-Ohana Y,Mittelman M,Gabet Y,Neumann D
12-1191 was used in Flow cytometry/Cell sorting to reveal that in multiple myeloma, erythropoietin may act as a double-edged sword by stimulating the immune response whilst accelerating bone resorption.
Tue Aug 02 00:00:00 UTC 2016
iNKT Cells Are Responsible for the Apoptotic Reduction of Basophils That Mediate Th2 Immune Responses Elicited by Papain in Mice Following γPGA Stimulation.
PloS one
Park HJ,Lee SW,Park SH,Hong S
12-1191 was used in Flow cytometry/Cell sorting to demonstrate that γPGA-induced iNKT cell polarization toward the Th1 phenotype induces apoptotic basophil depletion, leading to the suppression of Th2 immune responses.
Mon Aug 22 00:00:00 UTC 2016
Chronic oral infection with major periodontal bacteria Tannerella forsythia modulates systemic atherosclerosis risk factors and inflammatory markers.
Pathogens and disease
Chukkapalli SS,Rivera-Kweh MF,Velsko IM,Chen H,Zheng D,Bhattacharyya I,Gangula PR,Lucas AR,Kesavalu L
12-1191 was used in Flow cytometry/Cell sorting to investigate the potential of a known bacterial pathogen found in atherosclerotic plaque in humans to accelerate atherosclerosis in hyperlipdemic mice.
Wed Apr 01 00:00:00 UTC 2015
CD8+ T Cells Mediate Female-Dominant IL-4 Production and Airway Inflammation in Allergic Asthma.
PloS one
Ito C,Okuyama-Dobashi K,Miyasaka T,Masuda C,Sato M,Kawano T,Ohkawara Y,Kikuchi T,Takayanagi M,Ohno I
12-1191 was used in Flow cytometry/Cell sorting to determine the mechanisms behind the sex differences in asthmatic responses after puberty.
Wed Jul 06 00:00:00 UTC 2016
Polymicrobial Oral Infection with Four Periodontal Bacteria Orchestrates a Distinct Inflammatory Response and Atherosclerosis in ApoE null Mice.
PloS one
Chukkapalli SS,Velsko IM,Rivera-Kweh MF,Zheng D,Lucas AR,Kesavalu L
12-1191 was used in Flow cytometry/Cell sorting to study the levels of inflammatory molecules and cells types involved in mounting an immune response to peridontal infection over time.
Fri Jun 17 00:00:00 UTC 2016
A role for the ITAM signaling module in specifying cytokine-receptor functions.
Nature immunology
Bezbradica JS,Rosenstein RK,DeMarco RA,Brodsky I,Medzhitov R
Published figure using CD119 (IFN gamma Receptor 1) monoclonal antibody (Product # 12-1191-82) in Flow Cytometry
Tue Apr 01 00:00:00 UTC 2014
A type I IFN-Flt3 ligand axis augments plasmacytoid dendritic cell development from common lymphoid progenitors.
The Journal of experimental medicine
Chen YL,Chen TT,Pai LM,Wesoly J,Bluyssen HA,Lee CK
12-1191 was used in Flow cytometry/Cell sorting to define a critical role for the FL/IFN-I/Flt3 axis in plasmacytoid dendritic cell differentiation from common lymphoid progenitors.
Mon Nov 18 00:00:00 UTC 2013
Prostaglandin E₂ promotes Th1 differentiation via synergistic amplification of IL-12 signalling by cAMP and PI3-kinase.
Nature communications
Yao C,Hirata T,Soontrapa K,Ma X,Takemori H,Narumiya S
12-1191 was used in Flow cytometry/Cell sorting to investigate the role of cAMP in Th1 differentiation, showing that prostaglandin E2 promotes differentiation via synergistic amplification of IL-12 signalling by cAMP and PI3-kinase.
Tue Oct 22 00:00:00 UTC 2013