Product References
LAL deficiency induced myeloid-derived suppressor cells as targets and biomarkers for lung cancer.
Journal for immunotherapy of cancer
Zhao T,Liu S,Hanna NH,Jalal S,Ding X,Wan J,Yan C,Du H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Topical Diacerein Decreases Skin and Splenic CD11c+ Dendritic Cells in Psoriasis.
International journal of molecular sciences
Brunner SM,Ramspacher A,Rieser C,Leitner J,Heil H,Ablinger M,Tevini J,Wimmer M,Koller A,Piñón Hofbauer J,Felder TK,Bauer JW,Kofler B,Lang R,Wally V
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Feb 21 00:00:00 EST 2023
CD11c+ microglia promote white matter repair after ischemic stroke.
Cell death & disease
Jia J,Zheng L,Ye L,Chen J,Shu S,Xu S,Bao X,Xia S,Liu R,Xu Y,Zhang M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Feb 24 00:00:00 EST 2023
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Anti-PD-L1 antibody enhances curative effect of cryoablation via antibody-dependent cell-mediated cytotoxicity mediating PD-L1highCD11b+ cells elimination in hepatocellular carcinoma.
Acta pharmaceutica Sinica. B
Tan J,Liu T,Fan W,Wei J,Zhu B,Liu Y,Liu L,Zhang X,Chen S,Lin H,Zhang Y,Li J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2023
Oxygen-carrying nanoplatform to reprogram tumor immunosuppressive microenvironment and enhance photothermal-immunotherapy.
Materials today. Bio
Huang J,Leng X,Jiang T,Xu L,Zheng J,Fang M,Wang J,Wang Z,Zhang L
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Non-Oncogene Addiction of KRAS-Mutant Cancers to IL-1β via Versican and Mononuclear IKKβ.
Cancers
Spella M,Ntaliarda G,Skiadas G,Lamort AS,Vreka M,Marazioti A,Lilis I,Bouloukou E,Giotopoulou GA,Pepe MAA,Weiss SAI,Petrera A,Hauck SM,Koch I,Lindner M,Hatz RA,Behr J,Arendt KAM,Giopanou I,Brunn D,Savai R,Jenne DE,de Château M,Yull FE,Blackwell TS,Stathopoulos GT
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Mar 20 00:00:00 EDT 2023
Metabolic improvement and liver regeneration by inhibiting CXXC5 function for non-alcoholic steatohepatitis treatment.
Experimental & molecular medicine
Seo SH,Kim E,Yoon M,Lee SH,Park BH,Choi KY
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2022
Macrophages in epididymal adipose tissue secrete osteopontin to regulate bone homeostasis.
Nature communications
Dai B,Xu J,Li X,Huang L,Hopkins C,Wang H,Yao H,Mi J,Zheng L,Wang J,Tong W,Chow DH,Li Y,He X,Hu P,Chen Z,Zu H,Li Y,Yao Y,Jiang Q,Qin L
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jan 20 00:00:00 EST 2022
PI3Kγ stimulates a high molecular weight form of myosin light chain kinase to promote myeloid cell adhesion and tumor inflammation.
Nature communications
Schmid MC,Kang SW,Chen H,Paradise M,Ghebremedhin A,Kaneda MM,Chin SM,Do A,Watterson DM,Varner JA
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Apr 01 00:00:00 EDT 2022
Neutrophil-dendritic cell interaction plays an important role in live attenuated Leishmania vaccine induced immunity.
PLoS neglected tropical diseases
Bhattacharya P,Ismail N,Saxena A,Gannavaram S,Dey R,Oljuskin T,Akue A,Takeda K,Yu J,Karmakar S,Dagur PK,McCoy JP,Nakhasi HL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Feb 01 00:00:00 EST 2022
Influence of the Postmortem/Storage Time of Human Corneas on the Properties of Cultured Limbal Epithelial Cells.
Cells
Le-Bel G,Desjardins P,Gross C,Cortez Ghio S,Couture C,Germain L,Guérin SL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Western Blot
Wed Aug 31 00:00:00 EDT 2022
TIM3 Expression in Anaplastic-Thyroid-Cancer-Infiltrating Macrophages: An Emerging Immunotherapeutic Target.
Biology
Palacios LM,Peyret V,Viano ME,Geysels RC,Chocobar YA,Volpini X,Pellizas CG,Nicola JP,Motran CC,Rodriguez-Galan MC,Fozzatti L
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Nov 03 00:00:00 EDT 2022
Comparative analysis of thoracic and abdominal aortic aneurysms across the segment and species at the single-cell level.
Frontiers in pharmacology
Wu H,Xie C,Wang R,Cheng J,Xu Q,Zhao H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Feb 02 00:00:00 EST 2023
Microglial angiotensin type 2 receptors mediate sex-specific expression of inflammatory cytokines independently of circulating estrogen.
Glia
Garrido-Gil P,Pedrosa MA,Garcia-Garrote M,Pequeño-Valtierra A,Rodríguez-Castro J,García-Souto D,Rodríguez-Pérez AI,Labandeira-Garcia JL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2022
Regulation of chromatin accessibility by the histone chaperone CAF-1 sustains lineage fidelity.
Nature communications
Franklin R,Guo Y,He S,Chen M,Ji F,Zhou X,Frankhouser D,Do BT,Chiem C,Jang M,Blanco MA,Vander Heiden MG,Rockne RC,Ninova M,Sykes DB,Hochedlinger K,Lu R,Sadreyev RI,Murn J,Volk A,Cheloufi S
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Apr 29 00:00:00 EDT 2022
Biocompatible reduced graphene oxide stimulated BMSCs induce acceleration of bone remodeling and orthodontic tooth movement through promotion on osteoclastogenesis and angiogenesis.
Bioactive materials
Jiao D,Wang J,Yu W,Zhang K,Zhang N,Cao L,Jiang X,Bai Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2022
Epigenetic Studies in the Male APP/BIN1/COPS5 Triple-Transgenic Mouse Model of Alzheimer's Disease.
International journal of molecular sciences
Martínez-Iglesias O,Naidoo V,Carrera I,Cacabelos R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunohistochemistry
Wed Feb 23 00:00:00 EST 2022
Induction of alarmin S100A8/A9 mediates activation of aberrant neutrophils in the pathogenesis of COVID-19.
Cell host & microbe
Guo Q,Zhao Y,Li J,Liu J,Yang X,Guo X,Kuang M,Xia H,Zhang Z,Cao L,Luo Y,Bao L,Wang X,Wei X,Deng W,Wang N,Chen L,Chen J,Zhu H,Gao R,Qin C,Wang X,You F
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Feb 10 00:00:00 EST 2021
Mice with a specific deficiency of Pfkfb3 in myeloid cells are protected from hypoxia-induced pulmonary hypertension.
British journal of pharmacology
Wang L,Zhang X,Cao Y,Ma Q,Mao X,Xu J,Yang Q,Zhou Y,Lucas R,Fulton DJ,Su Y,Barman SA,Hong M,Liu Z,Huo Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Depletion of Adipocyte Becn1 Leads to Lipodystrophy and Metabolic Dysregulation.
Diabetes
Jin Y,Ji Y,Song Y,Choe SS,Jeon YG,Na H,Nam TW,Kim HJ,Nahmgoong H,Kim SM,Kim JW,Nam KT,Seong JK,Hwang D,Park CB,Lee IH,Kim JB,Lee HW
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunohistochemistry
Fri Jan 01 00:00:00 EST 2021
The effects of postmortem delay on mouse and human microglia gene expression.
Glia
Heng Y,Dubbelaar ML,Marie SKN,Boddeke EWGM,Eggen BJL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Intrinsic DNA damage repair deficiency results in progressive microglia loss and replacement.
Glia
Zhang X,Heng Y,Kooistra SM,van Weering HRJ,Brummer ML,Gerrits E,Wesseling EM,Brouwer N,Nijboer TW,Dubbelaar ML,Boddeke EWGM,Eggen BJL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
CD36 deficiency ameliorates drug-induced acute liver injury in mice.
Molecular medicine (Cambridge, Mass.)
Zhang C,Shi X,Su Z,Hu C,Mu X,Pan J,Li M,Teng F,Ling T,Zhao T,Xu C,Ji G,You Q
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Jun 06 00:00:00 EDT 2021
Advanced maternal age induces fetal growth restriction through decreased placental inflammatory cytokine expression and immune cell accumulation in mice.
The Journal of reproduction and development
Hirata Y,Katsukura Y,Henmi Y,Ozawa R,Shimazaki S,Kurosawa A,Torii Y,Takahashi H,Iwata H,Kuwayama T,Shirasuna K
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Aug 27 00:00:00 EDT 2021
Effect of IL-33 on pyroptosis of macrophages in mice with sepsis via NF-κB/p38 MAPK signaling pathway.
Acta cirurgica brasileira
Ke J,Cai G
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunohistochemistry (PFA fixed)
Fri Jun 18 00:00:00 EDT 2021
Brd4 modulates diet-induced obesity via PPARγ-dependent Gdf3 expression in adipose tissue macrophages.
JCI insight
Hu X,Dong X,Li G,Chen Y,Chen J,He X,Sun H,Kim DH,Kemper JK,Chen LF
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Apr 08 00:00:00 EDT 2021
Blockade of IDO-Kynurenine-AhR Axis Ameliorated Colitis-Associated Colon Cancer via Inhibiting Immune Tolerance.
Cellular and molecular gastroenterology and hepatology
Zhang X,Liu X,Zhou W,Du Q,Yang M,Ding Y,Hu R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunocytochemistry
Fri Mar 25 00:00:00 EDT 2022
Polarization of Type 1 Macrophages Is Associated with the Severity of Viral Encephalitis Caused by Japanese Encephalitis Virus and Dengue Virus.
Cells
Jhan MK,Chen CL,Shen TJ,Tseng PC,Wang YT,Satria RD,Yu CY,Lin CF
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Nov 15 00:00:00 EST 2021
Pancreatic Cancer Chemotherapy Is Potentiated by Induction of Tertiary Lymphoid Structures in Mice.
Cellular and molecular gastroenterology and hepatology
Delvecchio FR,Fincham REA,Spear S,Clear A,Roy-Luzarraga M,Balkwill FR,Gribben JG,Bombardieri M,Hodivala-Dilke K,Capasso M,Kocher HM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Mar 28 00:00:00 EDT 2022
Myeloid DJ-1 deficiency protects acetaminophen-induced acute liver injury through decreasing inflammatory response.
Aging
Wang B,Li J,Jiao J,Xu M,Luo Y,Wang F,Xia Q,Gao Y,Feng Y,Kong X,Sun X
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jul 21 00:00:00 EDT 2021
Mutant ASXL1 induces age-related expansion of phenotypic hematopoietic stem cells through activation of Akt/mTOR pathway.
Nature communications
Fujino T,Goyama S,Sugiura Y,Inoue D,Asada S,Yamasaki S,Matsumoto A,Yamaguchi K,Isobe Y,Tsuchiya A,Shikata S,Sato N,Morinaga H,Fukuyama T,Tanaka Y,Fukushima T,Takeda R,Yamamoto K,Honda H,Nishimura EK,Furukawa Y,Shibata T,Abdel-Wahab O,Suematsu M,Kitamura T
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Mar 23 00:00:00 EDT 2021
M2 macrophage microvesicle-inspired nanovehicles improve accessibility to cancer cells and cancer stem cells in tumors.
Journal of nanobiotechnology
Wang Y,Gong X,Li J,Wang H,Xu X,Wu Y,Wang J,Wang S,Li Y,Zhang Z
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Neutralization
Sat Nov 27 00:00:00 EST 2021
In vitro and ex vivo evaluation of tumor-derived exosome-induced dendritic cell dysfunction in mouse.
STAR protocols
Zeng W,Yin X,Jiang Y,Jin L,Liang W
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
Platelet-Like Fusogenic Liposome-Mediated Targeting Delivery of miR-21 Improves Myocardial Remodeling by Reprogramming Macrophages Post Myocardial Ischemia-Reperfusion Injury.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Tan H,Song Y,Chen J,Zhang N,Wang Q,Li Q,Gao J,Yang H,Dong Z,Weng X,Wang Z,Sun D,Yakufu W,Pang Z,Huang Z,Ge J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Synergistic immunotherapy of glioblastoma by dual targeting of IL-6 and CD40.
Nature communications
Yang F,He Z,Duan H,Zhang D,Li J,Yang H,Dorsey JF,Zou W,Nabavizadeh SA,Bagley SJ,Abdullah K,Brem S,Zhang L,Xu X,Byrne KT,Vonderheide RH,Gong Y,Fan Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jun 08 00:00:00 EDT 2021
Mechanical Stress Induce PG-E2 in Murine Synovial Fibroblasts Originating from the Temporomandibular Joint.
Cells
Nazet U,Feulner L,Muschter D,Neubert P,Schatz V,Grässel S,Jantsch J,Proff P,Schröder A,Kirschneck C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Electronic Cigarette Exposure Enhances Lung Inflammatory and Fibrotic Responses in COPD Mice.
Frontiers in pharmacology
Han H,Peng G,Meister M,Yao H,Yang JJ,Zou MH,Liu ZR,Ji X
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Apr 02 00:00:00 EDT 2022
Serp-1 Promotes Corneal Wound Healing by Facilitating Re-epithelialization and Inhibiting Fibrosis and Angiogenesis.
Frontiers in cardiovascular medicine
Ju B,Guo O,Benissan-Messan DZ,Shawver MH,Chen P,Geng B,Wei S,Yaron JR,Lucas AR,Zhu H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunocytochemistry
Fri May 20 00:00:00 EDT 2022
STING Activated Tumor-Intrinsic Type I Interferon Signaling Promotes CXCR3 Dependent Antitumor Immunity in Pancreatic Cancer.
Cellular and molecular gastroenterology and hepatology
Vonderhaar EP,Barnekow NS,McAllister D,McOlash L,Eid MA,Riese MJ,Tarakanova VL,Johnson BD,Dwinell MB
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jan 24 00:00:00 EST 2022
Prevention of non-alcoholic steatohepatitis by long-term exercise via the induction of phenotypic changes in Kupffer cells of hyperphagic obese mice.
Physiological reports
Miura I,Komine S,Okada K,Wada S,Warabi E,Uchida F,Oh S,Suzuki H,Mizokami Y,Shoda J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat May 01 00:00:00 EDT 2021
Genetic fate-mapping reveals surface accumulation but not deep organ invasion of pleural and peritoneal cavity macrophages following injury.
Nature communications
Jin H,Liu K,Tang J,Huang X,Wang H,Zhang Q,Zhu H,Li Y,Pu W,Zhao H,He L,Li Y,Zhang S,Zhang Z,Zhao Y,Qin Y,Pflanz S,Kasmi KEI,Zhang W,Liu Z,Ginhoux F,Ji Y,He B,Wang L,Zhou B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon May 17 00:00:00 EDT 2021
Desiccation Induced Conjunctival Monocyte Recruitment and Activation - Implications for Keratoconjunctivitis.
Frontiers in immunology
Alam J,de Paiva CS,Pflugfelder SC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Dec 10 00:00:00 EST 2021
Inflammation-induced alterations in maternal-fetal Heme Oxygenase (HO) are associated with sustained innate immune cell dysregulation in mouse offspring.
PloS one
Ozen M,Zhao H,Kalish F,Yang Y,Jantzie LL,Wong RJ,Stevenson DK
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Nov 17 00:00:00 EST 2021
A Population of M2 Macrophages Associated With Bone Formation.
Frontiers in immunology
Olmsted-Davis E,Mejia J,Salisbury E,Gugala Z,Davis AR
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Dec 13 00:00:00 EST 2021
Enteric Glia Regulate Lymphocyte Activation via Autophagy-Mediated MHC-II Expression.
Cellular and molecular gastroenterology and hepatology
Chow AK,Grubišić V,Gulbransen BD
46-0112-82 was used in Immunohistochemistry-wholemount to determine whether type II major histocompatibility complex molecule expression in Crohn''s disease and Chagas disease is functional in enteric glial cells.
Fri Mar 25 00:00:00 EDT 2022
Abraxane-induced bone marrow CD11b+ myeloid cell depletion in tumor-bearing mice is visualized by μPET-CT with 64Cu-labeled anti-CD11b and prevented by anti-CSF-1.
Theranostics
Cao Q,Huang Q,Wang YA,Li C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jul 23 00:00:00 EDT 2021
Microglia RAGE exacerbates the progression of neurodegeneration within the SOD1G93A murine model of amyotrophic lateral sclerosis in a sex-dependent manner.
Journal of neuroinflammation
MacLean M,Juranek J,Cuddapah S,López-Díez R,Ruiz HH,Hu J,Frye L,Li H,Gugger PF,Schmidt AM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunohistochemistry (PFA fixed)
Tue Jun 15 00:00:00 EDT 2021
Small-molecule antagonist of VLA-4 (GW559090) attenuated neuro-inflammation by targeting Th17 cell trafficking across the blood-retinal barrier in experimental autoimmune uveitis.
Journal of neuroinflammation
Chen YH,Eskandarpour M,Zhang X,Galatowicz G,Greenwood J,Lightman S,Calder V
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Feb 18 00:00:00 EST 2021
Roles of lung-recruited monocytes and pulmonary Vascular Endothelial Growth Factor (VEGF) in resolving Ventilator-Induced Lung Injury (VILI).
PloS one
Lin CK,Huang TH,Yang CT,Shi CS
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Oct 12 00:00:00 EDT 2021
Systemic administration of β-glucan induces immune training in microglia.
Journal of neuroinflammation
Heng Y,Zhang X,Borggrewe M,van Weering HRJ,Brummer ML,Nijboer TW,Joosten LAB,Netea MG,Boddeke EWGM,Laman JD,Eggen BJL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Feb 22 00:00:00 EST 2021
Hyperactivation of sympathetic nerves drives depletion of melanocyte stem cells.
Nature
Zhang B,Ma S,Rachmin I,He M,Baral P,Choi S,Gonçalves WA,Shwartz Y,Fast EM,Su Y,Zon LI,Regev A,Buenrostro JD,Cunha TM,Chiu IM,Fisher DE,Hsu YC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunohistochemistry
Wed Jan 01 00:00:00 EST 2020
Dual-Wavelength Photosensitive Nano-in-Micro Scaffold Regulates Innate and Adaptive Immune Responses for Osteogenesis.
Nano-micro letters
Zhao Q,Shi M,Yin C,Zhao Z,Zhang J,Wang J,Shen K,Zhang L,Tang H,Xiao Y,Zhang Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Nov 21 00:00:00 EST 2020
IgE Contributes to Atherosclerosis and Obesity by Affecting Macrophage Polarization, Macrophage Protein Network, and Foam Cell Formation.
Arteriosclerosis, thrombosis, and vascular biology
Zhang X,Li J,Luo S,Wang M,Huang Q,Deng Z,de Febbo C,Daoui A,Liew PX,Sukhova GK,Metso J,Jauhiainen M,Shi GP,Guo J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Modulation of the functions of myeloid-derived suppressor cells : a new strategy of hydrogen sulfide anti-cancer effects.
British journal of pharmacology
De Cicco P,Ercolano G,Rubino V,Terrazzano G,Ruggiero G,Cirino G,Ianaro A
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
Alpha-hemolysin of uropathogenic Escherichia coli induces GM-CSF-mediated acute kidney injury.
Mucosal immunology
Wang C,Li Q,Lv J,Sun X,Cao Y,Yu K,Miao C,Zhang ZS,Yao Z,Wang Q
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
Temporospatial distribution and transcriptional profile of retinal microglia in the oxygen-induced retinopathy mouse model.
Glia
Boeck M,Thien A,Wolf J,Hagemeyer N,Laich Y,Yusuf D,Backofen R,Zhang P,Boneva S,Stahl A,Hilgendorf I,Agostini H,Prinz M,Wieghofer P,Schlunck G,Schlecht A,Lange C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Characterization and regulation of osteoclast precursors following chronic Porphyromonas gingivalis infection.
Journal of leukocyte biology
Zhao Y,Li Z,Su L,Ballesteros-Tato A,Katz J,Michalek SM,Feng X,Zhang P
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Circulating Neutrophils of Nonalcoholic Steatohepatitis Patients Show an Activated Phenotype and Suppress T Lymphocytes Activity.
Journal of immunology research
Antonucci L,Porcu C,Timperi E,Santini SJ,Iannucci G,Balsano C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri May 21 00:00:00 EDT 2021
Mouse bone marrow mesenchymal stem cells with distinct p53 statuses display differential characteristics.
Molecular medicine reports
Wang B,Wang L,Mao J,Wen H,Xu L,Ren Y,Du H,Yang H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
Senolytic Agent Navitoclax Inhibits Angiotensin II-Induced Heart Failure in Mice.
Journal of cardiovascular pharmacology
Jia K,Dai Y,Liu A,Li X,Wu L,Lu L,Bao Y,Jin Q
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Immunohistochemistry
Thu Oct 01 00:00:00 EDT 2020
Effects of Hypertonic Saline and Hydroxyethyl Starch on Myeloid-Derived Suppressor Cells in Hemorrhagic Shock Mice under Secondary Bacterial Attack.
BioMed research international
Jiang JK,Hong LJ,Lu YQ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Dec 16 00:00:00 EST 2020
Irradiation induces cancer lung metastasis through activation of the cGAS-STING-CCL5 pathway in mesenchymal stromal cells.
Cell death & disease
Zheng Z,Jia S,Shao C,Shi Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu May 07 00:00:00 EDT 2020
Therapeutic Role of Inducible Nitric Oxide Synthase Expressing Myeloid-Derived Suppressor Cells in Acetaminophen-Induced Murine Liver Failure.
Frontiers in immunology
Hsu CY,Lin YC,Chang LY,Huang SK,Huang CH,Yang CK,Huang CT,Lin CY
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jun 15 00:00:00 EDT 2021
Long Non-Coding RNA Hotairm1 Promotes S100A9 Support of MDSC Expansion during Sepsis.
Journal of clinical & cellular immunology
Alkhateeb T,Bah I,Kumbhare A,Youssef D,Yao ZQ,McCall CE,Gazzar ME
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Aug 13 00:00:00 EDT 2022
GDF3 Protects Mice against Sepsis-Induced Cardiac Dysfunction and Mortality by Suppression of Macrophage Pro-Inflammatory Phenotype.
Cells
Wang L,Li Y,Wang X,Wang P,Essandoh K,Cui S,Huang W,Mu X,Liu Z,Wang Y,Peng T,Fan GC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jan 03 00:00:00 EST 2020
Differential Activation of Macrophages Based on Their Environment in Advanced Age.
Chonnam medical journal
Lee DY,Lim JS,Cho KA
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Exploring the Role of Osteosarcoma-Derived Extracellular Vesicles in Pre-Metastatic Niche Formation and Metastasis in the 143-B Xenograft Mouse Osteosarcoma Model.
Cancers
Mazumdar A,Urdinez J,Boro A,Arlt MJE,Egli FE,Niederöst B,Jaeger PK,Moschini G,Muff R,Fuchs B,Snedeker JG,Gvozdenovic A
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Nov 20 00:00:00 EST 2020
Lipin-1 Contributes to IL-4 Mediated Macrophage Polarization.
Frontiers in immunology
Chandran S,Schilke RM,Blackburn CMR,Yurochko A,Mirza R,Scott RS,Finck BN,Woolard MD
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Mar 29 00:00:00 EDT 2021
Ly6cLo non-classical monocytes promote resolution of rhesus rotavirus-mediated perinatal hepatic inflammation.
Scientific reports
Alkhani A,Levy CS,Tsui M,Rosenberg KA,Polovina K,Mattis AN,Mack M,Van Dyken S,Wang BM,Maher JJ,Nijagal A
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Apr 28 00:00:00 EDT 2020
Stromal CCL2 Signaling Promotes Mammary Tumor Fibrosis through Recruitment of Myeloid-Lineage Cells.
Cancers
Kuziel G,Thompson V,D'Amato JV,Arendt LM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jul 28 00:00:00 EDT 2020
The immune response after noise damage in the cochlea is characterized by a heterogeneous mix of adaptive and innate immune cells.
Scientific reports
Rai V,Wood MB,Feng H,Schabla NM,Tu S,Zuo J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Sep 16 00:00:00 EDT 2020
Macrophage-Associated Lipin-1 Promotes β-Oxidation in Response to Proresolving Stimuli.
ImmunoHorizons
Schilke RM,Blackburn CMR,Rao S,Krzywanski DM,Finck BN,Woolard MD
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Oct 19 00:00:00 EDT 2020
TNFAIP3 Plays a Role in Aging of the Hematopoietic System.
Frontiers in immunology
Smith MA,Culver-Cochran AE,Adelman ER,Rhyasen GW,Ma A,Figueroa ME,Starczynowski DT
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Apr 28 00:00:00 EDT 2021
Analysis of the immune response to sciatic nerve injury identifies efferocytosis as a key mechanism of nerve debridement.
eLife
Kalinski AL,Yoon C,Huffman LD,Duncker PC,Kohen R,Passino R,Hafner H,Johnson C,Kawaguchi R,Carbajal KS,Jara JS,Hollis E,Geschwind DH,Segal BM,Giger RJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Dec 02 00:00:00 EST 2020
CD103+ cDC1 and endogenous CD8+ T cells are necessary for improved CD40L-overexpressing CAR T cell antitumor function.
Nature communications
Kuhn NF,Lopez AV,Li X,Cai W,Daniyan AF,Brentjens RJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Dec 02 00:00:00 EST 2020
Authentication of Primary Murine Cell Lines by a Microfluidics-Based Lab-On-Chip System.
Biomedicines
Hong Y,Singh N,Bamopoulos S,Gjerga E,Schmalbrock LK,Balabanian K,Schick M,Keller U,Wirth M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Dec 09 00:00:00 EST 2020
The armed oncolytic adenovirus ZD55-IL-24 eradicates melanoma by turning the tumor cells from the self-state into the nonself-state besides direct killing.
Cell death & disease
Hu HJ,Liang X,Li HL,Du CM,Hao JL,Wang HY,Gu JF,Ni AM,Sun LY,Xiao J,Hu JQ,Yuan H,Dai YS,Jin XT,Zhang KJ,Liu XY
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Nov 30 00:00:00 EST 2020
L-Selectin/CD62L is a Key Driver of Non-Alcoholic Steatohepatitis in Mice and Men.
Cells
Drescher HK,Schippers A,Rosenhain S,Gremse F,Bongiovanni L,Bruin A,Eswaran S,Gallage SU,Pfister D,Szydlowska M,Heikenwalder M,Weiskirchen S,Wagner N,Trautwein C,Weiskirchen R,Kroy DC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Apr 29 00:00:00 EDT 2020
Ovarian granulosa cell tumor characterization identifies FOXL2 as an immunotherapeutic target.
JCI insight
Pierini S,Tanyi JL,Simpkins F,George E,Uribe-Herranz M,Drapkin R,Burger R,Morgan MA,Facciabene A
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Aug 20 00:00:00 EDT 2020
Protein Tyrosine Phosphatase Non-Receptor Type 2 Function in Dendritic Cells Is Crucial to Maintain Tissue Tolerance.
Frontiers in immunology
Hering L,Katkeviciute E,Schwarzfischer M,Busenhart P,Gottier C,Mrdjen D,Komuczki J,Wawrzyniak M,Lang S,Atrott K,Becher B,Rogler G,Scharl M,Spalinger MR
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Apr 12 00:00:00 EDT 2021
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Aug 27 00:00:00 EDT 2020
Hyaluronan control of the primary vascular barrier during early mouse pregnancy is mediated by uterine NK cells.
JCI insight
Hadas R,Gershon E,Cohen A,Atrakchi O,Lazar S,Golani O,Dassa B,Elbaz M,Cohen G,Eilam R,Dekel N,Neeman M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Nov 19 00:00:00 EST 2020
Loss of Atg7 causes chaotic nucleosome assembly of mouse bone marrow CD11b+Ly6G- myeloid cells.
Aging
Fang Y,Gu Y,Li L,Zhu L,Qian J,Zhao C,Xu L,Wei W,Du Y,Yuan N,Zhang S,Yuan Y,Xu Y,Jiang C,Wang J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Nov 24 00:00:00 EST 2020
Bivalent Inhibitor with Selectivity for Trimeric MMP-9 Amplifies Neutrophil Chemotaxis and Enables Functional Studies on MMP-9 Proteoforms.
Cells
Nuti E,Rossello A,Cuffaro D,Camodeca C,Van Bael J,van der Maat D,Martens E,Fiten P,Pereira RVS,Ugarte-Berzal E,Gouwy M,Opdenakker G,Vandooren J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jul 07 00:00:00 EDT 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Nov 27 00:00:00 EST 2020
Loss of FGFR3 Delays Acute Myeloid Leukemogenesis by Programming Weakly Pathogenic CD117-Positive Leukemia Stem-Like Cells.
Frontiers in pharmacology
Guo C,Ran Q,Sun C,Zhou T,Yang X,Zhang J,Pang S,Xiao Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Feb 16 00:00:00 EST 2021
Chemokine Receptor Redundancy and Specificity Are Context Dependent.
Immunity
Dyer DP,Medina-Ruiz L,Bartolini R,Schuette F,Hughes CE,Pallas K,Vidler F,Macleod MKL,Kelly CJ,Lee KM,Hansell CAH,Graham GJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Feb 19 00:00:00 EST 2019
Basophil-derived tumor necrosis factor can enhance survival in a sepsis model in mice.
Nature immunology
Piliponsky AM,Shubin NJ,Lahiri AK,Truong P,Clauson M,Niino K,Tsuha AL,Nedospasov SA,Karasuyama H,Reber LL,Tsai M,Mukai K,Galli SJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2019
Central nervous system regeneration is driven by microglia necroptosis and repopulation.
Nature neuroscience
Lloyd AF,Davies CL,Holloway RK,Labrak Y,Ireland G,Carradori D,Dillenburg A,Borger E,Soong D,Richardson JC,Kuhlmann T,Williams A,Pollard JW,des Rieux A,Priller J,Miron VE
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
NLRX1 inhibits the early stages of CNS inflammation and prevents the onset of spontaneous autoimmunity.
PLoS biology
Gharagozloo M,Mahmoud S,Simard C,Yamamoto K,Bobbala D,Ilangumaran S,Smith MD,Lamontagne A,Jarjoura S,Denault JB,Blais V,Gendron L,Vilariño-Güell C,Sadovnick AD,Ting JP,Calabresi PA,Amrani A,Gris D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2019
Seselin ameliorates inflammation via targeting Jak2 to suppress the proinflammatory phenotype of macrophages.
British journal of pharmacology
Feng L,Sun Y,Song P,Xu L,Wu X,Wu X,Shen Y,Sun Y,Kong L,Wu X,Xu Q
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2019
Innate immune memory through TLR2 and NOD2 contributes to the control of Leptospira interrogans infection.
PLoS pathogens
Santecchia I,Vernel-Pauillac F,Rasid O,Quintin J,Gomes-Solecki M,Boneca IG,Werts C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2019
Distinct Tie2 tyrosine phosphorylation sites dictate phenotypic switching in endothelial progenitor cells.
Journal of cellular physiology
Siavashi V,Nassiri SM,Rahbarghazi R,Mohseni Z,Sharifi AM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2019
Extracellular RNAs from lung cancer cells activate epithelial cells and induce neutrophil extracellular traps.
International journal of oncology
Li Y,Yang Y,Gan T,Zhou J,Hu F,Hao N,Yuan B,Chen Y,Zhang M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
Antiallodynic Effects of Cannabinoid Receptor 2 (CB2R) Agonists on Retrovirus Infection-Induced Neuropathic Pain.
Pain research & management
Sheng WS,Chauhan P,Hu S,Prasad S,Lokensgard JR
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Dec 16 00:00:00 EST 2019
Antitumor and immunomodulatory effects of a novel multitarget inhibitor, CS2164, in mouse hepatocellular carcinoma models.
Anti-cancer drugs
Zhou Y,Fu C,Kong Y,Pan D,Wang Y,Huang S,Li Z,Ning Z,Lu X,Shan S,Xin L
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
Absence of TGFβ signaling in retinal microglia induces retinal degeneration and exacerbates choroidal neovascularization.
eLife
Ma W,Silverman SM,Zhao L,Villasmil R,Campos MM,Amaral J,Wong WT
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jan 22 00:00:00 EST 2019
TRIM9-Mediated Resolution of Neuroinflammation Confers Neuroprotection upon Ischemic Stroke in Mice.
Cell reports
Zeng J,Wang Y,Luo Z,Chang LC,Yoo JS,Yan H,Choi Y,Xie X,Deverman BE,Gradinaru V,Gupton SL,Zlokovic BV,Zhao Z,Jung JU
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Apr 09 00:00:00 EDT 2019
Ubiquitination of RIPK1 suppresses programmed cell death by regulating RIPK1 kinase activation during embryogenesis.
Nature communications
Zhang X,Zhang H,Xu C,Li X,Li M,Wu X,Pu W,Zhou B,Wang H,Li D,Ding Q,Ying H,Wang H,Zhang H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Sep 13 00:00:00 EDT 2019
Dependence on Myb expression is attenuated in myeloid leukaemia with N-terminal CEBPA mutations.
Life science alliance
Volpe G,Cauchy P,Walton DS,Ward C,Blakemore D,Bayley R,Clarke ML,Schmidt L,Nerlov C,Garcia P,Dumon S,Grebien F,Frampton J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2019
Soluble TREM2 ameliorates pathological phenotypes by modulating microglial functions in an Alzheimer's disease model.
Nature communications
Zhong L,Xu Y,Zhuo R,Wang T,Wang K,Huang R,Wang D,Gao Y,Zhu Y,Sheng X,Chen K,Wang N,Zhu L,Can D,Marten Y,Shinohara M,Liu CC,Du D,Sun H,Wen L,Xu H,Bu G,Chen XF
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Mar 25 00:00:00 EDT 2019
Neural anti-inflammatory action mediated by two types of acetylcholine receptors in the small intestine.
Scientific reports
Kimura H,Imura YK,Tomiyasu H,Mihara T,Kaji N,Ohno K,Unno T,Tanahashi Y,Jan TR,Tsubone H,Ozaki H,Hori M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Apr 10 00:00:00 EDT 2019
MDM2 inhibitor APG-115 synergizes with PD-1 blockade through enhancing antitumor immunity in the tumor microenvironment.
Journal for immunotherapy of cancer
Fang DD,Tang Q,Kong Y,Wang Q,Gu J,Fang X,Zou P,Rong T,Wang J,Yang D,Zhai Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Nov 28 00:00:00 EST 2019
Muscularis Propria Macrophages Alter the Proportion of Nitrergic but Not Cholinergic Gastric Myenteric Neurons.
Cellular and molecular gastroenterology and hepatology
Cipriani G,Terhaar ML,Eisenman ST,Ji S,Linden DR,Wright AM,Sha L,Ordog T,Szurszewski JH,Gibbons SJ,Farrugia G
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jun 04 00:00:00 EDT 2019
Tpl2 Protects Against Fulminant Hepatitis Through Mobilization of Myeloid-Derived Suppressor Cells.
Frontiers in immunology
Xu J,Pei S,Wang Y,Liu J,Qian Y,Huang M,Zhang Y,Xiao Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Oct 14 00:00:00 EDT 2020
Exposure to Cigarette Smoke Augments Post-ischemic Brain Injury and Inflammation via Mobilization of Neutrophils and Monocytes.
Frontiers in immunology
Li H,Li X,Gao S,Wang D,Gao X,Li Y,Wang X,Cui Z,Ma H,Liu Q,Li M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Knockdown of Ant2 Reduces Adipocyte Hypoxia And Improves Insulin Resistance in Obesity.
Nature metabolism
Seo JB,Riopel M,Cabrales P,Huh JY,Bandyopadhyay GK,Andreyev AY,Murphy AN,Beeman SC,Smith GI,Klein S,Lee YS,Olefsky JM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2019
Inducing cancer indolence by targeting mitochondrial Complex I is potentiated by blocking macrophage-mediated adaptive responses.
Nature communications
Kurelac I,Iommarini L,Vatrinet R,Amato LB,De Luise M,Leone G,Girolimetti G,Umesh Ganesh N,Bridgeman VL,Ombrato L,Columbaro M,Ragazzi M,Gibellini L,Sollazzo M,Feichtinger RG,Vidali S,Baldassarre M,Foriel S,Vidone M,Cossarizza A,Grifoni D,Kofler B,Malanchi I,Porcelli AM,Gasparre G
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Feb 22 00:00:00 EST 2019
The nuclear variant of bone morphogenetic protein 2 (nBMP2) is expressed in macrophages and alters calcium response.
Scientific reports
Tellez Freitas CM,Burrell HR,Valdoz JC,Hamblin GJ,Raymond CM,Cox TD,Johnson DK,Andersen JL,Weber KS,Bridgewater LC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jan 30 00:00:00 EST 2019
Costimulation of type-2 innate lymphoid cells by GITR promotes effector function and ameliorates type 2 diabetes.
Nature communications
Galle-Treger L,Sankaranarayanan I,Hurrell BP,Howard E,Lo R,Maazi H,Lewis G,Banie H,Epstein AL,Hu P,Rehan VK,Gilliland FD,Allayee H,Soroosh P,Sharpe AH,Akbari O
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Feb 12 00:00:00 EST 2019
Toll-Like Receptor 7 Agonist-Induced Dermatitis Causes Severe Dextran Sulfate Sodium Colitis by Altering the Gut Microbiome and Immune Cells.
Cellular and molecular gastroenterology and hepatology
Kiyohara H,Sujino T,Teratani T,Miyamoto K,Arai MM,Nomura E,Harada Y,Aoki R,Koda Y,Mikami Y,Mizuno S,Naganuma M,Hisamatsu T,Kanai T
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Apr 29 00:00:00 EDT 2019
Inflammation induced by incomplete radiofrequency ablation accelerates tumor progression and hinders PD-1 immunotherapy.
Nature communications
Shi L,Wang J,Ding N,Zhang Y,Zhu Y,Dong S,Wang X,Peng C,Zhou C,Zhou L,Li X,Shi H,Wu W,Long X,Wu C,Liao W
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Nov 28 00:00:00 EST 2019
Acute tobacco smoke exposure exacerbates the inflammatory response to corneal wounds in mice via the sympathetic nervous system.
Communications biology
Xiao C,Wu M,Liu J,Gu J,Jiao X,Lu D,He J,Lin C,Xue Y,Fu T,Wang H,Wang G,Yang X,Li Z
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Apr 02 00:00:00 EDT 2020
Efferocytosis induces a novel SLC program to promote glucose uptake and lactate release.
Nature
Morioka S,Perry JSA,Raymond MH,Medina CB,Zhu Y,Zhao L,Serbulea V,Onengut-Gumuscu S,Leitinger N,Kucenas S,Rathmell JC,Makowski L,Ravichandran KS
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2018
Microbial signals drive pre-leukaemic myeloproliferation in a Tet2-deficient host.
Nature
Meisel M,Hinterleitner R,Pacis A,Chen L,Earley ZM,Mayassi T,Pierre JF,Ernest JD,Galipeau HJ,Thuille N,Bouziat R,Buscarlet M,Ringus DL,Wang Y,Li Y,Dinh V,Kim SM,McDonald BD,Zurenski MA,Musch MW,Furtado GC,Lira SA,Baier G,Chang EB,Eren AM,Weber CR,Busque L,Godley LA,Verdú EF,Barreiro LB,Jabri B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2018
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jan 25 00:00:00 EST 2018
Mitophagy in Intestinal Epithelial Cells Triggers Adaptive Immunity during Tumorigenesis.
Cell
Ziegler PK,Bollrath J,Pallangyo CK,Matsutani T,Canli Ö,De Oliveira T,Diamanti MA,Müller N,Gamrekelashvili J,Putoczki T,Horst D,Mankan AK,Öner MG,Müller S,Müller-Höcker J,Kirchner T,Slotta-Huspenina J,Taketo MM,Reinheckel T,Dröse S,Larner AC,Wels WS,Ernst M,Greten TF,Arkan MC,Korn T,Wirth D,Greten FR
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jun 28 00:00:00 EDT 2018
Chanzyme TRPM7 Mediates the Ca2+ Influx Essential for Lipopolysaccharide-Induced Toll-Like Receptor 4 Endocytosis and Macrophage Activation.
Immunity
Schappe MS,Szteyn K,Stremska ME,Mendu SK,Downs TK,Seegren PV,Mahoney MA,Dixit S,Krupa JK,Stipes EJ,Rogers JS,Adamson SE,Leitinger N,Desai BN
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jan 16 00:00:00 EST 2018
E3 Ligase VHL Promotes Group 2 Innate Lymphoid Cell Maturation and Function via Glycolysis Inhibition and Induction of Interleukin-33 Receptor.
Immunity
Li Q,Li D,Zhang X,Wan Q,Zhang W,Zheng M,Zou L,Elly C,Lee JH,Liu YC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Feb 20 00:00:00 EST 2018
Autoimmune Th17 Cells Induced Synovial Stromal and Innate Lymphoid Cell Secretion of the Cytokine GM-CSF to Initiate and Augment Autoimmune Arthritis.
Immunity
Hirota K,Hashimoto M,Ito Y,Matsuura M,Ito H,Tanaka M,Watanabe H,Kondoh G,Tanaka A,Yasuda K,Kopf M,Potocnik AJ,Stockinger B,Sakaguchi N,Sakaguchi S
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jun 19 00:00:00 EDT 2018
Insulin resistance causes inflammation in adipose tissue.
The Journal of clinical investigation
Shimobayashi M,Albert V,Woelnerhanssen B,Frei IC,Weissenberger D,Meyer-Gerspach AC,Clement N,Moes S,Colombi M,Meier JA,Swierczynska MM,Jenö P,Beglinger C,Peterli R,Hall MN
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Apr 02 00:00:00 EDT 2018
Granulocyte-CSF links destructive inflammation and comorbidities in obstructive lung disease.
The Journal of clinical investigation
Tsantikos E,Lau M,Castelino CM,Maxwell MJ,Passey SL,Hansen MJ,McGregor NE,Sims NA,Steinfort DP,Irving LB,Anderson GP,Hibbs ML
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Tamm-Horsfall Protein Regulates Mononuclear Phagocytes in the Kidney.
Journal of the American Society of Nephrology : JASN
Micanovic R,Khan S,Janosevic D,Lee ME,Hato T,Srour EF,Winfree S,Ghosh J,Tong Y,Rice SE,Dagher PC,Wu XR,El-Achkar TM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2018
11 Beta-hydroxysteroid dehydrogenase type 1 regulates synovitis, joint destruction, and systemic bone loss in chronic polyarthritis.
Journal of autoimmunity
Hardy RS,Fenton C,Croft AP,Naylor AJ,Begum R,Desanti G,Buckley CD,Lavery G,Cooper MS,Raza K
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
MK2 contributes to tumor progression by promoting M2 macrophage polarization and tumor angiogenesis.
Proceedings of the National Academy of Sciences of the United States of America
Suarez-Lopez L,Sriram G,Kong YW,Morandell S,Merrick KA,Hernandez Y,Haigis KM,Yaffe MB
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2018
Myeloid ERK5 deficiency suppresses tumor growth by blocking protumor macrophage polarization via STAT3 inhibition.
Proceedings of the National Academy of Sciences of the United States of America
Giurisato E,Xu Q,Lonardi S,Telfer B,Russo I,Pearson A,Finegan KG,Wang W,Wang J,Gray NS,Vermi W,Xia Z,Tournier C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Mar 20 00:00:00 EDT 2018
VISTA expression by microglia decreases during inflammation and is differentially regulated in CNS diseases.
Glia
Borggrewe M,Grit C,Den Dunnen WFA,Burm SM,Bajramovic JJ,Noelle RJ,Eggen BJL,Laman JD
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Bone loss in C57BL/6J-OlaHsd mice, a substrain of C57BL/6J carrying mutated alpha-synuclein and multimerin-1 genes.
Journal of cellular physiology
Liron T,Raphael B,Hiram-Bab S,Bab IA,Gabet Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Inactivation of mTORC1 Signaling in Osterix-Expressing Cells Impairs B-cell Differentiation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang Y,Xiao M,Tao C,Chen J,Wang Z,Yang J,Chen Z,Zou Z,Liu A,Cai D,Jiang Y,Ding C,Li M,Bai X
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Frontline Science: Macrophage-derived exosomes promote neutrophil necroptosis following hemorrhagic shock.
Journal of leukocyte biology
Jiao Y,Li Z,Loughran PA,Fan EK,Scott MJ,Li Y,Billiar TR,Wilson MA,Shi X,Fan J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Feb 01 00:00:00 EST 2018
Dual specificity phosphatase DUSP6 promotes endothelial inflammation through inducible expression of ICAM-1.
The FEBS journal
Hsu SF,Lee YB,Lee YC,Chung AL,Apaya MK,Shyur LF,Cheng CF,Ho FM,Meng TC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2018
The Outcome of the Cryptococcus neoformans-Macrophage Interaction Depends on Phagolysosomal Membrane Integrity.
Journal of immunology (Baltimore, Md. : 1950)
De Leon-Rodriguez CM,Rossi DCP,Fu MS,Dragotakes Q,Coelho C,Guerrero Ros I,Caballero B,Nolan SJ,Casadevall A
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Jul 15 00:00:00 EDT 2018
Primary cilia disruption differentially affects the infiltrating and resident macrophage compartment in the liver.
American journal of physiology. Gastrointestinal and liver physiology
Zimmerman KA,Song CJ,Gonzalez-Mize N,Li Z,Yoder BK
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Anti‑IL‑39 (IL‑23p19/Ebi3) polyclonal antibodies ameliorate autoimmune symptoms in lupus‑like mice.
Molecular medicine reports
Wang X,Zhang Y,Wang Z,Liu X,Zhu G,Han G,Chen G,Hou C,Wang T,Shen B,Li Y,Xiao H,Ma N,Wang R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
A novel method to isolate mesenchymal stem cells from mouse umbilical cord.
Molecular medicine reports
Zhang B,Zhang J,Shi H,Mao F,Wang J,Yan Y,Zhang X,Qian H,Xu W
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Simultaneous isolation of mesenchymal stem cells and endothelial progenitor cells derived from murine bone marrow.
Experimental and therapeutic medicine
Wang X,Zhao Z,Zhang H,Hou J,Feng W,Zhang M,Guo J,Xia J,Ge Q,Chen X,Wu X
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
iRhom2-mediated proinflammatory signalling regulates heart repair following myocardial infarction.
JCI insight
Barnette DN,Cahill TJ,Gunadasa-Rohling M,Carr CA,Freeman M,Riley PR
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Feb 08 00:00:00 EST 2018
A cancer vaccine-mediated postoperative immunotherapy for recurrent and metastatic tumors.
Nature communications
Wang T,Wang D,Yu H,Feng B,Zhou F,Zhang H,Zhou L,Jiao S,Li Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Apr 18 00:00:00 EDT 2018
Diet-Induced Obesity Elicits Macrophage Infiltration and Reduction in Spine Density in the Hypothalami of Male but Not Female Mice.
Frontiers in immunology
Lainez NM,Jonak CR,Nair MG,Ethell IM,Wilson EH,Carson MJ,Coss D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Sep 25 00:00:00 EDT 2019
Microglia prevent peripheral immune cell invasion and promote an anti-inflammatory environment in the brain of APP-PS1 transgenic mice.
Journal of neuroinflammation
Unger MS,Schernthaner P,Marschallinger J,Mrowetz H,Aigner L
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Sep 21 00:00:00 EDT 2018
Investigation of tumor-tumor interactions in a double human cervical carcinoma xenograft model in nude mice.
Oncotarget
Mertens B,Cristina de Araujo Nogueira T,Topalis D,Stranska R,Snoeck R,Andrei G
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Apr 24 00:00:00 EDT 2018
BNIP3 modulates the interface between B16-F10 melanoma cells and immune cells.
Oncotarget
Romano E,Rufo N,Korf H,Mathieu C,Garg AD,Agostinis P
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Apr 03 00:00:00 EDT 2018
An immune cell spray (ICS) formulation allows for the delivery of functional monocyte/macrophages.
Scientific reports
Beneke V,Küster F,Neehus AL,Hesse C,Lopez-Rodriguez E,Haake K,Rafiei Hashtchin A,Schott JW,Walter D,Braun A,Wolkers WF,Ackermann M,Lachmann N
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Nov 02 00:00:00 EDT 2018
Macrophage Depletion Protects against Cigarette Smoke-Induced Inflammatory Response in the Mouse Colon and Lung.
Frontiers in physiology
Lim D,Kim W,Lee C,Bae H,Kim J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
RhoA, Rac1, and Cdc42 differentially regulate αSMA and collagen I expression in mesenchymal stem cells.
The Journal of biological chemistry
Ge J,Burnier L,Adamopoulou M,Kwa MQ,Schaks M,Rottner K,Brakebusch C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jun 15 00:00:00 EDT 2018
JNK1 negatively controls antifungal innate immunity by suppressing CD23 expression.
Nature medicine
Zhao X,Guo Y,Jiang C,Chang Q,Zhang S,Luo T,Zhang B,Jia X,Hung MC,Dong C,Lin X
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Myeloid progenitor cluster formation drives emergency and leukaemic myelopoiesis.
Nature
Hérault A,Binnewies M,Leong S,Calero-Nieto FJ,Zhang SY,Kang YA,Wang X,Pietras EM,Chu SH,Barry-Holson K,Armstrong S,Göttgens B,Passegué E
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Apr 06 00:00:00 EDT 2017
A Wnt-producing niche drives proliferative potential and progression in lung adenocarcinoma.
Nature
Tammela T,Sanchez-Rivera FJ,Cetinbas NM,Wu K,Joshi NS,Helenius K,Park Y,Azimi R,Kerper NR,Wesselhoeft RA,Gu X,Schmidt L,Cornwall-Brady M,Yilmaz ÖH,Xue W,Katajisto P,Bhutkar A,Jacks T
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu May 18 00:00:00 EDT 2017
Ubiquitination of hnRNPA1 by TRAF6 links chronic innate immune signaling with myelodysplasia.
Nature immunology
Fang J,Bolanos LC,Choi K,Liu X,Christie S,Akunuru S,Kumar R,Wang D,Chen X,Greis KD,Stoilov P,Filippi MD,Maciejewski JP,Garcia-Manero G,Weirauch MT,Salomonis N,Geiger H,Zheng Y,Starczynowski DT
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
A CCR4 antagonist reverses the tumor-promoting microenvironment of renal cancer.
The Journal of clinical investigation
Berlato C,Khan MN,Schioppa T,Thompson R,Maniati E,Montfort A,Jangani M,Canosa M,Kulbe H,Hagemann UB,Duncan AR,Fletcher L,Wilkinson RW,Powles T,Quezada SA,Balkwill FR
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Critical role for Sec22b-dependent antigen cross-presentation in antitumor immunity.
The Journal of experimental medicine
Alloatti A,Rookhuizen DC,Joannas L,Carpier JM,Iborra S,Magalhaes JG,Yatim N,Kozik P,Sancho D,Albert ML,Amigorena S
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Aug 07 00:00:00 EDT 2017
Deletion of PIKfyve alters alveolar macrophage populations and exacerbates allergic inflammation in mice.
The EMBO journal
Kawasaki T,Ito K,Miyata H,Akira S,Kawai T
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jun 14 00:00:00 EDT 2017
Brd4 modulates the innate immune response through Mnk2-eIF4E pathway-dependent translational control of IκBα.
Proceedings of the National Academy of Sciences of the United States of America
Bao Y,Wu X,Chen J,Hu X,Zeng F,Cheng J,Jin H,Lin X,Chen LF
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue May 16 00:00:00 EDT 2017
Prolonged Deleterious Influences of Chemotherapeutic Agent CPT-11 on Resident Peritoneal Macrophages and B1 Cells.
Frontiers in immunology
Bai WJ,Li CG,Zhang CC,Xu LH,Zeng QZ,Hu B,Hong Z,He XH,Ouyang DY
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
p40 -Deficient Mice Exhibit Impaired Bacterial Clearance and Enhanced Pro-inflammatory Responses during Salmonella enterica serovar Typhimurium Infection.
Frontiers in immunology
Li Y,Lv M,Su C,Long S,Zhang W,Conway KL,Li W,Xavier RJ,Shi HN
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
CCR2- and CCR2+ corneal macrophages exhibit distinct characteristics and balance inflammatory responses after epithelial abrasion.
Mucosal immunology
Liu J,Xue Y,Dong D,Xiao C,Lin C,Wang H,Song F,Fu T,Wang Z,Chen J,Pan H,Li Y,Cai D,Li Z
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2017
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Lymphatic Endothelial Cells Produce M-CSF, Causing Massive Bone Loss in Mice.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang W,Wang H,Zhou X,Li X,Sun W,Dellinger M,Boyce BF,Xing L
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
Distribution and differentiation of myeloid-derived suppressor cells after fluid resuscitation in mice with hemorrhagic shock.
Journal of Zhejiang University. Science. B
Jiang JK,Fang W,Hong LJ,Lu YQ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jul 11 00:00:00 EDT 2017
Lung Epithelial Cell-Derived Microvesicles Regulate Macrophage Migration via MicroRNA-17/221-Induced Integrin β1 Recycling.
Journal of immunology (Baltimore, Md. : 1950)
Lee H,Zhang D,Wu J,Otterbein LE,Jin Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Aug 15 00:00:00 EDT 2017
Silybin attenuates LPS-induced lung injury in mice by inhibiting NF-κB signaling and NLRP3 activation.
International journal of molecular medicine
Zhang B,Wang B,Cao S,Wang Y,Wu D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
ADAR1 deletion induces NFκB and interferon signaling dependent liver inflammation and fibrosis.
RNA biology
Ben-Shoshan SO,Kagan P,Sultan M,Barabash Z,Dor C,Jacob-Hirsch J,Harmelin A,Pappo O,Marcu-Malina V,Ben-Ari Z,Amariglio N,Rechavi G,Goldstein I,Safran M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu May 04 00:00:00 EDT 2017
β3-Adrenergic Regulation of EPC Features Through Manipulation of the Bone Marrow MSC Niche.
Journal of cellular biochemistry
Vafaei R,Nassiri SM,Siavashi V
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
Coinjection of IL2 DNA enhances E7-specific antitumor immunity elicited by intravaginal therapeutic HPV DNA vaccination with electroporation.
Gene therapy
Sun Y,Peng S,Yang A,Farmer E,Wu TC,Hung CF
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2017
Growth inhibition of formed corneal neovascularization following Fosaprepitant treatment.
Acta ophthalmologica
Bignami F,Lorusso A,Rama P,Ferrari G
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Nov 01 00:00:00 EDT 2017
Ethanol, TLR3, and TLR4 Agonists Have Unique Innate Immune Responses in Neuron-Like SH-SY5Y and Microglia-Like BV2.
Alcoholism, clinical and experimental research
Lawrimore CJ,Crews FT
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
Myeloid-specific genetic ablation of ATP-binding cassette transporter ABCA1 is protective against cancer.
Oncotarget
Zamanian-Daryoush M,Lindner DJ,DiDonato JA,Wagner M,Buffa J,Rayman P,Parks JS,Westerterp M,Tall AR,Hazen SL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Sep 22 00:00:00 EDT 2017
Armored CAR T cells enhance antitumor efficacy and overcome the tumor microenvironment.
Scientific reports
Yeku OO,Purdon TJ,Koneru M,Spriggs D,Brentjens RJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Sep 05 00:00:00 EDT 2017
MiR-125a Is a critical modulator for neutrophil development.
PLoS genetics
Qin Y,Wu L,Ouyang Y,Zhou P,Zhou H,Wang Y,Ma J,Zhang J,Chen Y,Qian J,Tang Y,Shen N
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Oct 01 00:00:00 EDT 2017
Human amnion cells reverse acute and chronic pulmonary damage in experimental neonatal lung injury.
Stem cell research & therapy
Zhu D,Tan J,Maleken AS,Muljadi R,Chan ST,Lau SN,Elgass K,Leaw B,Mockler J,Chambers D,Leeman KT,Kim CF,Wallace EM,Lim R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Nov 10 00:00:00 EST 2017
Recipient bone marrow assimilates the myeloid/lymphoid reconstitution of distinct fetal hematopoietic stem cells.
Oncotarget
Guo XL,Chu L,Ke F,Mu LL,Li Z,Cai JJ,Li HF,Hong DL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Dec 12 00:00:00 EST 2017
Berberine augments ATP-induced inflammasome activation in macrophages by enhancing AMPK signaling.
Oncotarget
Li CG,Yan L,Jing YY,Xu LH,Liang YD,Wei HX,Hu B,Pan H,Zha QB,Ouyang DY,He XH
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jan 03 00:00:00 EST 2017
Loss of epidermal AP1 transcription factor function reduces filaggrin level, alters chemokine expression and produces an ichthyosis-related phenotype.
Cell death & disease
Young CA,Rorke EA,Adhikary G,Xu W,Eckert RL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Neutrophil swarming and extracellular trap formation play a significant role in Alum adjuvant activity.
NPJ vaccines
Stephen J,Scales HE,Benson RA,Erben D,Garside P,Brewer JM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Apr 08 00:00:00 EDT 2022
In vivo quantification of quantum dot systemic transport in C57BL/6 hairless mice following skin application post-ultraviolet radiation.
Particle and fibre toxicology
Jatana S,Palmer BC,Phelan SJ,Gelein R,DeLouise LA
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Apr 14 00:00:00 EDT 2017
Experimental lupus is aggravated in mouse strains with impaired induction of neutrophil extracellular traps.
JCI insight
Kienhöfer D,Hahn J,Stoof J,Csepregi JZ,Reinwald C,Urbonaviciute V,Johnsson C,Maueröder C,Podolska MJ,Biermann MH,Leppkes M,Harrer T,Hultqvist M,Olofsson P,Munoz LE,Mocsai A,Herrmann M,Schett G,Holmdahl R,Hoffmann MH
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu May 18 00:00:00 EDT 2017
Regulation of endothelial intracellular adenosine via adenosine kinase epigenetically modulates vascular inflammation.
Nature communications
Xu Y,Wang Y,Yan S,Yang Q,Zhou Y,Zeng X,Liu Z,An X,Toque HA,Dong Z,Jiang X,Fulton DJ,Weintraub NL,Li Q,Bagi Z,Hong M,Boison D,Wu C,Huo Y
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Oct 16 00:00:00 EDT 2017
CIITA-driven MHC class II expressing tumor cells can efficiently prime naive CD4+ TH cells in vivo and vaccinate the host against parental MHC-II-negative tumor cells.
Oncoimmunology
Bou Nasser Eddine F,Forlani G,Lombardo L,Tedeschi A,Tosi G,Accolla RS
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Combination immunotherapy with TLR agonists and checkpoint inhibitors suppresses head and neck cancer.
JCI insight
Sato-Kaneko F,Yao S,Ahmadi A,Zhang SS,Hosoya T,Kaneda MM,Varner JA,Pu M,Messer KS,Guiducci C,Coffman RL,Kitaura K,Matsutani T,Suzuki R,Carson DA,Hayashi T,Cohen EE
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 21 00:00:00 EDT 2017
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Oct 06 00:00:00 EDT 2016
The Deubiquitinase OTULIN Is an Essential Negative Regulator of Inflammation and Autoimmunity.
Cell
Damgaard RB,Walker JA,Marco-Casanova P,Morgan NV,Titheradge HL,Elliott PR,McHale D,Maher ER,McKenzie ANJ,Komander D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Aug 25 00:00:00 EDT 2016
Classical dendritic cells are required for dietary antigen-mediated induction of peripheral T(reg) cells and tolerance.
Nature immunology
Esterházy D,Loschko J,London M,Jove V,Oliveira TY,Mucida D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
Oligodendrocyte death results in immune-mediated CNS demyelination.
Nature neuroscience
Traka M,Podojil JR,McCarthy DP,Miller SD,Popko B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
The breast tumor microenvironment alters the phenotype and function of natural killer cells.
Cellular & molecular immunology
Krneta T,Gillgrass A,Chew M,Ashkar AA
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2016
Alternatively activated macrophages determine repair of the infarcted adult murine heart.
The Journal of clinical investigation
Shiraishi M,Shintani Y,Shintani Y,Ishida H,Saba R,Yamaguchi A,Adachi H,Yashiro K,Suzuki K
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
ATP-Induced Inflammasome Activation and Pyroptosis Is Regulated by AMP-Activated Protein Kinase in Macrophages.
Frontiers in immunology
Zha QB,Wei HX,Li CG,Liang YD,Xu LH,Bai WJ,Pan H,He XH,Ouyang DY
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Opposite Effects of Coinjection and Distant Injection of Mesenchymal Stem Cells on Breast Tumor Cell Growth.
Stem cells translational medicine
Zheng H,Zou W,Shen J,Xu L,Wang S,Fu YX,Fan W
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2016
Macrophages transfer antigens to dendritic cells by releasing exosomes containing dead-cell-associated antigens partially through a ceramide-dependent pathway to enhance CD4(+) T-cell responses.
Immunology
Xu Y,Liu Y,Yang C,Kang L,Wang M,Hu J,He H,Song W,Tang H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2016
Ginsenoside Rd alleviates mouse acute renal ischemia/reperfusion injury by modulating macrophage phenotype.
Journal of ginseng research
Ren K,Jin C,Ma P,Ren Q,Jia Z,Zhu D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Apr 01 00:00:00 EDT 2016
Multimodal analysis of ocular inflammation using the endotoxin-induced uveitis mouse model.
Disease models & mechanisms
Chu CJ,Gardner PJ,Copland DA,Liyanage SE,Gonzalez-Cordero A,Kleine Holthaus SM,Luhmann UF,Smith AJ,Ali RR,Dick AD
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Apr 01 00:00:00 EDT 2016
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2016
Effects of ketanserin on experimental colitis in mice and macrophage function.
International journal of molecular medicine
Xiao J,Shao L,Shen J,Jiang W,Feng Y,Zheng P,Liu F
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
ECM-Dependence of Endothelial Progenitor Cell Features.
Journal of cellular biochemistry
Siavashi V,Nassiri SM,Rahbarghazi R,Vafaei R,Sariri R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Sertoli cell condition medium can induce germ like cells from bone marrow derived mesenchymal stem cells.
Iranian journal of basic medical sciences
Monfared MH,Minaee B,Rastegar T,Khrazinejad E,Barbarestani M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Activated c-Kit receptor in the heart promotes cardiac repair and regeneration after injury.
Cell death & disease
Di Siena S,Gimmelli R,Nori SL,Barbagallo F,Campolo F,Dolci S,Rossi P,Venneri MA,Giannetta E,Gianfrilli D,Feigenbaum L,Lenzi A,Naro F,Cianflone E,Mancuso T,Torella D,Isidori AM,Pellegrini M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jul 28 00:00:00 EDT 2016
Ranitidine modifies myeloid cell populations and inhibits breast tumor development and spread in mice.
Oncoimmunology
Vila-Leahey A,Oldford SA,Marignani PA,Wang J,Haidl ID,Marshall JS
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
Interleukin 6 regulates psoriasiform inflammation-associated thrombosis.
JCI insight
Wang Y,Golden JB,Fritz Y,Zhang X,Diaconu D,Camhi MI,Gao H,Dawes SM,Xing X,Ganesh SK,Gudjonsson JE,Simon DI,McCormick TS,Ward NL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Dec 08 00:00:00 EST 2016
Stroke sensitivity in the aged: sex chromosome complement vs. gonadal hormones.
Aging
McCullough LD,Mirza MA,Xu Y,Bentivegna K,Steffens EB,Ritzel R,Liu F
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
NK cell development requires Tsc1-dependent negative regulation of IL-15-triggered mTORC1 activation.
Nature communications
Yang M,Chen S,Du J,He J,Wang Y,Li Z,Liu G,Peng W,Zeng X,Li D,Xu P,Guo W,Chang Z,Wang S,Tian Z,Dong Z
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Sep 07 00:00:00 EDT 2016
The impact of ranitidine on monocyte responses in the context of solid tumors.
Oncotarget
Vila-Leahey A,Rogers D,Marshall JS
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Mar 08 00:00:00 EST 2016
Impaired differentiation of macrophage lineage cells attenuates bone remodeling and inflammatory angiogenesis in Ndrg1 deficient mice.
Scientific reports
Watari K,Shibata T,Nabeshima H,Shinoda A,Fukunaga Y,Kawahara A,Karasuyama K,Fukushi J,Iwamoto Y,Kuwano M,Ono M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jan 18 00:00:00 EST 2016
Macrophagic control of the response to uropathogenic E. coli infection by regulation of iron retention in an IL-6-dependent manner.
Immunity, inflammation and disease
Owusu-Boaitey N,Bauckman KA,Zhang T,Mysorekar IU
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Agonist anti-GITR monoclonal antibody and stereotactic radiation induce immune-mediated survival advantage in murine intracranial glioma.
Journal for immunotherapy of cancer
Patel MA,Kim JE,Theodros D,Tam A,Velarde E,Kochel CM,Francica B,Nirschl TR,Ghasemzadeh A,Mathios D,Harris-Bookman S,Jackson CC,Jackson C,Ye X,Tran PT,Tyler B,Coric V,Selby M,Brem H,Drake CG,Pardoll DM,Lim M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed May 18 00:00:00 EDT 2016
Irradiation of necrotic cancer cells, employed for pulsing dendritic cells (DCs), potentiates DC vaccine-induced antitumor immunity against high-grade glioma.
Oncoimmunology
Vandenberk L,Garg AD,Verschuere T,Koks C,Belmans J,Beullens M,Agostinis P,De Vleeschouwer S,Van Gool SW
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Diet-dependent, microbiota-independent regulation of IL-10-producing lamina propria macrophages in the small intestine.
Scientific reports
Ochi T,Feng Y,Kitamoto S,Nagao-Kitamoto H,Kuffa P,Atarashi K,Honda K,Teitelbaum DH,Kamada N
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jun 15 00:00:00 EDT 2016
KLF4-dependent phenotypic modulation of smooth muscle cells has a key role in atherosclerotic plaque pathogenesis.
Nature medicine
Shankman LS,Gomez D,Cherepanova OA,Salmon M,Alencar GF,Haskins RM,Swiatlowska P,Newman AA,Greene ES,Straub AC,Isakson B,Randolph GJ,Owens GK
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Host microbiota constantly control maturation and function of microglia in the CNS.
Nature neuroscience
Erny D,Hrabě de Angelis AL,Jaitin D,Wieghofer P,Staszewski O,David E,Keren-Shaul H,Mahlakoiv T,Jakobshagen K,Buch T,Schwierzeck V,Utermöhlen O,Chun E,Garrett WS,McCoy KD,Diefenbach A,Staeheli P,Stecher B,Amit I,Prinz M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Functional-genetic dissection of HDAC dependencies in mouse lymphoid and myeloid malignancies.
Blood
Matthews GM,Mehdipour P,Cluse LA,Falkenberg KJ,Wang E,Roth M,Santoro F,Vidacs E,Stanley K,House CM,Rusche JR,Vakoc CR,Zuber J,Minucci S,Johnstone RW
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Nov 19 00:00:00 EST 2015
Apoptotic cells trigger a membrane-initiated pathway to increase ABCA1.
The Journal of clinical investigation
Fond AM,Lee CS,Schulman IG,Kiss RS,Ravichandran KS
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
MicroRNA-223 is a crucial mediator of PPARγ-regulated alternative macrophage activation.
The Journal of clinical investigation
Ying W,Tseng A,Chang RC,Morin A,Brehm T,Triff K,Nair V,Zhuang G,Song H,Kanameni S,Wang H,Golding MC,Bazer FW,Chapkin RS,Safe S,Zhou B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Nov 02 00:00:00 EST 2015
Aging-associated inflammation promotes selection for adaptive oncogenic events in B cell progenitors.
The Journal of clinical investigation
Henry CJ,Casás-Selves M,Kim J,Zaberezhnyy V,Aghili L,Daniel AE,Jimenez L,Azam T,McNamee EN,Clambey ET,Klawitter J,Serkova NJ,Tan AC,Dinarello CA,DeGregori J
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
STK4 regulates TLR pathways and protects against chronic inflammation-related hepatocellular carcinoma.
The Journal of clinical investigation
Li W,Xiao J,Zhou X,Xu M,Hu C,Xu X,Lu Y,Liu C,Xue S,Nie L,Zhang H,Li Z,Zhang Y,Ji F,Hui L,Tao W,Wei B,Wang H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Nov 02 00:00:00 EST 2015
Retinoid X receptor activation reverses age-related deficiencies in myelin debris phagocytosis and remyelination.
Brain : a journal of neurology
Natrajan MS,de la Fuente AG,Crawford AH,Linehan E,Nuñez V,Johnson KR,Wu T,Fitzgerald DC,Ricote M,Bielekova B,Franklin RJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
Tamm-Horsfall Protein Regulates Granulopoiesis and Systemic Neutrophil Homeostasis.
Journal of the American Society of Nephrology : JASN
Micanovic R,Chitteti BR,Dagher PC,Srour EF,Khan S,Hato T,Lyle A,Tong Y,Wu XR,El-Achkar TM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Cis-regulatory mechanisms governing stem and progenitor cell transitions.
Science advances
Johnson KD,Kong G,Gao X,Chang YI,Hewitt KJ,Sanalkumar R,Prathibha R,Ranheim EA,Dewey CN,Zhang J,Bresnick EH
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Identification of a chronic non-neurodegenerative microglia activation state in a mouse model of peroxisomal β-oxidation deficiency.
Glia
Verheijden S,Beckers L,Casazza A,Butovsky O,Mazzone M,Baes M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Aging-like changes in the transcriptome of irradiated microglia.
Glia
Li MD,Burns TC,Kumar S,Morgan AA,Sloan SA,Palmer TD
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2015
Local delivery of VEGF and SDF enhances endothelial progenitor cell recruitment and resultant recovery from ischemia.
Tissue engineering. Part A
Anderson EM,Kwee BJ,Lewin SA,Raimondo T,Mehta M,Mooney DJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Reducing progression of experimental lupus nephritis via inhibition of the B7/CD28 signaling pathway.
Molecular medicine reports
Huang L,Kong Y,Wang J,Sun J,Shi Q,Qiu YH
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Targeted Mutation of Nuclear Bone Morphogenetic Protein 2 Impairs Secondary Immune Response in a Mouse Model.
BioMed research international
Olsen DS,Goar WA,Nichols BA,Bailey KT,Christensen SL,Merriam KR,Reynolds PR,Wilson E,Weber KS,Bridgewater LC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Injectable cryogel-based whole-cell cancer vaccines.
Nature communications
Bencherif SA,Warren Sands R,Ali OA,Li WA,Lewin SA,Braschler TM,Shih TY,Verbeke CS,Bhatta D,Dranoff G,Mooney DJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Aug 12 00:00:00 EDT 2015
Granulocyte/Macrophage Colony-stimulating Factor-dependent Dendritic Cells Restrain Lean Adipose Tissue Expansion.
The Journal of biological chemistry
Pamir N,Liu NC,Irwin A,Becker L,Peng Y,Ronsein GE,Bornfeldt KE,Duffield JS,Heinecke JW
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jun 05 00:00:00 EDT 2015
The HIF-1/glial TIM-3 axis controls inflammation-associated brain damage under hypoxia.
Nature communications
Koh HS,Chang CY,Jeon SB,Yoon HJ,Ahn YH,Kim HS,Kim IH,Jeon SH,Johnson RS,Park EJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Mar 20 00:00:00 EDT 2015
Cancer-associated ASXL1 mutations may act as gain-of-function mutations of the ASXL1-BAP1 complex.
Nature communications
Balasubramani A,Larjo A,Bassein JA,Chang X,Hastie RB,Togher SM,Lähdesmäki H,Rao A
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jun 22 00:00:00 EDT 2015
Macrophages Contribute to the Spermatogonial Niche in the Adult Testis.
Cell reports
DeFalco T,Potter SJ,Williams AV,Waller B,Kan MJ,Capel B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Aug 18 00:00:00 EDT 2015
Glioma-associated microglia/macrophages display an expression profile different from M1 and M2 polarization and highly express Gpnmb and Spp1.
PloS one
Szulzewsky F,Pelz A,Feng X,Synowitz M,Markovic D,Langmann T,Holtman IR,Wang X,Eggen BJ,Boddeke HW,Hambardzumyan D,Wolf SA,Kettenmann H
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2016
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Sep 15 00:00:00 EDT 2015
Targeted Gene Correction in Osteopetrotic-Induced Pluripotent Stem Cells for the Generation of Functional Osteoclasts.
Stem cell reports
Neri T,Muggeo S,Paulis M,Caldana ME,Crisafulli L,Strina D,Focarelli ML,Faggioli F,Recordati C,Scaramuzza S,Scanziani E,Mantero S,Buracchi C,Sobacchi C,Lombardo A,Naldini L,Vezzoni P,Villa A,Ficara F
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Oct 13 00:00:00 EDT 2015
Pathogenic Yersinia Promotes Its Survival by Creating an Acidic Fluid-Accessible Compartment on the Macrophage Surface.
PloS one
Bahnan W,Boettner DR,Westermark L,Fällman M,Schesser K
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue May 10 00:00:00 EDT 2016
Differential Immunometabolic Phenotype in Th1 and Th2 Dominant Mouse Strains in Response to High-Fat Feeding.
PloS one
Jovicic N,Jeftic I,Jovanovic I,Radosavljevic G,Arsenijevic N,Lukic ML,Pejnovic N
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue May 03 00:00:00 EDT 2016
Sunitinib depletes myeloid-derived suppressor cells and synergizes with a cancer vaccine to enhance antigen-specific immune responses and tumor eradication.
Oncoimmunology
Draghiciu O,Nijman HW,Hoogeboom BN,Meijerhof T,Daemen T
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2015
Functional polarization of tumour-associated macrophages by tumour-derived lactic acid.
Nature
Colegio OR,Chu NQ,Szabo AL,Chu T,Rhebergen AM,Jairam V,Cyrus N,Brokowski CE,Eisenbarth SC,Phillips GM,Cline GW,Phillips AJ,Medzhitov R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 25 00:00:00 EDT 2014
Heme-mediated SPI-C induction promotes monocyte differentiation into iron-recycling macrophages.
Cell
Haldar M,Kohyama M,So AY,Kc W,Wu X,Briseño CG,Satpathy AT,Kretzer NM,Arase H,Rajasekaran NS,Wang L,Egawa T,Igarashi K,Baltimore D,Murphy TL,Murphy KM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Mar 13 00:00:00 EDT 2014
A role for the ITAM signaling module in specifying cytokine-receptor functions.
Nature immunology
Bezbradica JS,Rosenstein RK,DeMarco RA,Brodsky I,Medzhitov R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Apr 01 00:00:00 EDT 2014
RAGE expression in tumor-associated macrophages promotes angiogenesis in glioma.
Cancer research
Chen X,Zhang L,Zhang IY,Liang J,Wang H,Ouyang M,Wu S,da Fonseca ACC,Weng L,Yamamoto Y,Yamamoto H,Natarajan R,Badie B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Dec 15 00:00:00 EST 2014
Macrophages contribute to the cyclic activation of adult hair follicle stem cells.
PLoS biology
Castellana D,Paus R,Perez-Moreno M
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Dec 01 00:00:00 EST 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Aug 12 00:00:00 EDT 2014
Astrocytic transforming growth factor-beta signaling reduces subacute neuroinflammation after stroke in mice.
Glia
Cekanaviciute E,Fathali N,Doyle KP,Williams AM,Han J,Buckwalter MS
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
A crucial role for the ubiquitously expressed transcription factor Sp1 at early stages of hematopoietic specification.
Development (Cambridge, England)
Gilmour J,Assi SA,Jaegle U,Kulu D,van de Werken H,Clarke D,Westhead DR,Philipsen S,Bonifer C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
Distinct spatial distribution of microglia and macrophages following mesenchymal stem cell implantation in mouse brain.
Immunology and cell biology
Le Blon D,Hoornaert C,Daans J,Santermans E,Hens N,Goossens H,Berneman Z,Ponsaerts P
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Tracking the elusive fibrocyte: identification and characterization of collagen-producing hematopoietic lineage cells during murine wound healing.
Stem cells (Dayton, Ohio)
Suga H,Rennert RC,Rodrigues M,Sorkin M,Glotzbach JP,Januszyk M,Fujiwara T,Longaker MT,Gurtner GC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu May 01 00:00:00 EDT 2014
Tryptophan catabolism restricts IFN-γ-expressing neutrophils and Clostridium difficile immunopathology.
Journal of immunology (Baltimore, Md. : 1950)
El-Zaatari M,Chang YM,Zhang M,Franz M,Shreiner A,McDermott AJ,van der Sluijs KF,Lutter R,Grasberger H,Kamada N,Young VB,Huffnagle GB,Kao JY
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Jul 15 00:00:00 EDT 2014
Increased expression of stress inducible protein 1 in glioma-associated microglia/macrophages.
Journal of neuroimmunology
Carvalho da Fonseca AC,Wang H,Fan H,Chen X,Zhang I,Zhang L,Lima FR,Badie B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Sep 15 00:00:00 EDT 2014
Metastasis is regulated via microRNA-200/ZEB1 axis control of tumour cell PD-L1 expression and intratumoral immunosuppression.
Nature communications
Chen L,Gibbons DL,Goswami S,Cortez MA,Ahn YH,Byers LA,Zhang X,Yi X,Dwyer D,Lin W,Diao L,Wang J,Roybal J,Patel M,Ungewiss C,Peng D,Antonia S,Mediavilla-Varela M,Robertson G,Suraokar M,Welsh JW,Erez B,Wistuba II,Chen L,Peng D,Wang S,Ullrich SE,Heymach JV,Kurie JM,Qin FX
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Oct 28 00:00:00 EDT 2014
Amniotic fluid stem cells prevent follicle atresia and rescue fertility of mice with premature ovarian failure induced by chemotherapy.
PloS one
Xiao GY,Liu IH,Cheng CC,Chang CC,Lee YH,Cheng WT,Wu SC
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu May 28 00:00:00 EDT 2015
Anti-tumor innate immunity activated by intermittent metronomic cyclophosphamide treatment of 9L brain tumor xenografts is preserved by anti-angiogenic drugs that spare VEGF receptor 2.
Molecular cancer
Doloff JC,Chen CS,Waxman DJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jun 26 00:00:00 EDT 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jun 10 00:00:00 EDT 2015
P2X7 receptor modulates inflammatory and functional pulmonary changes induced by silica.
PloS one
Monção-Ribeiro LC,Faffe DS,Santana PT,Vieira FS,da Graça CL,Marques-da-Silva C,Machado MN,Caruso-Neves C,Zin WA,Borojevic R,Takiya CM,Coutinho-Silva R
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2015
The nucleotide-binding leucine-rich repeat (NLR) family member NLRX1 mediates protection against experimental autoimmune encephalomyelitis and represses macrophage/microglia-induced inflammation.
The Journal of biological chemistry
Eitas TK,Chou WC,Wen H,Gris D,Robbins GR,Brickey J,Oyama Y,Ting JP
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Feb 14 00:00:00 EST 2014
Selective regulation of lymphopoiesis and leukemogenesis by individual zinc fingers of Ikaros.
Nature immunology
Schjerven H,McLaughlin J,Arenzana TL,Frietze S,Cheng D,Wadsworth SE,Lawson GW,Bensinger SJ,Farnham PJ,Witte ON,Smale ST
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
S100B promotes glioma growth through chemoattraction of myeloid-derived macrophages.
Clinical cancer research : an official journal of the American Association for Cancer Research
Wang H,Zhang L,Zhang IY,Chen X,Da Fonseca A,Wu S,Ren H,Badie S,Sadeghi S,Ouyang M,Warden CD,Badie B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Jul 15 00:00:00 EDT 2013
A network of interactions enables CCM3 and STK24 to coordinate UNC13D-driven vesicle exocytosis in neutrophils.
Developmental cell
Zhang Y,Tang W,Zhang H,Niu X,Xu Y,Zhang J,Gao K,Pan W,Boggon TJ,Toomre D,Min W,Wu D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Oct 28 00:00:00 EDT 2013
Arrayed lentiviral barcoding for quantification analysis of hematopoietic dynamics.
Stem cells (Dayton, Ohio)
Grosselin J,Sii-Felice K,Payen E,Chretien S,Tronik-Le Roux D,Leboulch P
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
Migration of bone marrow-derived cells into the central nervous system in models of neurodegeneration.
The Journal of comparative neurology
Lampron A,Pimentel-Coelho PM,Rivest S
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2013
Myxomaviral Anti-Inflammatory Serpin Reduces Myeloid-Derived Suppressor Cells and Human Pancreatic Cancer Cell Growth in Mice.
Journal of cancer science & therapy
Zheng D,Chen H,Bartee MY,Williams J,Davids JA,Lomas DA,McFadden G,Lucas AR
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Aug 19 00:00:00 EDT 2013
The cardioprotective protein apolipoprotein A1 promotes potent anti-tumorigenic effects.
The Journal of biological chemistry
Zamanian-Daryoush M,Lindner D,Tallant TC,Wang Z,Buffa J,Klipfell E,Parker Y,Hatala D,Parsons-Wingerter P,Rayman P,Yusufishaq MSS,Fisher EA,Smith JD,Finke J,DiDonato JA,Hazen SL
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jul 19 00:00:00 EDT 2013
Identification of the low density lipoprotein (LDL) receptor-related protein-1 interactome in central nervous system myelin suggests a role in the clearance of necrotic cell debris.
The Journal of biological chemistry
Fernandez-Castaneda A,Arandjelovic S,Stiles TL,Schlobach RK,Mowen KA,Gonias SL,Gaultier A
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Feb 15 00:00:00 EST 2013
Endothelial cells provide an instructive niche for the differentiation and functional polarization of M2-like macrophages.
Blood
He H,Xu J,Warren CM,Duan D,Li X,Wu L,Iruela-Arispe ML
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Oct 11 00:00:00 EDT 2012
Eltrombopag inhibits the proliferation of leukemia cells via reduction of intracellular iron and induction of differentiation.
Blood
Roth M,Will B,Simkin G,Narayanagari S,Barreyro L,Bartholdy B,Tamari R,Mitsiades CS,Verma A,Steidl U
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Jul 12 00:00:00 EDT 2012
Cis-element mutated in GATA2-dependent immunodeficiency governs hematopoiesis and vascular integrity.
The Journal of clinical investigation
Johnson KD,Hsu AP,Ryu MJ,Wang J,Gao X,Boyer ME,Liu Y,Lee Y,Calvo KR,Keles S,Zhang J,Holland SM,Bresnick EH
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2012
Induction of histiocytic sarcoma in mouse skeletal muscle.
PloS one
Liu J,Hettmer S,Milsom MD,Hofmann I,Hua F,Miller C,Bronson RT,Wagers AJ
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Thu Feb 07 00:00:00 EST 2013
Cell-selective inhibition of NF-κB signaling improves therapeutic index in a melanoma chemotherapy model.
Cancer discovery
Enzler T,Sano Y,Choo MK,Cottam HB,Karin M,Tsao H,Park JM
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2011
Sensitized phenotypic screening identifies gene dosage sensitive region on chromosome 11 that predisposes to disease in mice.
EMBO molecular medicine
Ermakova O,Piszczek L,Luciani L,Cavalli FM,Ferreira T,Farley D,Rizzo S,Paolicelli RC,Al-Banchaabouchi M,Nerlov C,Moriggl R,Luscombe NM,Gross C
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2011
A tritherapy combination of a fusion protein vaccine with immune-modulating doses of sequential chemotherapies in an optimized regimen completely eradicates large tumors in mice.
International journal of cancer
Song X,Guo W,Cui J,Qian X,Yi L,Chang M,Cai Q,Zhao Q
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2011
PAD4-mediated neutrophil extracellular trap formation is not required for immunity against influenza infection.
PloS one
Hemmers S,Teijaro JR,Arandjelovic S,Mowen KA
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Nov 15 00:00:00 EST 2011
Wnt7a activates the planar cell polarity pathway to drive the symmetric expansion of satellite stem cells.
Cell stem cell
Le Grand F,Jones AE,Seale V,Scimè A,Rudnicki MA
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Jun 05 00:00:00 EDT 2009
Combination of all-trans retinoic acid and a human papillomavirus therapeutic vaccine suppresses the number and function of immature myeloid cells and enhances antitumor immunity.
Cancer science
Song X,Ye D,Liu B,Cui J,Zhao X,Yi L,Liang J,Song J,Zhang Z,Zhao Q
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Feb 01 00:00:00 EST 2009
Cellular expression and antimicrobial function of a phylogenetically conserved novel histone 1x-like protein on mouse cells: a potential new class of pattern recognition receptor.
Journal of leukocyte biology
Evans DL,Connor MA,Moss LD,Lackay S,Leary JH,Krunkosky T,Jaso-Friedmann L
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2009
Region-specific expression of immunoregulatory proteins on microglia in the healthy CNS.
Glia
de Haas AH,Boddeke HW,Biber K
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2008
Phospholipase Cgamma2 mediates RANKL-stimulated lymph node organogenesis and osteoclastogenesis.
The Journal of biological chemistry
Chen Y,Wang X,Di L,Fu G,Chen Y,Bai L,Liu J,Feng X,McDonald JM,Michalek S,He Y,Yu M,Fu YX,Wen R,Wu H,Wang D
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Fri Oct 24 00:00:00 EDT 2008
Optimized isolation enables ex vivo analysis of microglia from various central nervous system regions.
Glia
de Haas AH,Boddeke HW,Brouwer N,Biber K
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2007
Dendritic cells purified from myeloma are primed with tumor-specific antigen (idiotype) and activate CD4+ T cells.
Proceedings of the National Academy of Sciences of the United States of America
Dembic Z,Schenck K,Bogen B
Published figure using CD11b monoclonal antibody (Product # 46-0112-82) in Flow Cytometry
Tue Mar 14 00:00:00 EST 2000